Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 129389488-129390058 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHCYL2
Entrez Gene ID
23382
Gene Name
adenosylhomocysteinase like 2
Gene Aliases
ADOHCYASE3, IRBIT2
UniGene
Hs.600789
Gene Chromosome Location
Chr.7: 129225014-129430211 on Build GRCh38
Transcripts
NM_001130720.2 NM_001130722.2 NM_001130723.2 NM_015328.3 XM_005250237.1 XM_011515987.2 XM_011515988.2 XM_017011904.1 XM_017011905.1 XM_017011906.1 XM_017011907.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3698167 c.399G>C p.L133F Chr.7: 129389656-129389656 on Build GRCh38
3698165 c.642G>C p.L214F Chr.7: 129389656-129389656 on Build GRCh38
4359665 c.445G>A p.V149M Chr.7: 129389702-129389702 on Build GRCh38
3878037 c.452G>A p.C151Y Chr.7: 129389709-129389709 on Build GRCh38
2769735 c.379G>A p.V127M Chr.7: 129389702-129389702 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148283331 Chr.7: 129389528-129389528 on Build GRCh38
rs768535161 Chr.7: 129389613-129389613 on Build GRCh38
rs949765470 Chr.7: 129389817-129389817 on Build GRCh38
rs916897428 Chr.7: 129389793-129389793 on Build GRCh38
rs1022831215 Chr.7: 129389939-129389939 on Build GRCh38
rs962817816 Chr.7: 129389645-129389645 on Build GRCh38
rs955609340 Chr.7: 129390002-129390002 on Build GRCh38
rs748186443 Chr.7: 129389594-129389596 on Build GRCh38
rs956150676 Chr.7: 129389545-129389545 on Build GRCh38
rs968062945 Chr.7: 129389940-129389941 on Build GRCh38
rs753024167 Chr.7: 129389784-129389784 on Build GRCh38
rs1541711 Chr.7: 129389550-129389550 on Build GRCh38
rs1002626075 Chr.7: 129389900-129389900 on Build GRCh38
rs934125594 Chr.7: 129389701-129389701 on Build GRCh38
rs766911111 Chr.7: 129389738-129389738 on Build GRCh38
rs561749176 Chr.7: 129389578-129389578 on Build GRCh38
rs754302285 Chr.7: 129389748-129389748 on Build GRCh38
rs1029694285 Chr.7: 129389559-129389559 on Build GRCh38
rs528477771 Chr.7: 129389564-129389564 on Build GRCh38
rs568475618 Chr.7: 129389725-129389725 on Build GRCh38
rs186304826 Chr.7: 129389846-129389846 on Build GRCh38
rs111680903 Chr.7: 129390018-129390018 on Build GRCh38
rs572759814 Chr.7: 129389913-129389913 on Build GRCh38
rs765638905 Chr.7: 129389755-129389755 on Build GRCh38
rs535622217 Chr.7: 129389754-129389754 on Build GRCh38
rs924164923 Chr.7: 129389983-129389983 on Build GRCh38
rs376055240 Chr.7: 129389737-129389737 on Build GRCh38
rs920336788 Chr.7: 129389705-129389705 on Build GRCh38
rs141148420 Chr.7: 129389571-129389571 on Build GRCh38
rs955373741 Chr.7: 129389925-129389925 on Build GRCh38
rs891874395 Chr.7: 129389865-129389865 on Build GRCh38
rs369017192 Chr.7: 129389818-129389818 on Build GRCh38
rs771827228 Chr.7: 129389773-129389774 on Build GRCh38
rs539865201 Chr.7: 129389933-129389933 on Build GRCh38
rs749102491 Chr.7: 129389597-129389597 on Build GRCh38
rs1033057614 Chr.7: 129389538-129389538 on Build GRCh38
rs1051280385 Chr.7: 129389722-129389722 on Build GRCh38
rs1010345075 Chr.7: 129389895-129389895 on Build GRCh38
rs774740816 Chr.7: 129389638-129389638 on Build GRCh38
rs117913215 Chr.7: 129389562-129389562 on Build GRCh38
rs555413952 Chr.7: 129390028-129390028 on Build GRCh38
rs779785208 Chr.7: 129389585-129389585 on Build GRCh38
rs994260022 Chr.7: 129389829-129389829 on Build GRCh38
rs191546470 Chr.7: 129389855-129389855 on Build GRCh38
rs905779496 Chr.7: 129389841-129389841 on Build GRCh38
rs978508986 Chr.7: 129389944-129389944 on Build GRCh38
rs747135290 Chr.7: 129389901-129389901 on Build GRCh38
rs762179977 Chr.7: 129389650-129389650 on Build GRCh38
rs371323706 Chr.7: 129389785-129389785 on Build GRCh38
rs767604280 Chr.7: 129389659-129389659 on Build GRCh38
rs113620628 Chr.7: 129390017-129390017 on Build GRCh38
rs1054326093 Chr.7: 129389859-129389859 on Build GRCh38
rs974250879 Chr.7: 129389702-129389702 on Build GRCh38

View Full Product Details