Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 89431566-89432147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC16A12
Entrez Gene ID
387700
Gene Name
solute carrier family 16 member 12
Gene Aliases
CJMG, CRT2, MCT12
UniGene
Hs.530338
Gene Chromosome Location
Chr.10: 89430294-89536028 on Build GRCh38
Transcripts
NM_213606.3 XM_017016237.1 XM_017016238.1 XM_017016239.1

SNP Detail

SNP ID Chromosome Location
rs1023917975 Chr.10: 89431894-89431894 on Build GRCh38
rs773401477 Chr.10: 89431674-89431674 on Build GRCh38
rs903897526 Chr.10: 89432045-89432045 on Build GRCh38
rs532774325 Chr.10: 89431993-89431993 on Build GRCh38
rs983231164 Chr.10: 89431994-89431994 on Build GRCh38
rs907621239 Chr.10: 89431996-89431996 on Build GRCh38
rs999727909 Chr.10: 89431654-89431654 on Build GRCh38
rs756312822 Chr.10: 89432015-89432015 on Build GRCh38
rs187148527 Chr.10: 89431689-89431689 on Build GRCh38
rs903941298 Chr.10: 89431617-89431617 on Build GRCh38
rs7342121 Chr.10: 89432002-89432002 on Build GRCh38
rs576759554 Chr.10: 89431802-89431802 on Build GRCh38
rs749436298 Chr.10: 89431759-89431759 on Build GRCh38
rs146415721 Chr.10: 89431624-89431624 on Build GRCh38
rs1014764051 Chr.10: 89431907-89431907 on Build GRCh38
rs74146950 Chr.10: 89432001-89432001 on Build GRCh38
rs1028683706 Chr.10: 89431781-89431781 on Build GRCh38
rs535569682 Chr.10: 89432094-89432094 on Build GRCh38
rs550042853 Chr.10: 89431923-89431923 on Build GRCh38
rs996206008 Chr.10: 89431590-89431590 on Build GRCh38
rs1055185759 Chr.10: 89431587-89431587 on Build GRCh38
rs1031036874 Chr.10: 89431733-89431733 on Build GRCh38
rs773914112 Chr.10: 89432010-89432010 on Build GRCh38
rs770929947 Chr.10: 89431818-89431818 on Build GRCh38
rs1013689127 Chr.10: 89431869-89431869 on Build GRCh38
rs570714544 Chr.10: 89431695-89431695 on Build GRCh38
rs868063151 Chr.10: 89431928-89431928 on Build GRCh38
rs935332123 Chr.10: 89432095-89432095 on Build GRCh38
rs946845488 Chr.10: 89431599-89431599 on Build GRCh38
rs1043741834 Chr.10: 89432025-89432025 on Build GRCh38
rs142580584 Chr.10: 89431606-89431606 on Build GRCh38
rs140843919 Chr.10: 89431914-89431914 on Build GRCh38
rs970014086 Chr.10: 89431908-89431908 on Build GRCh38
rs1050506450 Chr.10: 89432106-89432106 on Build GRCh38
rs921892354 Chr.10: 89432012-89432012 on Build GRCh38
rs951876444 Chr.10: 89431874-89431874 on Build GRCh38
rs902721480 Chr.10: 89431610-89431610 on Build GRCh38
rs991209231 Chr.10: 89431977-89431977 on Build GRCh38
rs960716457 Chr.10: 89431925-89431925 on Build GRCh38
rs565096673 Chr.10: 89431716-89431716 on Build GRCh38
rs1006008540 Chr.10: 89431889-89431889 on Build GRCh38
rs868191680 Chr.10: 89432019-89432019 on Build GRCh38

View Full Product Details