Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 32629815-32630376 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYNJ1
Entrez Gene ID
8867
Gene Name
synaptojanin 1
Gene Aliases
INPP5G, PARK20
UniGene
Hs.473632
Gene Chromosome Location
Chr.21: 32628759-32728128 on Build GRCh38
Transcripts
NM_001160302.1 NM_001160306.1 NM_003895.3 NM_203446.2 XM_017028494.1 XM_017028495.1 XM_017028496.1 XM_017028497.1 XM_017028498.1 XM_017028499.1 XM_017028500.1 XM_017028501.1 XM_017028502.1 XM_017028503.1 XM_017028504.1 XM_017028505.1

SNP Detail

SNP ID Chromosome Location
rs955406523 Chr.21: 32630013-32630013 on Build GRCh38
rs577256755 Chr.21: 32630295-32630295 on Build GRCh38
rs1018072586 Chr.21: 32630041-32630041 on Build GRCh38
rs971505293 Chr.21: 32629843-32629843 on Build GRCh38
rs147590143 Chr.21: 32630070-32630070 on Build GRCh38
rs117085153 Chr.21: 32629916-32629916 on Build GRCh38
rs1034871752 Chr.21: 32630108-32630108 on Build GRCh38
rs1043584870 Chr.21: 32629975-32629975 on Build GRCh38
rs541111549 Chr.21: 32630301-32630301 on Build GRCh38
rs920856797 Chr.21: 32629875-32629875 on Build GRCh38
rs374027671 Chr.21: 32629844-32629844 on Build GRCh38
rs1043719838 Chr.21: 32630098-32630098 on Build GRCh38
rs990965883 Chr.21: 32630258-32630258 on Build GRCh38
rs756991215 Chr.21: 32630110-32630110 on Build GRCh38
rs979507206 Chr.21: 32630294-32630294 on Build GRCh38
rs549227725 Chr.21: 32629917-32629917 on Build GRCh38
rs1010868914 Chr.21: 32629953-32629953 on Build GRCh38
rs1029481849 Chr.21: 32630010-32630010 on Build GRCh38
rs916664919 Chr.21: 32630081-32630081 on Build GRCh38
rs902457269 Chr.21: 32629977-32629977 on Build GRCh38
rs531384486 Chr.21: 32630276-32630276 on Build GRCh38
rs113590478 Chr.21: 32630208-32630208 on Build GRCh38
rs1031790483 Chr.21: 32630342-32630346 on Build GRCh38
rs751220190 Chr.21: 32630076-32630076 on Build GRCh38
rs927750031 Chr.21: 32629862-32629862 on Build GRCh38
rs1027887646 Chr.21: 32630054-32630054 on Build GRCh38
rs1040526801 Chr.21: 32629958-32629958 on Build GRCh38
rs995142869 Chr.21: 32630198-32630198 on Build GRCh38
rs973739762 Chr.21: 32630053-32630053 on Build GRCh38
rs984138233 Chr.21: 32630058-32630058 on Build GRCh38
rs983719746 Chr.21: 32630230-32630230 on Build GRCh38
rs895141708 Chr.21: 32629947-32629947 on Build GRCh38
rs572436751 Chr.21: 32630116-32630116 on Build GRCh38
rs916635139 Chr.21: 32630267-32630267 on Build GRCh38
rs538298535 Chr.21: 32630068-32630068 on Build GRCh38
rs781689047 Chr.21: 32630055-32630055 on Build GRCh38
rs1023507092 Chr.21: 32629837-32629838 on Build GRCh38
rs1006637683 Chr.21: 32630032-32630032 on Build GRCh38
rs1050076581 Chr.21: 32629918-32629918 on Build GRCh38
rs960865409 Chr.21: 32630254-32630254 on Build GRCh38
rs576203373 Chr.21: 32630213-32630213 on Build GRCh38
rs777138831 Chr.21: 32629902-32629902 on Build GRCh38
rs935362454 Chr.21: 32630347-32630347 on Build GRCh38
rs549581750 Chr.21: 32630048-32630048 on Build GRCh38
rs762510102 Chr.21: 32630057-32630057 on Build GRCh38
rs543902312 Chr.21: 32630222-32630222 on Build GRCh38
rs956498873 Chr.21: 32630111-32630111 on Build GRCh38
rs1017999444 Chr.21: 32630153-32630153 on Build GRCh38
rs1051491428 Chr.21: 32630112-32630112 on Build GRCh38
rs897958019 Chr.21: 32630031-32630031 on Build GRCh38
rs752150443 Chr.21: 32629903-32629903 on Build GRCh38
rs192520528 Chr.21: 32629923-32629923 on Build GRCh38
rs536533392 Chr.21: 32630127-32630127 on Build GRCh38
rs915167432 Chr.21: 32630135-32630135 on Build GRCh38
rs1007145450 Chr.21: 32630152-32630152 on Build GRCh38
rs565192434 Chr.21: 32630282-32630282 on Build GRCh38
rs114913358 Chr.21: 32629845-32629845 on Build GRCh38
rs929503028 Chr.21: 32630220-32630220 on Build GRCh38
rs367580525 Chr.21: 32629855-32629855 on Build GRCh38
rs757738370 Chr.21: 32629964-32629964 on Build GRCh38
rs902550029 Chr.21: 32630107-32630107 on Build GRCh38
rs116092770 Chr.21: 32630049-32630049 on Build GRCh38
rs923938949 Chr.21: 32630340-32630340 on Build GRCh38
rs946694935 Chr.21: 32630088-32630088 on Build GRCh38
rs537953447 Chr.21: 32629930-32629930 on Build GRCh38
rs999736300 Chr.21: 32630000-32630000 on Build GRCh38
rs919438461 Chr.21: 32630214-32630214 on Build GRCh38
rs763733765 Chr.21: 32630073-32630073 on Build GRCh38
rs1002972482 Chr.21: 32630328-32630328 on Build GRCh38
rs928137108 Chr.21: 32630061-32630061 on Build GRCh38
rs759982775 Chr.21: 32629910-32629910 on Build GRCh38
rs1035687941 Chr.21: 32630231-32630231 on Build GRCh38
rs971481470 Chr.21: 32630086-32630086 on Build GRCh38
rs554758438 Chr.21: 32630166-32630166 on Build GRCh38

View Full Product Details