Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 45183243-45183799 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUP50
Entrez Gene ID
10762
Gene Name
nucleoporin 50
Gene Aliases
NPAP60, NPAP60L
UniGene
Hs.475103
Gene Chromosome Location
Chr.22: 45163845-45188011 on Build GRCh38
Transcripts
NM_007172.3 NM_153645.2 XM_005261312.1 XM_005261314.1 XM_006724103.1 XM_006724104.1 XM_011529833.1 XM_017028537.1 XM_017028538.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3713747 c.1192G>C p.D398H Chr.22: 45183508-45183508 on Build GRCh38
4791142 c.1148T>C p.L383S Chr.22: 45183464-45183464 on Build GRCh38
1034998 c.1186C>T p.R396W Chr.22: 45183502-45183502 on Build GRCh38
1644844 c.1176G>C p.Q392H Chr.22: 45183492-45183492 on Build GRCh38
1644843 c.1177C>T p.L393F Chr.22: 45183493-45183493 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779183174 Chr.22: 45183565-45183565 on Build GRCh38
rs146749329 Chr.22: 45183486-45183486 on Build GRCh38
rs373726931 Chr.22: 45183515-45183515 on Build GRCh38
rs1040596441 Chr.22: 45183534-45183534 on Build GRCh38
rs746869312 Chr.22: 45183488-45183488 on Build GRCh38
rs953130818 Chr.22: 45183763-45183763 on Build GRCh38
rs767507661 Chr.22: 45183602-45183603 on Build GRCh38
rs896719685 Chr.22: 45183720-45183720 on Build GRCh38
rs974687607 Chr.22: 45183289-45183289 on Build GRCh38
rs6006950 Chr.22: 45183541-45183541 on Build GRCh38
rs956401910 Chr.22: 45183747-45183747 on Build GRCh38
rs372228306 Chr.22: 45183554-45183554 on Build GRCh38
rs774630842 Chr.22: 45183703-45183703 on Build GRCh38
rs941951593 Chr.22: 45183644-45183647 on Build GRCh38
rs760925721 Chr.22: 45183385-45183385 on Build GRCh38
rs758885314 Chr.22: 45183567-45183567 on Build GRCh38
rs779493727 Chr.22: 45183470-45183470 on Build GRCh38
rs923221059 Chr.22: 45183291-45183291 on Build GRCh38
rs975875202 Chr.22: 45183616-45183616 on Build GRCh38
rs991716703 Chr.22: 45183414-45183414 on Build GRCh38
rs376810757 Chr.22: 45183566-45183566 on Build GRCh38
rs752396802 Chr.22: 45183448-45183448 on Build GRCh38
rs750276372 Chr.22: 45183409-45183409 on Build GRCh38
rs368107021 Chr.22: 45183651-45183654 on Build GRCh38
rs781294855 Chr.22: 45183548-45183548 on Build GRCh38
rs113286908 Chr.22: 45183511-45183511 on Build GRCh38
rs778353508 Chr.22: 45183357-45183357 on Build GRCh38
rs932961519 Chr.22: 45183331-45183331 on Build GRCh38
rs769727564 Chr.22: 45183562-45183562 on Build GRCh38
rs368029667 Chr.22: 45183361-45183361 on Build GRCh38
rs1050337659 Chr.22: 45183542-45183542 on Build GRCh38
rs759601125 Chr.22: 45183616-45183617 on Build GRCh38
rs772951553 Chr.22: 45183540-45183540 on Build GRCh38
rs752789409 Chr.22: 45183600-45183602 on Build GRCh38
rs150983434 Chr.22: 45183756-45183756 on Build GRCh38
rs533961795 Chr.22: 45183695-45183698 on Build GRCh38
rs780610633 Chr.22: 45183436-45183436 on Build GRCh38
rs985957959 Chr.22: 45183773-45183773 on Build GRCh38
rs759815351 Chr.22: 45183562-45183563 on Build GRCh38
rs776812227 Chr.22: 45183580-45183580 on Build GRCh38
rs535057659 Chr.22: 45183620-45183620 on Build GRCh38
rs561636003 Chr.22: 45183304-45183304 on Build GRCh38
rs759461413 Chr.22: 45183549-45183549 on Build GRCh38
rs59420220 Chr.22: 45183658-45183664 on Build GRCh38
rs142735960 Chr.22: 45183601-45183601 on Build GRCh38
rs760280714 Chr.22: 45183617-45183617 on Build GRCh38
rs1006271665 Chr.22: 45183359-45183359 on Build GRCh38
rs61743071 Chr.22: 45183490-45183490 on Build GRCh38
rs778229170 Chr.22: 45183457-45183457 on Build GRCh38
rs780051036 Chr.22: 45183572-45183572 on Build GRCh38
rs999075679 Chr.22: 45183724-45183724 on Build GRCh38
rs1013803516 Chr.22: 45183737-45183737 on Build GRCh38
rs763082785 Chr.22: 45183601-45183602 on Build GRCh38
rs61740782 Chr.22: 45183411-45183411 on Build GRCh38
rs536965670 Chr.22: 45183549-45183551 on Build GRCh38
rs761495720 Chr.22: 45183759-45183761 on Build GRCh38
rs757616478 Chr.22: 45183563-45183563 on Build GRCh38
rs747724982 Chr.22: 45183267-45183267 on Build GRCh38
rs745897715 Chr.22: 45183560-45183560 on Build GRCh38
rs6007493 Chr.22: 45183346-45183346 on Build GRCh38
rs536041634 Chr.22: 45183598-45183598 on Build GRCh38
rs1021771039 Chr.22: 45183706-45183706 on Build GRCh38
rs766137434 Chr.22: 45183544-45183544 on Build GRCh38
rs533049374 Chr.22: 45183503-45183503 on Build GRCh38
rs774224974 Chr.22: 45183375-45183375 on Build GRCh38
rs377483891 Chr.22: 45183407-45183407 on Build GRCh38
rs909127690 Chr.22: 45183643-45183643 on Build GRCh38
rs762811449 Chr.22: 45183425-45183425 on Build GRCh38
rs774075071 Chr.22: 45183595-45183595 on Build GRCh38
rs758119099 Chr.22: 45183284-45183284 on Build GRCh38
rs777855653 Chr.22: 45183323-45183323 on Build GRCh38
rs1055244023 Chr.22: 45183681-45183681 on Build GRCh38
rs547367260 Chr.22: 45183378-45183378 on Build GRCh38
rs191619998 Chr.22: 45183279-45183279 on Build GRCh38
rs1036404123 Chr.22: 45183559-45183559 on Build GRCh38
rs777596315 Chr.22: 45183561-45183561 on Build GRCh38
rs897841355 Chr.22: 45183657-45183657 on Build GRCh38
rs775684635 Chr.22: 45183575-45183575 on Build GRCh38
rs765453469 Chr.22: 45183564-45183565 on Build GRCh38
rs566370858 Chr.22: 45183602-45183602 on Build GRCh38
rs775665771 Chr.22: 45183382-45183382 on Build GRCh38
rs759463284 Chr.22: 45183597-45183597 on Build GRCh38
rs1057223558 Chr.22: 45183666-45183666 on Build GRCh38
rs768986222 Chr.22: 45183626-45183626 on Build GRCh38
rs374015864 Chr.22: 45183355-45183355 on Build GRCh38
rs750985816 Chr.22: 45183428-45183428 on Build GRCh38
rs749669968 Chr.22: 45183459-45183459 on Build GRCh38
rs778586429 Chr.22: 45183264-45183264 on Build GRCh38
rs1035887960 Chr.22: 45183396-45183396 on Build GRCh38
rs777418734 Chr.22: 45183376-45183376 on Build GRCh38
rs923658961 Chr.22: 45183615-45183615 on Build GRCh38
rs755891777 Chr.22: 45183452-45183452 on Build GRCh38
rs748361021 Chr.22: 45183591-45183591 on Build GRCh38
rs1042811665 Chr.22: 45183622-45183622 on Build GRCh38
rs571505558 Chr.22: 45183665-45183665 on Build GRCh38
rs938180675 Chr.22: 45183654-45183654 on Build GRCh38
rs771298463 Chr.22: 45183469-45183469 on Build GRCh38
rs75474767 Chr.22: 45183577-45183577 on Build GRCh38
rs768879297 Chr.22: 45183538-45183538 on Build GRCh38
rs143503455 Chr.22: 45183430-45183430 on Build GRCh38
rs752272433 Chr.22: 45183558-45183558 on Build GRCh38
rs765086886 Chr.22: 45183360-45183360 on Build GRCh38
rs770757402 Chr.22: 45183593-45183593 on Build GRCh38
rs1013034885 Chr.22: 45183687-45183687 on Build GRCh38
rs766373082 Chr.22: 45183426-45183426 on Build GRCh38
rs2075952 Chr.22: 45183621-45183621 on Build GRCh38
rs771132625 Chr.22: 45183418-45183421 on Build GRCh38
rs768817951 Chr.22: 45183579-45183579 on Build GRCh38
rs968928664 Chr.22: 45183715-45183715 on Build GRCh38
rs112466118 Chr.22: 45183280-45183280 on Build GRCh38
rs975014258 Chr.22: 45183589-45183589 on Build GRCh38
rs760872582 Chr.22: 45183521-45183521 on Build GRCh38
rs1016469927 Chr.22: 45183574-45183574 on Build GRCh38

View Full Product Details