Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 40769544-40770123 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A17
Entrez Gene ID
10478
Gene Name
solute carrier family 25 member 17
Gene Aliases
PMP34
UniGene
Hs.474938
Gene Chromosome Location
Chr.22: 40769630-40819399 on Build GRCh38
Transcripts
NM_001282726.1 NM_001282727.1 NM_006358.3 NR_104235.1 NR_104236.1 NR_104237.1 NR_104238.1

SNP Detail

SNP ID Chromosome Location
rs769542336 Chr.22: 40769565-40769565 on Build GRCh38
rs754370869 Chr.22: 40769913-40769913 on Build GRCh38
rs997339809 Chr.22: 40769612-40769612 on Build GRCh38
rs922664414 Chr.22: 40769919-40769919 on Build GRCh38
rs185814758 Chr.22: 40769737-40769737 on Build GRCh38
rs745362484 Chr.22: 40769981-40769981 on Build GRCh38
rs551912505 Chr.22: 40769848-40769848 on Build GRCh38
rs544211687 Chr.22: 40769656-40769656 on Build GRCh38
rs1050125120 Chr.22: 40769589-40769589 on Build GRCh38
rs904471791 Chr.22: 40770046-40770046 on Build GRCh38
rs569140363 Chr.22: 40770045-40770045 on Build GRCh38
rs564564368 Chr.22: 40769637-40769637 on Build GRCh38
rs992937914 Chr.22: 40769819-40769819 on Build GRCh38
rs756939407 Chr.22: 40769882-40769882 on Build GRCh38
rs1007899323 Chr.22: 40769702-40769702 on Build GRCh38
rs1037116172 Chr.22: 40769585-40769585 on Build GRCh38
rs960205741 Chr.22: 40769736-40769736 on Build GRCh38
rs969477991 Chr.22: 40769683-40769683 on Build GRCh38
rs1014550678 Chr.22: 40770089-40770089 on Build GRCh38
rs117012345 Chr.22: 40769791-40769791 on Build GRCh38
rs970302137 Chr.22: 40769856-40769856 on Build GRCh38
rs191113447 Chr.22: 40769774-40769774 on Build GRCh38
rs41281281 Chr.22: 40769674-40769674 on Build GRCh38
rs555099719 Chr.22: 40770077-40770077 on Build GRCh38
rs12158059 Chr.22: 40769942-40769942 on Build GRCh38
rs1050488519 Chr.22: 40769879-40769879 on Build GRCh38
rs960748591 Chr.22: 40770081-40770081 on Build GRCh38
rs959810516 Chr.22: 40769763-40769763 on Build GRCh38
rs907678464 Chr.22: 40769849-40769849 on Build GRCh38
rs767636122 Chr.22: 40769686-40769686 on Build GRCh38
rs116005203 Chr.22: 40770076-40770076 on Build GRCh38
rs569661968 Chr.22: 40770022-40770022 on Build GRCh38
rs745814544 Chr.22: 40769913-40769914 on Build GRCh38
rs550979135 Chr.22: 40769943-40769943 on Build GRCh38
rs1010906414 Chr.22: 40769660-40769660 on Build GRCh38
rs774617099 Chr.22: 40769621-40769621 on Build GRCh38
rs1021254652 Chr.22: 40769703-40769703 on Build GRCh38
rs984783131 Chr.22: 40769866-40769866 on Build GRCh38
rs1053001887 Chr.22: 40769632-40769632 on Build GRCh38
rs940080986 Chr.22: 40769578-40769578 on Build GRCh38
rs901293940 Chr.22: 40769599-40769599 on Build GRCh38
rs367858950 Chr.22: 40769890-40769890 on Build GRCh38
rs890682232 Chr.22: 40769611-40769611 on Build GRCh38
rs751305662 Chr.22: 40769749-40769749 on Build GRCh38
rs560146230 Chr.22: 40770064-40770064 on Build GRCh38
rs992771980 Chr.22: 40769771-40769771 on Build GRCh38
rs892759136 Chr.22: 40769990-40769990 on Build GRCh38
rs920335992 Chr.22: 40769862-40769862 on Build GRCh38
rs940009871 Chr.22: 40769908-40769908 on Build GRCh38
rs931772136 Chr.22: 40769873-40769873 on Build GRCh38
rs757630538 Chr.22: 40769991-40769991 on Build GRCh38
rs961878339 Chr.22: 40769855-40769855 on Build GRCh38
rs1031321753 Chr.22: 40770091-40770091 on Build GRCh38
rs752855012 Chr.22: 40769739-40769739 on Build GRCh38
rs533291032 Chr.22: 40769899-40769899 on Build GRCh38
rs1031374148 Chr.22: 40769744-40769744 on Build GRCh38
rs375343274 Chr.22: 40770102-40770102 on Build GRCh38
rs909882355 Chr.22: 40769572-40769572 on Build GRCh38
rs904187516 Chr.22: 40769707-40769707 on Build GRCh38
rs527486344 Chr.22: 40769804-40769804 on Build GRCh38

View Full Product Details