Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 29131048-29131625 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLIP4
Entrez Gene ID
79745
Gene Name
CAP-Gly domain containing linker protein family member 4
Gene Aliases
RSNL2
UniGene
Hs.122927
Gene Chromosome Location
Chr.2: 29097676-29183813 on Build GRCh38
Transcripts
NM_001287527.1 NM_001287528.1 NM_024692.5 NR_109844.1 XM_005264562.2 XM_005264563.1 XM_006712104.2 XM_006712105.3 XM_011533109.1 XM_011533110.1 XM_017004958.1 XM_017004959.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1201395 c.148G>T p.D50Y Chr.2: 29131272-29131272 on Build GRCh38
5862403 c.194C>T p.T65I Chr.2: 29131318-29131318 on Build GRCh38
6885595 c.243C>T p.V81V Chr.2: 29131367-29131367 on Build GRCh38
5598151 c.233T>C p.V78A Chr.2: 29131357-29131357 on Build GRCh38
3910205 c.140C>T p.S47F Chr.2: 29131264-29131264 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1017695133 Chr.2: 29131139-29131139 on Build GRCh38
rs746264181 Chr.2: 29131489-29131489 on Build GRCh38
rs755957985 Chr.2: 29131296-29131296 on Build GRCh38
rs201143374 Chr.2: 29131287-29131287 on Build GRCh38
rs780931043 Chr.2: 29131244-29131244 on Build GRCh38
rs544328850 Chr.2: 29131203-29131203 on Build GRCh38
rs1007187457 Chr.2: 29131139-29131139 on Build GRCh38
rs768136303 Chr.2: 29131425-29131425 on Build GRCh38
rs904230341 Chr.2: 29131140-29131140 on Build GRCh38
rs140458378 Chr.2: 29131383-29131383 on Build GRCh38
rs569538610 Chr.2: 29131601-29131601 on Build GRCh38
rs759660473 Chr.2: 29131390-29131390 on Build GRCh38
rs963397152 Chr.2: 29131348-29131348 on Build GRCh38
rs549053519 Chr.2: 29131473-29131473 on Build GRCh38
rs773384173 Chr.2: 29131378-29131378 on Build GRCh38
rs757080798 Chr.2: 29131319-29131319 on Build GRCh38
rs891563193 Chr.2: 29131446-29131446 on Build GRCh38
rs940222139 Chr.2: 29131152-29131152 on Build GRCh38
rs775334422 Chr.2: 29131346-29131346 on Build GRCh38
rs1028562400 Chr.2: 29131169-29131169 on Build GRCh38
rs762311023 Chr.2: 29131213-29131213 on Build GRCh38
rs758406789 Chr.2: 29131382-29131382 on Build GRCh38
rs969958541 Chr.2: 29131470-29131470 on Build GRCh38
rs752893012 Chr.2: 29131292-29131292 on Build GRCh38
rs537800795 Chr.2: 29131545-29131545 on Build GRCh38
rs377578157 Chr.2: 29131177-29131177 on Build GRCh38
rs78332192 Chr.2: 29131147-29131148 on Build GRCh38
rs775487928 Chr.2: 29131448-29131448 on Build GRCh38
rs369245161 Chr.2: 29131416-29131416 on Build GRCh38
rs770227864 Chr.2: 29131373-29131373 on Build GRCh38
rs1011377100 Chr.2: 29131259-29131259 on Build GRCh38
rs141091030 Chr.2: 29131343-29131343 on Build GRCh38
rs938386575 Chr.2: 29131543-29131543 on Build GRCh38
rs763331094 Chr.2: 29131244-29131244 on Build GRCh38
rs201325033 Chr.2: 29131387-29131387 on Build GRCh38
rs928194787 Chr.2: 29131490-29131490 on Build GRCh38
rs79643436 Chr.2: 29131071-29131071 on Build GRCh38
rs906909345 Chr.2: 29131170-29131170 on Build GRCh38
rs749848482 Chr.2: 29131436-29131437 on Build GRCh38
rs372696264 Chr.2: 29131266-29131266 on Build GRCh38
rs201841032 Chr.2: 29131139-29131140 on Build GRCh38
rs777389312 Chr.2: 29131311-29131311 on Build GRCh38
rs751580548 Chr.2: 29131268-29131268 on Build GRCh38
rs973419804 Chr.2: 29131527-29131527 on Build GRCh38
rs999897515 Chr.2: 29131140-29131140 on Build GRCh38
rs537924532 Chr.2: 29131376-29131376 on Build GRCh38
rs371657198 Chr.2: 29131404-29131404 on Build GRCh38
rs778977081 Chr.2: 29131321-29131321 on Build GRCh38
rs763530959 Chr.2: 29131381-29131381 on Build GRCh38
rs192903361 Chr.2: 29131084-29131084 on Build GRCh38
rs188032968 Chr.2: 29131412-29131412 on Build GRCh38
rs767535341 Chr.2: 29131394-29131394 on Build GRCh38
rs781585661 Chr.2: 29131481-29131481 on Build GRCh38
rs746596896 Chr.2: 29131356-29131356 on Build GRCh38
rs1014057140 Chr.2: 29131459-29131460 on Build GRCh38
rs1055579838 Chr.2: 29131208-29131208 on Build GRCh38
rs767766211 Chr.2: 29131291-29131291 on Build GRCh38
rs752946214 Chr.2: 29131400-29131400 on Build GRCh38
rs775281038 Chr.2: 29131209-29131209 on Build GRCh38
rs982421680 Chr.2: 29131483-29131483 on Build GRCh38
rs903109262 Chr.2: 29131299-29131299 on Build GRCh38
rs749654045 Chr.2: 29131436-29131436 on Build GRCh38
rs567295356 Chr.2: 29131480-29131480 on Build GRCh38
rs760791990 Chr.2: 29131449-29131449 on Build GRCh38
rs980079265 Chr.2: 29131211-29131211 on Build GRCh38
rs759993710 Chr.2: 29131439-29131439 on Build GRCh38
rs915667389 Chr.2: 29131561-29131561 on Build GRCh38
rs750964537 Chr.2: 29131233-29131233 on Build GRCh38
rs192977173 Chr.2: 29131592-29131592 on Build GRCh38
rs548019804 Chr.2: 29131182-29131182 on Build GRCh38
rs961537293 Chr.2: 29131464-29131464 on Build GRCh38
rs749249736 Chr.2: 29131313-29131313 on Build GRCh38
rs963345395 Chr.2: 29131515-29131515 on Build GRCh38
rs776334545 Chr.2: 29131431-29131431 on Build GRCh38
rs185158020 Chr.2: 29131100-29131100 on Build GRCh38
rs779673429 Chr.2: 29131413-29131413 on Build GRCh38
rs148556850 Chr.2: 29131159-29131159 on Build GRCh38
rs748483674 Chr.2: 29131243-29131244 on Build GRCh38
rs757074289 Chr.2: 29131409-29131409 on Build GRCh38
rs745442308 Chr.2: 29131341-29131341 on Build GRCh38
rs202154099 Chr.2: 29131438-29131438 on Build GRCh38
rs756220997 Chr.2: 29131402-29131402 on Build GRCh38
rs943609009 Chr.2: 29131430-29131430 on Build GRCh38
rs908808642 Chr.2: 29131120-29131120 on Build GRCh38
rs767586402 Chr.2: 29131444-29131444 on Build GRCh38
rs765499141 Chr.2: 29131224-29131224 on Build GRCh38
rs912043637 Chr.2: 29131598-29131598 on Build GRCh38
rs142900518 Chr.2: 29131191-29131191 on Build GRCh38
rs1052772418 Chr.2: 29131445-29131445 on Build GRCh38
rs770154345 Chr.2: 29131250-29131251 on Build GRCh38

View Full Product Details