Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 86450852-86451338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDM3A
Entrez Gene ID
55818
Gene Name
lysine demethylase 3A
Gene Aliases
JHDM2A, JHMD2A, JMJD1, JMJD1A, TSGA
UniGene
Hs.557425
Gene Chromosome Location
Chr.2: 86440647-86492716 on Build GRCh38
Transcripts
NM_001146688.1 NM_018433.5 XM_006712051.3 XM_017004493.1 XM_017004494.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2823023 c.395G>A p.R132H Chr.2: 86451155-86451155 on Build GRCh38
3739962 c.420A>G p.V140V Chr.2: 86451180-86451180 on Build GRCh38
6586019 c.386C>T p.T129I Chr.2: 86451146-86451146 on Build GRCh38
6092465 c.448A>G p.I150V Chr.2: 86451208-86451208 on Build GRCh38
1307066 c.376G>A p.G126R Chr.2: 86451136-86451136 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776406459 Chr.2: 86451072-86451073 on Build GRCh38
rs567922063 Chr.2: 86450908-86450908 on Build GRCh38
rs761895936 Chr.2: 86451221-86451222 on Build GRCh38
rs769800555 Chr.2: 86451222-86451222 on Build GRCh38
rs766523661 Chr.2: 86451252-86451252 on Build GRCh38
rs375225586 Chr.2: 86451084-86451084 on Build GRCh38
rs933920310 Chr.2: 86450910-86450910 on Build GRCh38
rs534948590 Chr.2: 86451224-86451224 on Build GRCh38
rs34173194 Chr.2: 86450936-86450937 on Build GRCh38
rs149635248 Chr.2: 86451112-86451112 on Build GRCh38
rs908653641 Chr.2: 86450929-86450929 on Build GRCh38
rs757822227 Chr.2: 86451190-86451190 on Build GRCh38
rs976030119 Chr.2: 86450885-86450885 on Build GRCh38
rs1009838839 Chr.2: 86451002-86451002 on Build GRCh38
rs745590738 Chr.2: 86451210-86451210 on Build GRCh38
rs139162735 Chr.2: 86451040-86451040 on Build GRCh38
rs550700879 Chr.2: 86451056-86451056 on Build GRCh38
rs777518063 Chr.2: 86451197-86451197 on Build GRCh38
rs552842449 Chr.2: 86450904-86450905 on Build GRCh38
rs756448328 Chr.2: 86451081-86451081 on Build GRCh38
rs937946367 Chr.2: 86450978-86450978 on Build GRCh38
rs201247647 Chr.2: 86451198-86451198 on Build GRCh38
rs746865942 Chr.2: 86451105-86451105 on Build GRCh38
rs773544304 Chr.2: 86451245-86451245 on Build GRCh38
rs761752202 Chr.2: 86451141-86451141 on Build GRCh38
rs771785147 Chr.2: 86451101-86451101 on Build GRCh38
rs930699627 Chr.2: 86450959-86450959 on Build GRCh38
rs903655470 Chr.2: 86451260-86451260 on Build GRCh38
rs112007276 Chr.2: 86451304-86451305 on Build GRCh38
rs377098522 Chr.2: 86451074-86451074 on Build GRCh38
rs764264350 Chr.2: 86451062-86451062 on Build GRCh38
rs192577683 Chr.2: 86450971-86450971 on Build GRCh38
rs377634057 Chr.2: 86450970-86450970 on Build GRCh38
rs183597287 Chr.2: 86451059-86451059 on Build GRCh38
rs922801609 Chr.2: 86450895-86450895 on Build GRCh38
rs879559523 Chr.2: 86451151-86451151 on Build GRCh38
rs1031099817 Chr.2: 86451037-86451037 on Build GRCh38
rs749795762 Chr.2: 86451086-86451086 on Build GRCh38
rs767596150 Chr.2: 86451073-86451073 on Build GRCh38
rs747547707 Chr.2: 86451100-86451100 on Build GRCh38
rs779252326 Chr.2: 86451173-86451173 on Build GRCh38
rs761993122 Chr.2: 86451229-86451229 on Build GRCh38
rs774492469 Chr.2: 86451127-86451127 on Build GRCh38
rs370493777 Chr.2: 86451255-86451255 on Build GRCh38
rs1039149099 Chr.2: 86450953-86450953 on Build GRCh38
rs757513396 Chr.2: 86451067-86451067 on Build GRCh38
rs553179654 Chr.2: 86451253-86451253 on Build GRCh38
rs752271195 Chr.2: 86451174-86451174 on Build GRCh38
rs982942987 Chr.2: 86450923-86450923 on Build GRCh38
rs977836509 Chr.2: 86451006-86451006 on Build GRCh38
rs57145635 Chr.2: 86451001-86451001 on Build GRCh38
rs766438413 Chr.2: 86451052-86451052 on Build GRCh38
rs376727383 Chr.2: 86451064-86451064 on Build GRCh38
rs754544342 Chr.2: 86451090-86451090 on Build GRCh38
rs754148984 Chr.2: 86451155-86451155 on Build GRCh38
rs1038394822 Chr.2: 86450956-86450956 on Build GRCh38
rs1012466189 Chr.2: 86451243-86451243 on Build GRCh38
rs770906961 Chr.2: 86451200-86451200 on Build GRCh38
rs978160695 Chr.2: 86450902-86450902 on Build GRCh38
rs750579969 Chr.2: 86451149-86451149 on Build GRCh38
rs760904934 Chr.2: 86451247-86451247 on Build GRCh38
rs772689138 Chr.2: 86451104-86451104 on Build GRCh38
rs895338537 Chr.2: 86450986-86450986 on Build GRCh38
rs778500623 Chr.2: 86451096-86451096 on Build GRCh38
rs942519911 Chr.2: 86450926-86450926 on Build GRCh38
rs560700053 Chr.2: 86450952-86450952 on Build GRCh38
rs772042108 Chr.2: 86451239-86451239 on Build GRCh38
rs766719966 Chr.2: 86451154-86451154 on Build GRCh38
rs1003808634 Chr.2: 86450980-86450980 on Build GRCh38
rs939315040 Chr.2: 86451188-86451188 on Build GRCh38
rs941414700 Chr.2: 86450938-86450938 on Build GRCh38
rs536146514 Chr.2: 86450982-86450982 on Build GRCh38
rs147119313 Chr.2: 86451110-86451110 on Build GRCh38
rs1042219684 Chr.2: 86451256-86451256 on Build GRCh38
rs781147888 Chr.2: 86451201-86451201 on Build GRCh38
rs755483346 Chr.2: 86451168-86451168 on Build GRCh38
rs955761781 Chr.2: 86450911-86450911 on Build GRCh38
rs767831132 Chr.2: 86451142-86451142 on Build GRCh38
rs756538332 Chr.2: 86451150-86451150 on Build GRCh38
rs895391110 Chr.2: 86451208-86451208 on Build GRCh38
rs746047990 Chr.2: 86451004-86451004 on Build GRCh38

View Full Product Details