Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 29704160-29704754 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TJP1
Entrez Gene ID
7082
Gene Name
tight junction protein 1
Gene Aliases
ZO-1
UniGene
Hs.743990
Gene Chromosome Location
Chr.15: 29699367-29968919 on Build GRCh38
Transcripts
NM_001301025.1 NM_001301026.1 NM_003257.4 NM_175610.3 XM_005254617.3 XM_005254618.3 XM_005254619.3 XM_005254620.3 XM_011521972.2 XM_017022521.1 XM_017022522.1 XM_017022523.1 XM_017022524.1 XM_017022525.1 XM_017022526.1 XM_017022527.1 XM_017022528.1 XM_017022529.1 XM_017022530.1 XM_017022531.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5027937 c.5152+9A>G p.? Chr.15: 29704213-29704213 on Build GRCh38
2083955 c.5114T>C p.L1705P Chr.15: 29704260-29704260 on Build GRCh38
6372148 c.5129C>T p.P1710L Chr.15: 29704245-29704245 on Build GRCh38
3815809 c.5140C>T p.R1714C Chr.15: 29704234-29704234 on Build GRCh38
4578156 c.5094G>A p.V1698V Chr.15: 29704280-29704280 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371086865 Chr.15: 29704313-29704313 on Build GRCh38
rs756391449 Chr.15: 29704218-29704218 on Build GRCh38
rs1046549941 Chr.15: 29704342-29704342 on Build GRCh38
rs762689099 Chr.15: 29704325-29704325 on Build GRCh38
rs1033755917 Chr.15: 29704588-29704588 on Build GRCh38
rs774282673 Chr.15: 29704195-29704198 on Build GRCh38
rs751476589 Chr.15: 29704201-29704201 on Build GRCh38
rs771212279 Chr.15: 29704256-29704256 on Build GRCh38
rs1003568377 Chr.15: 29704558-29704558 on Build GRCh38
rs932804789 Chr.15: 29704491-29704491 on Build GRCh38
rs755047396 Chr.15: 29704202-29704202 on Build GRCh38
rs192418249 Chr.15: 29704692-29704692 on Build GRCh38
rs45576137 Chr.15: 29704414-29704414 on Build GRCh38
rs781101609 Chr.15: 29704217-29704217 on Build GRCh38
rs755828352 Chr.15: 29704416-29704416 on Build GRCh38
rs532894295 Chr.15: 29704584-29704584 on Build GRCh38
rs187986522 Chr.15: 29704483-29704483 on Build GRCh38
rs141190320 Chr.15: 29704470-29704470 on Build GRCh38
rs894016555 Chr.15: 29704514-29704514 on Build GRCh38
rs368959239 Chr.15: 29704492-29704492 on Build GRCh38
rs952641459 Chr.15: 29704560-29704560 on Build GRCh38
rs751410981 Chr.15: 29704334-29704334 on Build GRCh38
rs749707467 Chr.15: 29704468-29704468 on Build GRCh38
rs906529725 Chr.15: 29704553-29704553 on Build GRCh38
rs778206679 Chr.15: 29704234-29704234 on Build GRCh38
rs376984007 Chr.15: 29704246-29704246 on Build GRCh38
rs952598037 Chr.15: 29704686-29704686 on Build GRCh38
rs375658941 Chr.15: 29704333-29704333 on Build GRCh38
rs958548013 Chr.15: 29704412-29704412 on Build GRCh38
rs1020967702 Chr.15: 29704377-29704377 on Build GRCh38
rs769373681 Chr.15: 29704283-29704283 on Build GRCh38
rs775995856 Chr.15: 29704271-29704271 on Build GRCh38
rs906580550 Chr.15: 29704384-29704384 on Build GRCh38
rs916383639 Chr.15: 29704327-29704327 on Build GRCh38
rs749154223 Chr.15: 29704507-29704507 on Build GRCh38
rs1012729648 Chr.15: 29704652-29704652 on Build GRCh38
rs566518117 Chr.15: 29704731-29704731 on Build GRCh38
rs780710986 Chr.15: 29704448-29704449 on Build GRCh38
rs772695948 Chr.15: 29704666-29704666 on Build GRCh38
rs767548547 Chr.15: 29704335-29704335 on Build GRCh38
rs759250913 Chr.15: 29704322-29704323 on Build GRCh38
rs748549037 Chr.15: 29704657-29704657 on Build GRCh38
rs889260038 Chr.15: 29704447-29704447 on Build GRCh38
rs969610002 Chr.15: 29704635-29704635 on Build GRCh38
rs76628456 Chr.15: 29704447-29704447 on Build GRCh38
rs183325366 Chr.15: 29704480-29704480 on Build GRCh38
rs531991532 Chr.15: 29704367-29704367 on Build GRCh38
rs773494285 Chr.15: 29704706-29704706 on Build GRCh38
rs191200000 Chr.15: 29704378-29704378 on Build GRCh38
rs750218949 Chr.15: 29704181-29704181 on Build GRCh38
rs952649890 Chr.15: 29704419-29704419 on Build GRCh38
rs867256826 Chr.15: 29704189-29704189 on Build GRCh38
rs575536449 Chr.15: 29704280-29704280 on Build GRCh38
rs1003620663 Chr.15: 29704398-29704398 on Build GRCh38
rs981913488 Chr.15: 29704678-29704678 on Build GRCh38
rs916434630 Chr.15: 29704588-29704588 on Build GRCh38
rs918494224 Chr.15: 29704263-29704263 on Build GRCh38
rs377677916 Chr.15: 29704518-29704518 on Build GRCh38
rs982374869 Chr.15: 29704434-29704434 on Build GRCh38
rs557401553 Chr.15: 29704403-29704403 on Build GRCh38
rs756825098 Chr.15: 29704679-29704679 on Build GRCh38
rs555595998 Chr.15: 29704188-29704188 on Build GRCh38
rs546140877 Chr.15: 29704429-29704429 on Build GRCh38
rs375021009 Chr.15: 29704485-29704485 on Build GRCh38
rs367707884 Chr.15: 29704191-29704191 on Build GRCh38
rs573198559 Chr.15: 29704448-29704448 on Build GRCh38
rs1052104283 Chr.15: 29704699-29704699 on Build GRCh38
rs752819863 Chr.15: 29704343-29704343 on Build GRCh38
rs943742341 Chr.15: 29704456-29704456 on Build GRCh38
rs779424333 Chr.15: 29704261-29704261 on Build GRCh38
rs1042694472 Chr.15: 29704530-29704530 on Build GRCh38
rs895234802 Chr.15: 29704590-29704590 on Build GRCh38
rs983920159 Chr.15: 29704323-29704323 on Build GRCh38
rs377130760 Chr.15: 29704268-29704268 on Build GRCh38
rs983977364 Chr.15: 29704566-29704566 on Build GRCh38

View Full Product Details