Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 82042373-82042924 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEX3B
Entrez Gene ID
84206
Gene Name
mex-3 RNA binding family member B
Gene Aliases
MEX-3B, RKHD3, RNF195
UniGene
Hs.104744
Gene Chromosome Location
Chr.15: 82041778-82046143 on Build GRCh38
Transcripts
NM_032246.4

SNP Detail

SNP ID Chromosome Location
rs185165233 Chr.15: 82042888-82042888 on Build GRCh38
rs914677159 Chr.15: 82042554-82042554 on Build GRCh38
rs770419495 Chr.15: 82042569-82042573 on Build GRCh38
rs957895843 Chr.15: 82042553-82042553 on Build GRCh38
rs532632543 Chr.15: 82042537-82042537 on Build GRCh38
rs1011935393 Chr.15: 82042673-82042673 on Build GRCh38
rs995660107 Chr.15: 82042449-82042449 on Build GRCh38
rs777446461 Chr.15: 82042566-82042566 on Build GRCh38
rs1047217815 Chr.15: 82042636-82042636 on Build GRCh38
rs371568442 Chr.15: 82042430-82042430 on Build GRCh38
rs1040006824 Chr.15: 82042593-82042593 on Build GRCh38
rs192995917 Chr.15: 82042804-82042804 on Build GRCh38
rs142471581 Chr.15: 82042668-82042668 on Build GRCh38
rs969320884 Chr.15: 82042537-82042544 on Build GRCh38
rs770365206 Chr.15: 82042905-82042905 on Build GRCh38
rs951777038 Chr.15: 82042470-82042470 on Build GRCh38
rs976433822 Chr.15: 82042803-82042803 on Build GRCh38
rs947453065 Chr.15: 82042885-82042885 on Build GRCh38
rs1020796455 Chr.15: 82042755-82042755 on Build GRCh38
rs563797399 Chr.15: 82042534-82042534 on Build GRCh38
rs905217515 Chr.15: 82042684-82042685 on Build GRCh38
rs1023324852 Chr.15: 82042464-82042464 on Build GRCh38
rs753701346 Chr.15: 82042568-82042569 on Build GRCh38
rs1041915498 Chr.15: 82042569-82042569 on Build GRCh38
rs1057370809 Chr.15: 82042657-82042657 on Build GRCh38
rs903310745 Chr.15: 82042608-82042608 on Build GRCh38
rs931988011 Chr.15: 82042543-82042543 on Build GRCh38
rs953393689 Chr.15: 82042828-82042828 on Build GRCh38
rs769314250 Chr.15: 82042439-82042439 on Build GRCh38
rs1039941180 Chr.15: 82042432-82042432 on Build GRCh38
rs866476516 Chr.15: 82042716-82042716 on Build GRCh38
rs954662654 Chr.15: 82042774-82042774 on Build GRCh38
rs1002148219 Chr.15: 82042698-82042698 on Build GRCh38
rs947422121 Chr.15: 82042567-82042567 on Build GRCh38
rs556455781 Chr.15: 82042784-82042785 on Build GRCh38
rs1014678553 Chr.15: 82042511-82042511 on Build GRCh38
rs781555283 Chr.15: 82042428-82042436 on Build GRCh38
rs887537631 Chr.15: 82042694-82042694 on Build GRCh38
rs538338874 Chr.15: 82042874-82042874 on Build GRCh38
rs553800007 Chr.15: 82042743-82042743 on Build GRCh38
rs764039437 Chr.15: 82042440-82042444 on Build GRCh38
rs976022711 Chr.15: 82042529-82042529 on Build GRCh38
rs1005887319 Chr.15: 82042732-82042732 on Build GRCh38
rs113690500 Chr.15: 82042827-82042827 on Build GRCh38
rs556062014 Chr.15: 82042795-82042795 on Build GRCh38
rs879556388 Chr.15: 82042595-82042595 on Build GRCh38
rs1014750684 Chr.15: 82042779-82042779 on Build GRCh38
rs913680991 Chr.15: 82042565-82042565 on Build GRCh38
rs914540859 Chr.15: 82042872-82042872 on Build GRCh38
rs536467770 Chr.15: 82042762-82042762 on Build GRCh38
rs567951753 Chr.15: 82042586-82042586 on Build GRCh38
rs996116092 Chr.15: 82042623-82042623 on Build GRCh38
rs986141697 Chr.15: 82042841-82042841 on Build GRCh38
rs535241205 Chr.15: 82042689-82042689 on Build GRCh38
rs751057756 Chr.15: 82042676-82042676 on Build GRCh38
rs187824971 Chr.15: 82042728-82042728 on Build GRCh38
rs879657752 Chr.15: 82042708-82042708 on Build GRCh38
rs957946658 Chr.15: 82042731-82042731 on Build GRCh38
rs986642302 Chr.15: 82042548-82042548 on Build GRCh38
rs960396824 Chr.15: 82042406-82042406 on Build GRCh38
rs961938986 Chr.15: 82042527-82042527 on Build GRCh38
rs35065650 Chr.15: 82042488-82042489 on Build GRCh38
rs566075022 Chr.15: 82042612-82042612 on Build GRCh38
rs938967985 Chr.15: 82042644-82042644 on Build GRCh38
rs530325987 Chr.15: 82042447-82042447 on Build GRCh38
rs1051919624 Chr.15: 82042567-82042568 on Build GRCh38
rs567413437 Chr.15: 82042753-82042753 on Build GRCh38
rs139663241 Chr.15: 82042488-82042488 on Build GRCh38
rs780818948 Chr.15: 82042765-82042765 on Build GRCh38
rs898676283 Chr.15: 82042442-82042447 on Build GRCh38
rs943038157 Chr.15: 82042803-82042804 on Build GRCh38
rs977499231 Chr.15: 82042891-82042891 on Build GRCh38

View Full Product Details