Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 49523083-49523528 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHF11
Entrez Gene ID
51131
Gene Name
PHD finger protein 11
Gene Aliases
APY, BCAP, IGEL, IGER, IGHER, NY-REN-34, NYREN34
UniGene
Hs.369039
Gene Chromosome Location
Chr.13: 49495610-49528992 on Build GRCh38
Transcripts
NM_001040443.2 NM_001040444.2 NR_135322.1 NR_135323.1 NM_001320727.1 NR_135324.1

Gene Symbol
SETDB2-PHF11
Entrez Gene ID
107303344
Gene Name
SETDB2-PHF11 readthrough
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.13: 49444293-49528992 on Build GRCh38
Transcripts
NM_001040443.2 NM_001040444.2 NR_135322.1 NR_135323.1 NM_001320727.1 NR_135324.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4047972 c.576C>T p.P192P Chr.13: 49523180-49523180 on Build GRCh38
7256530 c.575C>A p.P192H Chr.13: 49523179-49523179 on Build GRCh38
4789288 c.623A>G p.H208R Chr.13: 49523227-49523227 on Build GRCh38
4047973 c.619G>T p.E207* Chr.13: 49523223-49523223 on Build GRCh38
3813960 c.585G>C p.M195I Chr.13: 49523189-49523189 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1045241070 Chr.13: 49523380-49523380 on Build GRCh38
rs947802145 Chr.13: 49523362-49523362 on Build GRCh38
rs577296558 Chr.13: 49523259-49523259 on Build GRCh38
rs915082589 Chr.13: 49523146-49523146 on Build GRCh38
rs891089727 Chr.13: 49523502-49523502 on Build GRCh38
rs1049777627 Chr.13: 49523501-49523501 on Build GRCh38
rs752594648 Chr.13: 49523202-49523202 on Build GRCh38
rs970150960 Chr.13: 49523206-49523206 on Build GRCh38
rs748545420 Chr.13: 49523129-49523129 on Build GRCh38
rs574646784 Chr.13: 49523377-49523377 on Build GRCh38
rs756355580 Chr.13: 49523246-49523246 on Build GRCh38
rs751155910 Chr.13: 49523161-49523162 on Build GRCh38
rs79165934 Chr.13: 49523487-49523487 on Build GRCh38
rs755441481 Chr.13: 49523162-49523163 on Build GRCh38
rs368988100 Chr.13: 49523181-49523181 on Build GRCh38
rs189067017 Chr.13: 49523349-49523349 on Build GRCh38
rs765184927 Chr.13: 49523198-49523198 on Build GRCh38
rs918872463 Chr.13: 49523325-49523325 on Build GRCh38
rs1030919625 Chr.13: 49523451-49523451 on Build GRCh38
rs906185085 Chr.13: 49523407-49523407 on Build GRCh38
rs781656818 Chr.13: 49523257-49523258 on Build GRCh38
rs944058006 Chr.13: 49523307-49523307 on Build GRCh38
rs766051904 Chr.13: 49523180-49523180 on Build GRCh38
rs371769077 Chr.13: 49523199-49523199 on Build GRCh38
rs989352804 Chr.13: 49523133-49523133 on Build GRCh38
rs921860598 Chr.13: 49523444-49523444 on Build GRCh38
rs977016080 Chr.13: 49523309-49523309 on Build GRCh38
rs763921888 Chr.13: 49523208-49523208 on Build GRCh38
rs368738874 Chr.13: 49523141-49523141 on Build GRCh38
rs541512162 Chr.13: 49523331-49523331 on Build GRCh38
rs760305458 Chr.13: 49523160-49523160 on Build GRCh38
rs558894359 Chr.13: 49523248-49523248 on Build GRCh38
rs1038047044 Chr.13: 49523381-49523381 on Build GRCh38
rs748715177 Chr.13: 49523288-49523288 on Build GRCh38
rs956518729 Chr.13: 49523114-49523114 on Build GRCh38
rs907735495 Chr.13: 49523341-49523341 on Build GRCh38
rs1053200266 Chr.13: 49523503-49523503 on Build GRCh38
rs1003654376 Chr.13: 49523413-49523413 on Build GRCh38
rs1048504658 Chr.13: 49523336-49523336 on Build GRCh38
rs139878500 Chr.13: 49523491-49523491 on Build GRCh38
rs751353603 Chr.13: 49523223-49523223 on Build GRCh38
rs759031178 Chr.13: 49523187-49523187 on Build GRCh38
rs956197011 Chr.13: 49523140-49523140 on Build GRCh38
rs372203739 Chr.13: 49523142-49523142 on Build GRCh38
rs758297378 Chr.13: 49523207-49523207 on Build GRCh38
rs369257467 Chr.13: 49523272-49523272 on Build GRCh38
rs547797568 Chr.13: 49523247-49523247 on Build GRCh38
rs533916455 Chr.13: 49523113-49523113 on Build GRCh38
rs959784573 Chr.13: 49523326-49523326 on Build GRCh38
rs911203929 Chr.13: 49523276-49523276 on Build GRCh38
rs755059426 Chr.13: 49523254-49523254 on Build GRCh38
rs576668503 Chr.13: 49523116-49523116 on Build GRCh38

View Full Product Details