Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 72579058-72579644 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHKA1
Entrez Gene ID
5255
Gene Name
phosphorylase kinase regulatory subunit alpha 1
Gene Aliases
PHKA
UniGene
Hs.201379
Gene Chromosome Location
Chr.X: 72578814-72714181 on Build GRCh38
Transcripts
NM_001122670.1 NM_001172436.1 NM_002637.3 XM_006724661.2

SNP Detail

SNP ID Chromosome Location
rs782659275 Chr.X: 72579137-72579137 on Build GRCh38
rs782553783 Chr.X: 72579096-72579096 on Build GRCh38
rs965598112 Chr.X: 72579139-72579139 on Build GRCh38
rs782589535 Chr.X: 72579553-72579553 on Build GRCh38
rs782370686 Chr.X: 72579391-72579391 on Build GRCh38
rs782017533 Chr.X: 72579344-72579344 on Build GRCh38
rs782174511 Chr.X: 72579278-72579278 on Build GRCh38
rs782434043 Chr.X: 72579325-72579325 on Build GRCh38
rs187366909 Chr.X: 72579457-72579457 on Build GRCh38
rs782074677 Chr.X: 72579498-72579498 on Build GRCh38
rs968587859 Chr.X: 72579169-72579169 on Build GRCh38
rs782338691 Chr.X: 72579230-72579230 on Build GRCh38
rs182287866 Chr.X: 72579190-72579190 on Build GRCh38
rs138901379 Chr.X: 72579274-72579274 on Build GRCh38
rs975219995 Chr.X: 72579153-72579153 on Build GRCh38
rs782495214 Chr.X: 72579507-72579507 on Build GRCh38
rs1004349600 Chr.X: 72579104-72579104 on Build GRCh38
rs781851235 Chr.X: 72579444-72579444 on Build GRCh38
rs979460666 Chr.X: 72579193-72579193 on Build GRCh38
rs34811076 Chr.X: 72579588-72579588 on Build GRCh38
rs925256988 Chr.X: 72579217-72579217 on Build GRCh38
rs551977025 Chr.X: 72579383-72579383 on Build GRCh38
rs782700376 Chr.X: 72579550-72579550 on Build GRCh38
rs905489059 Chr.X: 72579595-72579595 on Build GRCh38
rs921185285 Chr.X: 72579156-72579156 on Build GRCh38
rs781909431 Chr.X: 72579583-72579583 on Build GRCh38

View Full Product Details