Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 133229723-133230308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WISP1
Entrez Gene ID
8840
Gene Name
WNT1 inducible signaling pathway protein 1
Gene Aliases
CCN4, WISP1c, WISP1i, WISP1tc
UniGene
Hs.492974
Gene Chromosome Location
Chr.8: 133191039-133231690 on Build GRCh38
Transcripts
NM_001204869.1 NM_001204870.1 NM_003882.3 NM_080838.2 NR_037944.1

SNP Detail

SNP ID Chromosome Location
rs548959266 Chr.8: 133230238-133230238 on Build GRCh38
rs955239570 Chr.8: 133230254-133230254 on Build GRCh38
rs1054834278 Chr.8: 133230149-133230149 on Build GRCh38
rs1043524547 Chr.8: 133230078-133230078 on Build GRCh38
rs993574386 Chr.8: 133229932-133229932 on Build GRCh38
rs763422145 Chr.8: 133229832-133229832 on Build GRCh38
rs142427022 Chr.8: 133230160-133230160 on Build GRCh38
rs553399878 Chr.8: 133230108-133230108 on Build GRCh38
rs117072299 Chr.8: 133229757-133229757 on Build GRCh38
rs925051954 Chr.8: 133230112-133230112 on Build GRCh38
rs922482260 Chr.8: 133229922-133229922 on Build GRCh38
rs939893297 Chr.8: 133229858-133229858 on Build GRCh38
rs2977549 Chr.8: 133229790-133229790 on Build GRCh38
rs922430001 Chr.8: 133230244-133230244 on Build GRCh38
rs1026831190 Chr.8: 133229944-133229944 on Build GRCh38
rs2929972 Chr.8: 133230212-133230212 on Build GRCh38
rs560933676 Chr.8: 133230264-133230264 on Build GRCh38
rs970122155 Chr.8: 133230174-133230174 on Build GRCh38
rs761936939 Chr.8: 133230100-133230100 on Build GRCh38
rs933912518 Chr.8: 133229948-133229948 on Build GRCh38
rs764344005 Chr.8: 133229996-133229996 on Build GRCh38
rs972534681 Chr.8: 133230217-133230217 on Build GRCh38
rs907341350 Chr.8: 133230175-133230175 on Build GRCh38
rs984780696 Chr.8: 133229973-133229973 on Build GRCh38
rs550033135 Chr.8: 133230197-133230197 on Build GRCh38
rs73362557 Chr.8: 133230093-133230093 on Build GRCh38
rs1046424811 Chr.8: 133230175-133230175 on Build GRCh38
rs972632269 Chr.8: 133229894-133229894 on Build GRCh38
rs1026381143 Chr.8: 133230224-133230224 on Build GRCh38
rs1054698739 Chr.8: 133229771-133229771 on Build GRCh38
rs2929971 Chr.8: 133230129-133230129 on Build GRCh38
rs373530385 Chr.8: 133230136-133230136 on Build GRCh38
rs2929973 Chr.8: 133230265-133230265 on Build GRCh38
rs909703857 Chr.8: 133229841-133229841 on Build GRCh38
rs76779268 Chr.8: 133230095-133230095 on Build GRCh38
rs984593097 Chr.8: 133230196-133230196 on Build GRCh38
rs555686520 Chr.8: 133229992-133229992 on Build GRCh38
rs563542574 Chr.8: 133230146-133230146 on Build GRCh38
rs971976735 Chr.8: 133229895-133229895 on Build GRCh38
rs946451988 Chr.8: 133230042-133230042 on Build GRCh38
rs889465185 Chr.8: 133230007-133230007 on Build GRCh38
rs578093828 Chr.8: 133230123-133230123 on Build GRCh38
rs923089223 Chr.8: 133230082-133230082 on Build GRCh38

View Full Product Details