Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 228145179-228145731 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GUK1
Entrez Gene ID
2987
Gene Name
guanylate kinase 1
Gene Aliases
GMK
UniGene
Hs.376933
Gene Chromosome Location
Chr.1: 228139643-228148955 on Build GRCh38
Transcripts
NM_000858.5 NM_001159390.1 NM_001159391.1 NM_001242839.1 NM_001242840.1 XM_005273103.3 XM_005273104.4 XM_005273105.3 XM_005273106.2 XM_011544174.2 XM_017001091.1 XM_017001092.1 XM_017001093.1 XM_017001094.1 XM_017001095.1 XM_017001096.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5993448 c.304C>T p.R102W Chr.1: 228145599-228145599 on Build GRCh38
6061608 c.283G>A p.E95K Chr.1: 228145578-228145578 on Build GRCh38
4028867 c.30C>T p.S10S Chr.1: 228145542-228145542 on Build GRCh38
4028864 c.8G>C p.G3A Chr.1: 228145520-228145520 on Build GRCh38
5993450 c.87C>T p.S29S Chr.1: 228145599-228145599 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1052180945 Chr.1: 228145245-228145245 on Build GRCh38
rs549081705 Chr.1: 228145642-228145642 on Build GRCh38
rs898544342 Chr.1: 228145316-228145316 on Build GRCh38
rs368643123 Chr.1: 228145653-228145653 on Build GRCh38
rs1023083609 Chr.1: 228145650-228145650 on Build GRCh38
rs555937041 Chr.1: 228145340-228145340 on Build GRCh38
rs760489276 Chr.1: 228145494-228145494 on Build GRCh38
rs768734736 Chr.1: 228145450-228145450 on Build GRCh38
rs964071558 Chr.1: 228145624-228145624 on Build GRCh38
rs369835349 Chr.1: 228145654-228145654 on Build GRCh38
rs770886564 Chr.1: 228145696-228145696 on Build GRCh38
rs372956407 Chr.1: 228145521-228145521 on Build GRCh38
rs545173663 Chr.1: 228145399-228145399 on Build GRCh38
rs76106352 Chr.1: 228145488-228145488 on Build GRCh38
rs967937913 Chr.1: 228145697-228145697 on Build GRCh38
rs368505522 Chr.1: 228145517-228145517 on Build GRCh38
rs56234884 Chr.1: 228145311-228145311 on Build GRCh38
rs952924153 Chr.1: 228145546-228145546 on Build GRCh38
rs752045940 Chr.1: 228145518-228145518 on Build GRCh38
rs565919531 Chr.1: 228145690-228145690 on Build GRCh38
rs932355338 Chr.1: 228145588-228145588 on Build GRCh38
rs542954199 Chr.1: 228145679-228145679 on Build GRCh38
rs748265827 Chr.1: 228145662-228145662 on Build GRCh38
rs761782492 Chr.1: 228145468-228145468 on Build GRCh38
rs565054231 Chr.1: 228145417-228145417 on Build GRCh38
rs200162832 Chr.1: 228145495-228145495 on Build GRCh38
rs772781548 Chr.1: 228145504-228145504 on Build GRCh38
rs757777889 Chr.1: 228145520-228145520 on Build GRCh38
rs1051319665 Chr.1: 228145289-228145289 on Build GRCh38
rs970976975 Chr.1: 228145242-228145242 on Build GRCh38
rs879336994 Chr.1: 228145282-228145282 on Build GRCh38
rs552281965 Chr.1: 228145688-228145688 on Build GRCh38
rs748097697 Chr.1: 228145592-228145592 on Build GRCh38
rs764781035 Chr.1: 228145510-228145510 on Build GRCh38
rs15112 Chr.1: 228145596-228145596 on Build GRCh38
rs373627017 Chr.1: 228145599-228145599 on Build GRCh38
rs756401161 Chr.1: 228145543-228145543 on Build GRCh38
rs377185198 Chr.1: 228145475-228145475 on Build GRCh38
rs576213063 Chr.1: 228145385-228145385 on Build GRCh38
rs979586005 Chr.1: 228145703-228145703 on Build GRCh38
rs747874663 Chr.1: 228145235-228145235 on Build GRCh38
rs946496631 Chr.1: 228145270-228145270 on Build GRCh38
rs939200710 Chr.1: 228145240-228145240 on Build GRCh38
rs774207783 Chr.1: 228145456-228145456 on Build GRCh38
rs772149695 Chr.1: 228145597-228145597 on Build GRCh38
rs980676423 Chr.1: 228145418-228145418 on Build GRCh38
rs894884030 Chr.1: 228145438-228145438 on Build GRCh38
rs759195112 Chr.1: 228145625-228145625 on Build GRCh38
rs1000698950 Chr.1: 228145365-228145365 on Build GRCh38
rs151031420 Chr.1: 228145269-228145269 on Build GRCh38
rs891342326 Chr.1: 228145606-228145606 on Build GRCh38
rs1015768927 Chr.1: 228145315-228145315 on Build GRCh38
rs1035701845 Chr.1: 228145669-228145669 on Build GRCh38
rs898685313 Chr.1: 228145632-228145632 on Build GRCh38
rs776194580 Chr.1: 228145610-228145610 on Build GRCh38
rs140917603 Chr.1: 228145551-228145551 on Build GRCh38
rs199673509 Chr.1: 228145617-228145617 on Build GRCh38
rs529314238 Chr.1: 228145281-228145281 on Build GRCh38
rs1034974805 Chr.1: 228145448-228145448 on Build GRCh38
rs200265279 Chr.1: 228145627-228145627 on Build GRCh38
rs146668917 Chr.1: 228145641-228145641 on Build GRCh38
rs776309977 Chr.1: 228145497-228145497 on Build GRCh38
rs536265267 Chr.1: 228145288-228145288 on Build GRCh38
rs920917312 Chr.1: 228145586-228145586 on Build GRCh38
rs749506347 Chr.1: 228145565-228145565 on Build GRCh38
rs891288537 Chr.1: 228145290-228145290 on Build GRCh38
rs773127917 Chr.1: 228145476-228145476 on Build GRCh38
rs759079640 Chr.1: 228145516-228145516 on Build GRCh38
rs770798621 Chr.1: 228145602-228145602 on Build GRCh38
rs367796074 Chr.1: 228145600-228145600 on Build GRCh38
rs762387688 Chr.1: 228145640-228145640 on Build GRCh38
rs140072078 Chr.1: 228145542-228145542 on Build GRCh38
rs979230614 Chr.1: 228145569-228145569 on Build GRCh38
rs766701509 Chr.1: 228145652-228145652 on Build GRCh38
rs779050123 Chr.1: 228145580-228145580 on Build GRCh38
rs762617116 Chr.1: 228145551-228145551 on Build GRCh38
rs369394197 Chr.1: 228145636-228145636 on Build GRCh38
rs931399365 Chr.1: 228145272-228145272 on Build GRCh38
rs141117528 Chr.1: 228145539-228145539 on Build GRCh38
rs995621695 Chr.1: 228145321-228145321 on Build GRCh38
rs1023368030 Chr.1: 228145359-228145359 on Build GRCh38
rs1040993105 Chr.1: 228145305-228145305 on Build GRCh38
rs370118444 Chr.1: 228145502-228145502 on Build GRCh38
rs901772819 Chr.1: 228145307-228145307 on Build GRCh38
rs766320920 Chr.1: 228145297-228145297 on Build GRCh38
rs371593150 Chr.1: 228145601-228145601 on Build GRCh38
rs755056625 Chr.1: 228145572-228145572 on Build GRCh38

View Full Product Details