Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 116111641-116112055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAB21L3
Entrez Gene ID
126868
Gene Name
mab-21 like 3
Gene Aliases
C1orf161
UniGene
Hs.376194
Gene Chromosome Location
Chr.1: 116072744-116135240 on Build GRCh38
Transcripts
NM_152367.2 XM_011540623.2 XM_011540625.2 XM_011540626.2 XM_017000260.1

SNP Detail

SNP ID Chromosome Location
rs969837983 Chr.1: 116111794-116111794 on Build GRCh38
rs1006425630 Chr.1: 116111685-116111685 on Build GRCh38
rs1021163568 Chr.1: 116111922-116111922 on Build GRCh38
rs770101366 Chr.1: 116111951-116111951 on Build GRCh38
rs565241303 Chr.1: 116111713-116111713 on Build GRCh38
rs886902357 Chr.1: 116111854-116111854 on Build GRCh38
rs79725369 Chr.1: 116111874-116111874 on Build GRCh38
rs1013829485 Chr.1: 116111912-116111912 on Build GRCh38
rs532590350 Chr.1: 116111763-116111763 on Build GRCh38
rs552693610 Chr.1: 116111988-116111988 on Build GRCh38
rs923333787 Chr.1: 116111901-116111901 on Build GRCh38
rs925676518 Chr.1: 116112014-116112014 on Build GRCh38
rs935644087 Chr.1: 116111775-116111775 on Build GRCh38
rs762184259 Chr.1: 116112019-116112019 on Build GRCh38
rs377036452 Chr.1: 116111948-116111948 on Build GRCh38
rs761759429 Chr.1: 116111933-116111933 on Build GRCh38
rs774661132 Chr.1: 116111859-116111859 on Build GRCh38
rs370092883 Chr.1: 116111865-116111865 on Build GRCh38
rs186526649 Chr.1: 116111928-116111928 on Build GRCh38
rs772780171 Chr.1: 116112006-116112007 on Build GRCh38
rs768866238 Chr.1: 116111824-116111824 on Build GRCh38
rs925739793 Chr.1: 116111748-116111748 on Build GRCh38
rs1004056242 Chr.1: 116111857-116111857 on Build GRCh38
rs888045790 Chr.1: 116111983-116111983 on Build GRCh38
rs148944185 Chr.1: 116111911-116111911 on Build GRCh38
rs1039396824 Chr.1: 116111795-116111795 on Build GRCh38
rs993484763 Chr.1: 116111667-116111667 on Build GRCh38
rs1005826879 Chr.1: 116111830-116111830 on Build GRCh38
rs914532202 Chr.1: 116111864-116111864 on Build GRCh38
rs899737610 Chr.1: 116111820-116111820 on Build GRCh38
rs979644133 Chr.1: 116111976-116111976 on Build GRCh38
rs531022381 Chr.1: 116111947-116111947 on Build GRCh38
rs745743925 Chr.1: 116111852-116111852 on Build GRCh38
rs138713237 Chr.1: 116111908-116111908 on Build GRCh38
rs945795470 Chr.1: 116111709-116111709 on Build GRCh38
rs1006847605 Chr.1: 116112002-116112002 on Build GRCh38
rs190917975 Chr.1: 116112023-116112023 on Build GRCh38
rs888024802 Chr.1: 116111685-116111685 on Build GRCh38
rs960935295 Chr.1: 116111892-116111892 on Build GRCh38
rs548469991 Chr.1: 116111918-116111918 on Build GRCh38
rs779841689 Chr.1: 116111873-116111873 on Build GRCh38
rs911005599 Chr.1: 116111870-116111870 on Build GRCh38
rs144773588 Chr.1: 116111668-116111668 on Build GRCh38
rs1050914420 Chr.1: 116111851-116111851 on Build GRCh38
rs977542766 Chr.1: 116111723-116111723 on Build GRCh38
rs199576437 Chr.1: 116112020-116112020 on Build GRCh38
rs543934898 Chr.1: 116111710-116111710 on Build GRCh38

View Full Product Details