Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 119262925-119263480 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STEAP3
Entrez Gene ID
55240
Gene Name
STEAP3 metalloreductase
Gene Aliases
AHMIO2, STMP3, TSAP6, dudlin-2, dudulin-2, pHyde
UniGene
Hs.647822
Gene Chromosome Location
Chr.2: 119222476-119265652 on Build GRCh38
Transcripts
NM_001008410.1 NM_018234.2 NM_138637.2 NM_182915.2 XM_006712614.3 XM_006712615.1 XM_011511403.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3961051 c.1329G>A p.T443T Chr.2: 119263170-119263170 on Build GRCh38
4730504 c.1348C>A p.L450I Chr.2: 119263219-119263219 on Build GRCh38
39553 c.1321G>A p.V441I Chr.2: 119263192-119263192 on Build GRCh38
4527413 c.1462G>A p.D488N Chr.2: 119263303-119263303 on Build GRCh38
1669232 c.1240G>A p.G414S Chr.2: 119263111-119263111 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752735656 Chr.2: 119263080-119263080 on Build GRCh38
rs768264100 Chr.2: 119263036-119263036 on Build GRCh38
rs760495654 Chr.2: 119263255-119263255 on Build GRCh38
rs139180247 Chr.2: 119263238-119263238 on Build GRCh38
rs537470551 Chr.2: 119263332-119263332 on Build GRCh38
rs200076386 Chr.2: 119263100-119263100 on Build GRCh38
rs35619518 Chr.2: 119263292-119263292 on Build GRCh38
rs762593731 Chr.2: 119263067-119263067 on Build GRCh38
rs933604351 Chr.2: 119263417-119263417 on Build GRCh38
rs758832778 Chr.2: 119263226-119263226 on Build GRCh38
rs753992611 Chr.2: 119263151-119263151 on Build GRCh38
rs745497289 Chr.2: 119263298-119263298 on Build GRCh38
rs777797898 Chr.2: 119263092-119263092 on Build GRCh38
rs774627341 Chr.2: 119263409-119263409 on Build GRCh38
rs11885203 Chr.2: 119263059-119263059 on Build GRCh38
rs761969248 Chr.2: 119263208-119263208 on Build GRCh38
rs200363679 Chr.2: 119263065-119263065 on Build GRCh38
rs781682924 Chr.2: 119263278-119263278 on Build GRCh38
rs139983521 Chr.2: 119263186-119263186 on Build GRCh38
rs574854030 Chr.2: 119263408-119263408 on Build GRCh38
rs376692800 Chr.2: 119263123-119263123 on Build GRCh38
rs546900400 Chr.2: 119263128-119263128 on Build GRCh38
rs148332588 Chr.2: 119263110-119263110 on Build GRCh38
rs561256507 Chr.2: 119263118-119263118 on Build GRCh38
rs886518078 Chr.2: 119263289-119263289 on Build GRCh38
rs778595169 Chr.2: 119263297-119263297 on Build GRCh38
rs999476826 Chr.2: 119263391-119263391 on Build GRCh38
rs766265633 Chr.2: 119263364-119263365 on Build GRCh38
rs764443015 Chr.2: 119263143-119263143 on Build GRCh38
rs915064861 Chr.2: 119262978-119262978 on Build GRCh38
rs1003178106 Chr.2: 119262964-119262964 on Build GRCh38
rs768319229 Chr.2: 119263111-119263111 on Build GRCh38
rs370568683 Chr.2: 119263380-119263380 on Build GRCh38
rs201034733 Chr.2: 119263204-119263204 on Build GRCh38
rs776629232 Chr.2: 119263312-119263312 on Build GRCh38
rs771480674 Chr.2: 119263020-119263020 on Build GRCh38
rs914811409 Chr.2: 119263035-119263035 on Build GRCh38
rs970462260 Chr.2: 119262963-119262963 on Build GRCh38
rs371117240 Chr.2: 119263293-119263293 on Build GRCh38
rs748236640 Chr.2: 119263317-119263317 on Build GRCh38
rs373263111 Chr.2: 119263237-119263237 on Build GRCh38
rs911951717 Chr.2: 119263026-119263026 on Build GRCh38
rs776470054 Chr.2: 119263050-119263050 on Build GRCh38
rs760800930 Chr.2: 119263195-119263195 on Build GRCh38
rs983616892 Chr.2: 119263419-119263419 on Build GRCh38
rs150254604 Chr.2: 119263326-119263326 on Build GRCh38
rs772740148 Chr.2: 119263121-119263121 on Build GRCh38
rs910924823 Chr.2: 119263189-119263189 on Build GRCh38
rs777179124 Chr.2: 119263235-119263235 on Build GRCh38
rs766410185 Chr.2: 119263071-119263071 on Build GRCh38
rs767900764 Chr.2: 119263432-119263432 on Build GRCh38
rs749324872 Chr.2: 119263406-119263406 on Build GRCh38
rs765728429 Chr.2: 119263039-119263040 on Build GRCh38
rs1012548941 Chr.2: 119263314-119263314 on Build GRCh38
rs761197484 Chr.2: 119263456-119263456 on Build GRCh38
rs753232184 Chr.2: 119263364-119263364 on Build GRCh38
rs775455207 Chr.2: 119263249-119263249 on Build GRCh38
rs1033767631 Chr.2: 119262965-119262965 on Build GRCh38
rs767394433 Chr.2: 119263129-119263129 on Build GRCh38
rs140239610 Chr.2: 119263308-119263308 on Build GRCh38
rs149348257 Chr.2: 119262973-119262973 on Build GRCh38
rs750665147 Chr.2: 119263267-119263267 on Build GRCh38
rs978221233 Chr.2: 119263175-119263175 on Build GRCh38
rs758064666 Chr.2: 119263373-119263373 on Build GRCh38
rs371313940 Chr.2: 119263152-119263152 on Build GRCh38
rs957778416 Chr.2: 119263345-119263345 on Build GRCh38
rs776593892 Chr.2: 119263256-119263256 on Build GRCh38
rs779111239 Chr.2: 119263155-119263155 on Build GRCh38
rs952169593 Chr.2: 119263457-119263457 on Build GRCh38
rs374652248 Chr.2: 119263170-119263170 on Build GRCh38
rs759879690 Chr.2: 119263333-119263333 on Build GRCh38
rs144758696 Chr.2: 119263139-119263139 on Build GRCh38
rs147303947 Chr.2: 119263365-119263365 on Build GRCh38
rs200025915 Chr.2: 119263101-119263101 on Build GRCh38
rs747001332 Chr.2: 119263173-119263173 on Build GRCh38
rs1015272512 Chr.2: 119262960-119262960 on Build GRCh38
rs765448684 Chr.2: 119263211-119263211 on Build GRCh38
rs780769514 Chr.2: 119263229-119263229 on Build GRCh38
rs768487970 Chr.2: 119263309-119263309 on Build GRCh38
rs774654393 Chr.2: 119263024-119263024 on Build GRCh38
rs977938794 Chr.2: 119263025-119263025 on Build GRCh38
rs368774790 Chr.2: 119263141-119263141 on Build GRCh38
rs755099329 Chr.2: 119263075-119263075 on Build GRCh38
rs771841410 Chr.2: 119263246-119263246 on Build GRCh38
rs769576483 Chr.2: 119263120-119263120 on Build GRCh38
rs1003897809 Chr.2: 119262954-119262954 on Build GRCh38
rs886635256 Chr.2: 119262948-119262948 on Build GRCh38
rs200231027 Chr.2: 119263257-119263257 on Build GRCh38
rs778232548 Chr.2: 119263019-119263019 on Build GRCh38
rs556534362 Chr.2: 119263394-119263394 on Build GRCh38
rs549220081 Chr.2: 119263114-119263114 on Build GRCh38
rs756350021 Chr.2: 119263090-119263090 on Build GRCh38
rs538794610 Chr.2: 119263169-119263169 on Build GRCh38
rs779528562 Chr.2: 119263106-119263106 on Build GRCh38
rs923578207 Chr.2: 119262975-119262975 on Build GRCh38
rs867457574 Chr.2: 119263197-119263197 on Build GRCh38
rs544816246 Chr.2: 119263381-119263381 on Build GRCh38
rs777530731 Chr.2: 119263176-119263176 on Build GRCh38
rs141086904 Chr.2: 119263325-119263325 on Build GRCh38
rs765446095 Chr.2: 119263261-119263261 on Build GRCh38
rs750824808 Chr.2: 119263212-119263212 on Build GRCh38
rs779851052 Chr.2: 119263306-119263306 on Build GRCh38
rs61746941 Chr.2: 119263179-119263179 on Build GRCh38
rs200886089 Chr.2: 119263318-119263318 on Build GRCh38
rs974618962 Chr.2: 119263434-119263434 on Build GRCh38
rs755463675 Chr.2: 119263273-119263273 on Build GRCh38
rs963767138 Chr.2: 119263368-119263368 on Build GRCh38
rs751932904 Chr.2: 119263271-119263271 on Build GRCh38
rs759267719 Chr.2: 119263191-119263191 on Build GRCh38
rs201774653 Chr.2: 119263058-119263058 on Build GRCh38
rs746192768 Chr.2: 119263156-119263156 on Build GRCh38
rs140893688 Chr.2: 119263438-119263438 on Build GRCh38
rs368978605 Chr.2: 119263194-119263194 on Build GRCh38
rs370504846 Chr.2: 119263028-119263028 on Build GRCh38
rs923536484 Chr.2: 119263375-119263375 on Build GRCh38
rs144013888 Chr.2: 119263269-119263269 on Build GRCh38
rs867505960 Chr.2: 119262992-119262992 on Build GRCh38
rs751439859 Chr.2: 119263074-119263074 on Build GRCh38
rs754125354 Chr.2: 119263205-119263205 on Build GRCh38
rs201407133 Chr.2: 119263372-119263372 on Build GRCh38
rs752094440 Chr.2: 119263230-119263230 on Build GRCh38
rs759874514 Chr.2: 119263426-119263426 on Build GRCh38
rs771138329 Chr.2: 119263393-119263393 on Build GRCh38
rs771999099 Chr.2: 119263192-119263192 on Build GRCh38
rs377489086 Chr.2: 119263242-119263242 on Build GRCh38
rs577685644 Chr.2: 119263055-119263055 on Build GRCh38
rs763463275 Chr.2: 119263268-119263268 on Build GRCh38
rs375934185 Chr.2: 119263301-119263301 on Build GRCh38
rs767555801 Chr.2: 119263078-119263078 on Build GRCh38
rs779621353 Chr.2: 119263379-119263379 on Build GRCh38
rs755691787 Chr.2: 119263233-119263233 on Build GRCh38
rs767974551 Chr.2: 119263341-119263341 on Build GRCh38
rs111723879 Chr.2: 119263122-119263122 on Build GRCh38
rs779830150 Chr.2: 119263161-119263161 on Build GRCh38
rs752786678 Chr.2: 119263138-119263138 on Build GRCh38

View Full Product Details