Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 96442367-96442858 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AK7
Entrez Gene ID
122481
Gene Name
adenylate kinase 7
Gene Aliases
AK 7
UniGene
Hs.667462
Gene Chromosome Location
Chr.14: 96392111-96489427 on Build GRCh38
Transcripts
NM_152327.3 XM_006720021.2 XM_006720022.2 XM_011536403.2 XM_017020956.1 XM_017020957.1 XM_017020958.1 XM_017020959.1 XM_017020960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7305354 c.705C>T p.G235G Chr.14: 96442744-96442744 on Build GRCh38
3815628 c.706G>A p.E236K Chr.14: 96442745-96442745 on Build GRCh38
699682 c.706G>T p.E236* Chr.14: 96442745-96442745 on Build GRCh38
6303750 c.779G>A p.G260E Chr.14: 96442818-96442818 on Build GRCh38
6870985 c.691-1G>A p.? Chr.14: 96442729-96442729 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs558036076 Chr.14: 96442407-96442407 on Build GRCh38
rs752469689 Chr.14: 96442735-96442735 on Build GRCh38
rs577915046 Chr.14: 96442608-96442608 on Build GRCh38
rs770027579 Chr.14: 96442627-96442627 on Build GRCh38
rs899971886 Chr.14: 96442494-96442494 on Build GRCh38
rs373955939 Chr.14: 96442771-96442771 on Build GRCh38
rs749498808 Chr.14: 96442686-96442686 on Build GRCh38
rs114393353 Chr.14: 96442745-96442745 on Build GRCh38
rs756645092 Chr.14: 96442774-96442774 on Build GRCh38
rs1049631821 Chr.14: 96442567-96442567 on Build GRCh38
rs761625542 Chr.14: 96442687-96442687 on Build GRCh38
rs934438544 Chr.14: 96442458-96442458 on Build GRCh38
rs1011772591 Chr.14: 96442538-96442538 on Build GRCh38
rs112200493 Chr.14: 96442775-96442775 on Build GRCh38
rs750987024 Chr.14: 96442772-96442772 on Build GRCh38
rs762525188 Chr.14: 96442744-96442744 on Build GRCh38
rs552890966 Chr.14: 96442603-96442603 on Build GRCh38
rs922434631 Chr.14: 96442513-96442513 on Build GRCh38
rs909834272 Chr.14: 96442580-96442580 on Build GRCh38
rs912193746 Chr.14: 96442789-96442789 on Build GRCh38
rs769661121 Chr.14: 96442832-96442832 on Build GRCh38
rs916503827 Chr.14: 96442678-96442678 on Build GRCh38
rs968348631 Chr.14: 96442429-96442429 on Build GRCh38
rs756794218 Chr.14: 96442761-96442761 on Build GRCh38
rs929766096 Chr.14: 96442754-96442754 on Build GRCh38
rs569274238 Chr.14: 96442480-96442480 on Build GRCh38
rs1003607772 Chr.14: 96442675-96442675 on Build GRCh38
rs941337844 Chr.14: 96442601-96442601 on Build GRCh38
rs1022502185 Chr.14: 96442697-96442697 on Build GRCh38
rs763111680 Chr.14: 96442713-96442713 on Build GRCh38
rs749654662 Chr.14: 96442807-96442807 on Build GRCh38
rs148054558 Chr.14: 96442656-96442656 on Build GRCh38
rs184769808 Chr.14: 96442484-96442484 on Build GRCh38
rs541164114 Chr.14: 96442572-96442572 on Build GRCh38
rs768831247 Chr.14: 96442821-96442821 on Build GRCh38
rs189498856 Chr.14: 96442829-96442829 on Build GRCh38
rs573640514 Chr.14: 96442553-96442553 on Build GRCh38
rs991051032 Chr.14: 96442390-96442390 on Build GRCh38
rs1035733654 Chr.14: 96442605-96442605 on Build GRCh38
rs370055465 Chr.14: 96442825-96442825 on Build GRCh38
rs770897429 Chr.14: 96442694-96442694 on Build GRCh38
rs932480583 Chr.14: 96442533-96442533 on Build GRCh38
rs982939189 Chr.14: 96442755-96442755 on Build GRCh38
rs761854868 Chr.14: 96442828-96442828 on Build GRCh38
rs1012866001 Chr.14: 96442693-96442693 on Build GRCh38
rs113115430 Chr.14: 96442544-96442544 on Build GRCh38
rs765049636 Chr.14: 96442718-96442718 on Build GRCh38
rs924344522 Chr.14: 96442451-96442451 on Build GRCh38
rs999804107 Chr.14: 96442803-96442803 on Build GRCh38
rs978438201 Chr.14: 96442450-96442450 on Build GRCh38
rs764335929 Chr.14: 96442719-96442719 on Build GRCh38
rs897173330 Chr.14: 96442648-96442648 on Build GRCh38
rs929630498 Chr.14: 96442416-96442416 on Build GRCh38
rs12431832 Chr.14: 96442688-96442688 on Build GRCh38
rs1046748680 Chr.14: 96442445-96442445 on Build GRCh38
rs776529954 Chr.14: 96442698-96442698 on Build GRCh38
rs967485431 Chr.14: 96442560-96442560 on Build GRCh38
rs1040370135 Chr.14: 96442464-96442464 on Build GRCh38
rs780567134 Chr.14: 96442800-96442800 on Build GRCh38
rs751475113 Chr.14: 96442742-96442742 on Build GRCh38
rs868058442 Chr.14: 96442751-96442751 on Build GRCh38
rs555371136 Chr.14: 96442502-96442502 on Build GRCh38
rs759421358 Chr.14: 96442707-96442707 on Build GRCh38

View Full Product Details