Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 82547121-82547634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPEB1
Entrez Gene ID
64506
Gene Name
cytoplasmic polyadenylation element binding protein 1
Gene Aliases
CPE-BP1, CPEB, CPEB-1, h-CPEB, hCPEB-1
UniGene
Hs.459132
Gene Chromosome Location
Chr.15: 82543201-82648016 on Build GRCh38
Transcripts
NM_001079533.1 NM_001079534.1 NM_001079535.1 NM_001288819.1 NM_001288820.1 NM_030594.4

SNP Detail

SNP ID Chromosome Location
rs755328188 Chr.15: 82547286-82547288 on Build GRCh38
rs200803573 Chr.15: 82547207-82547207 on Build GRCh38
rs944760500 Chr.15: 82547465-82547465 on Build GRCh38
rs904902116 Chr.15: 82547343-82547343 on Build GRCh38
rs1022201890 Chr.15: 82547428-82547428 on Build GRCh38
rs768160786 Chr.15: 82547161-82547161 on Build GRCh38
rs748591767 Chr.15: 82547290-82547291 on Build GRCh38
rs936796634 Chr.15: 82547511-82547511 on Build GRCh38
rs373749169 Chr.15: 82547184-82547184 on Build GRCh38
rs577671702 Chr.15: 82547457-82547457 on Build GRCh38
rs140456585 Chr.15: 82547441-82547441 on Build GRCh38
rs767234807 Chr.15: 82547269-82547269 on Build GRCh38
rs777300594 Chr.15: 82547290-82547304 on Build GRCh38
rs951357657 Chr.15: 82547423-82547423 on Build GRCh38
rs1001204679 Chr.15: 82547289-82547289 on Build GRCh38
rs755621912 Chr.15: 82547290-82547301 on Build GRCh38
rs563611424 Chr.15: 82547254-82547254 on Build GRCh38
rs995594364 Chr.15: 82547336-82547336 on Build GRCh38
rs1049298382 Chr.15: 82547216-82547216 on Build GRCh38
rs556449821 Chr.15: 82547357-82547357 on Build GRCh38
rs988526823 Chr.15: 82547537-82547537 on Build GRCh38
rs976291204 Chr.15: 82547481-82547481 on Build GRCh38
rs778396392 Chr.15: 82547315-82547346 on Build GRCh38
rs779375273 Chr.15: 82547151-82547151 on Build GRCh38
rs770669109 Chr.15: 82547290-82547300 on Build GRCh38
rs925378686 Chr.15: 82547501-82547501 on Build GRCh38
rs747825334 Chr.15: 82547165-82547165 on Build GRCh38
rs929477044 Chr.15: 82547530-82547530 on Build GRCh38
rs1035236108 Chr.15: 82547378-82547378 on Build GRCh38
rs35309965 Chr.15: 82547255-82547255 on Build GRCh38
rs71156035 Chr.15: 82547314-82547314 on Build GRCh38
rs750233563 Chr.15: 82547271-82547271 on Build GRCh38
rs772522093 Chr.15: 82547173-82547173 on Build GRCh38
rs1056200195 Chr.15: 82547595-82547595 on Build GRCh38
rs776571408 Chr.15: 82547224-82547224 on Build GRCh38
rs909461182 Chr.15: 82547542-82547542 on Build GRCh38
rs748919206 Chr.15: 82547289-82547290 on Build GRCh38
rs150409941 Chr.15: 82547495-82547495 on Build GRCh38
rs372556871 Chr.15: 82547487-82547487 on Build GRCh38
rs572060484 Chr.15: 82547358-82547358 on Build GRCh38
rs755999483 Chr.15: 82547274-82547274 on Build GRCh38
rs765426128 Chr.15: 82547249-82547249 on Build GRCh38
rs377016068 Chr.15: 82547284-82547284 on Build GRCh38
rs762667358 Chr.15: 82547576-82547576 on Build GRCh38
rs761026665 Chr.15: 82547190-82547190 on Build GRCh38
rs374011661 Chr.15: 82547185-82547185 on Build GRCh38
rs1003271890 Chr.15: 82547593-82547593 on Build GRCh38
rs748893773 Chr.15: 82547158-82547158 on Build GRCh38
rs781704966 Chr.15: 82547290-82547290 on Build GRCh38
rs547610843 Chr.15: 82547282-82547282 on Build GRCh38
rs373799548 Chr.15: 82547327-82547327 on Build GRCh38
rs560295615 Chr.15: 82547573-82547573 on Build GRCh38
rs766816215 Chr.15: 82547414-82547414 on Build GRCh38
rs949659321 Chr.15: 82547541-82547541 on Build GRCh38
rs576281305 Chr.15: 82547389-82547389 on Build GRCh38
rs146805892 Chr.15: 82547325-82547325 on Build GRCh38
rs1034064023 Chr.15: 82547290-82547290 on Build GRCh38
rs941006743 Chr.15: 82547554-82547554 on Build GRCh38
rs763194835 Chr.15: 82547256-82547256 on Build GRCh38
rs778756628 Chr.15: 82547162-82547162 on Build GRCh38
rs1025337714 Chr.15: 82547421-82547421 on Build GRCh38
rs913942016 Chr.15: 82547540-82547540 on Build GRCh38
rs904954785 Chr.15: 82547574-82547574 on Build GRCh38
rs368755551 Chr.15: 82547390-82547390 on Build GRCh38
rs752868471 Chr.15: 82547254-82547254 on Build GRCh38
rs956670185 Chr.15: 82547396-82547396 on Build GRCh38
rs907731787 Chr.15: 82547533-82547533 on Build GRCh38
rs759706108 Chr.15: 82547244-82547244 on Build GRCh38
rs986755822 Chr.15: 82547439-82547439 on Build GRCh38
rs986092848 Chr.15: 82547452-82547452 on Build GRCh38
rs775248512 Chr.15: 82547500-82547500 on Build GRCh38
rs557741204 Chr.15: 82547450-82547450 on Build GRCh38
rs969884038 Chr.15: 82547358-82547373 on Build GRCh38

View Full Product Details