Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 77199378-77199934 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MON1B
Entrez Gene ID
22879
Gene Name
MON1 homolog B, secretory trafficking associated
Gene Aliases
HSRG1, SAND2, SRG1
UniGene
Hs.513743
Gene Chromosome Location
Chr.16: 77190919-77199646 on Build GRCh38
Transcripts
NM_001286639.1 NM_001286640.1 NM_014940.3 NM_001129979.1

Gene Symbol
SYCE1L
Entrez Gene ID
100130958
Gene Name
synaptonemal complex central element protein 1 like
Gene Aliases
MRP2
UniGene
Hs.734779
Gene Chromosome Location
Chr.16: 77199452-77213079 on Build GRCh38
Transcripts
NM_001286639.1 NM_001286640.1 NM_014940.3 NM_001129979.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6581928 c.31G>A p.E11K Chr.16: 77199482-77199482 on Build GRCh38
4129374 c.10A>G p.K4E Chr.16: 77199461-77199461 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs974831608 Chr.16: 77199518-77199518 on Build GRCh38
rs573788823 Chr.16: 77199704-77199704 on Build GRCh38
rs1055717836 Chr.16: 77199486-77199486 on Build GRCh38
rs966668005 Chr.16: 77199430-77199430 on Build GRCh38
rs557773263 Chr.16: 77199630-77199630 on Build GRCh38
rs754218754 Chr.16: 77199510-77199510 on Build GRCh38
rs894013950 Chr.16: 77199655-77199655 on Build GRCh38
rs1040616072 Chr.16: 77199634-77199634 on Build GRCh38
rs899053487 Chr.16: 77199468-77199468 on Build GRCh38
rs563325510 Chr.16: 77199694-77199694 on Build GRCh38
rs927625931 Chr.16: 77199632-77199632 on Build GRCh38
rs951687935 Chr.16: 77199752-77199752 on Build GRCh38
rs920375567 Chr.16: 77199739-77199739 on Build GRCh38
rs1011141686 Chr.16: 77199415-77199415 on Build GRCh38
rs1054849137 Chr.16: 77199642-77199642 on Build GRCh38
rs761238363 Chr.16: 77199503-77199503 on Build GRCh38
rs1046168724 Chr.16: 77199465-77199465 on Build GRCh38
rs370319057 Chr.16: 77199735-77199735 on Build GRCh38
rs770974432 Chr.16: 77199448-77199448 on Build GRCh38
rs983904011 Chr.16: 77199734-77199734 on Build GRCh38
rs1040187050 Chr.16: 77199798-77199798 on Build GRCh38
rs79760539 Chr.16: 77199559-77199559 on Build GRCh38
rs552313156 Chr.16: 77199511-77199511 on Build GRCh38
rs904656183 Chr.16: 77199825-77199825 on Build GRCh38
rs757888203 Chr.16: 77199438-77199438 on Build GRCh38
rs1036440947 Chr.16: 77199466-77199466 on Build GRCh38
rs897979946 Chr.16: 77199885-77199885 on Build GRCh38
rs962114381 Chr.16: 77199547-77199547 on Build GRCh38
rs11544783 Chr.16: 77199548-77199548 on Build GRCh38
rs1042592814 Chr.16: 77199811-77199811 on Build GRCh38
rs966741546 Chr.16: 77199499-77199499 on Build GRCh38
rs1011078003 Chr.16: 77199656-77199656 on Build GRCh38
rs533039786 Chr.16: 77199429-77199429 on Build GRCh38
rs537786854 Chr.16: 77199669-77199669 on Build GRCh38
rs908248831 Chr.16: 77199457-77199457 on Build GRCh38
rs761911708 Chr.16: 77199477-77199477 on Build GRCh38
rs201098769 Chr.16: 77199853-77199853 on Build GRCh38
rs996698742 Chr.16: 77199707-77199707 on Build GRCh38
rs572639117 Chr.16: 77199616-77199616 on Build GRCh38
rs988281089 Chr.16: 77199461-77199461 on Build GRCh38
rs373375255 Chr.16: 77199566-77199566 on Build GRCh38
rs528352318 Chr.16: 77199773-77199773 on Build GRCh38
rs989255565 Chr.16: 77199404-77199404 on Build GRCh38
rs781002276 Chr.16: 77199851-77199851 on Build GRCh38
rs199890788 Chr.16: 77199512-77199512 on Build GRCh38
rs8398 Chr.16: 77199441-77199441 on Build GRCh38
rs183352586 Chr.16: 77199720-77199720 on Build GRCh38
rs944957128 Chr.16: 77199814-77199814 on Build GRCh38
rs1034593868 Chr.16: 77199530-77199530 on Build GRCh38
rs369924114 Chr.16: 77199456-77199456 on Build GRCh38
rs1034564539 Chr.16: 77199701-77199701 on Build GRCh38
rs952570111 Chr.16: 77199712-77199712 on Build GRCh38
rs1014391178 Chr.16: 77199709-77199709 on Build GRCh38
rs931232480 Chr.16: 77199445-77199445 on Build GRCh38
rs535625150 Chr.16: 77199587-77199587 on Build GRCh38
rs756075235 Chr.16: 77199471-77199472 on Build GRCh38
rs772245220 Chr.16: 77199496-77199496 on Build GRCh38
rs754198233 Chr.16: 77199422-77199422 on Build GRCh38
rs893942225 Chr.16: 77199490-77199490 on Build GRCh38
rs533125720 Chr.16: 77199897-77199897 on Build GRCh38
rs76154891 Chr.16: 77199617-77199617 on Build GRCh38
rs946897235 Chr.16: 77199803-77199803 on Build GRCh38
rs958858422 Chr.16: 77199531-77199531 on Build GRCh38
rs1011172908 Chr.16: 77199494-77199494 on Build GRCh38
rs4888607 Chr.16: 77199497-77199497 on Build GRCh38
rs570203837 Chr.16: 77199454-77199454 on Build GRCh38
rs1041809331 Chr.16: 77199657-77199657 on Build GRCh38
rs1020493882 Chr.16: 77199409-77199409 on Build GRCh38
rs1004896846 Chr.16: 77199896-77199896 on Build GRCh38
rs367810281 Chr.16: 77199400-77199400 on Build GRCh38
rs982009994 Chr.16: 77199760-77199760 on Build GRCh38
rs914977692 Chr.16: 77199799-77199799 on Build GRCh38
rs1040621061 Chr.16: 77199839-77199839 on Build GRCh38
rs952540401 Chr.16: 77199543-77199543 on Build GRCh38
rs77430739 Chr.16: 77199505-77199505 on Build GRCh38
rs11544784 Chr.16: 77199433-77199433 on Build GRCh38
rs894702062 Chr.16: 77199862-77199862 on Build GRCh38
rs996115405 Chr.16: 77199867-77199867 on Build GRCh38
rs902658503 Chr.16: 77199813-77199813 on Build GRCh38
rs1020972362 Chr.16: 77199421-77199421 on Build GRCh38
rs112674196 Chr.16: 77199473-77199473 on Build GRCh38
rs117687745 Chr.16: 77199638-77199638 on Build GRCh38
rs945260205 Chr.16: 77199817-77199817 on Build GRCh38
rs141392869 Chr.16: 77199895-77199895 on Build GRCh38
rs961608100 Chr.16: 77199741-77199741 on Build GRCh38
rs1027691791 Chr.16: 77199710-77199710 on Build GRCh38
rs927629928 Chr.16: 77199765-77199765 on Build GRCh38
rs937536329 Chr.16: 77199435-77199435 on Build GRCh38
rs944635901 Chr.16: 77199794-77199794 on Build GRCh38
rs111823947 Chr.16: 77199442-77199442 on Build GRCh38
rs145052441 Chr.16: 77199591-77199591 on Build GRCh38
rs367877599 Chr.16: 77199718-77199718 on Build GRCh38
rs937686082 Chr.16: 77199771-77199771 on Build GRCh38
rs112115473 Chr.16: 77199877-77199877 on Build GRCh38
rs775165602 Chr.16: 77199623-77199623 on Build GRCh38
rs751569005 Chr.16: 77199696-77199696 on Build GRCh38
rs532227355 Chr.16: 77199475-77199475 on Build GRCh38
rs921068696 Chr.16: 77199443-77199443 on Build GRCh38
rs752430911 Chr.16: 77199539-77199539 on Build GRCh38
rs939729932 Chr.16: 77199578-77199578 on Build GRCh38
rs539529755 Chr.16: 77199533-77199533 on Build GRCh38
rs1042686093 Chr.16: 77199666-77199666 on Build GRCh38
rs777479812 Chr.16: 77199562-77199562 on Build GRCh38
rs965171145 Chr.16: 77199516-77199516 on Build GRCh38
rs910434956 Chr.16: 77199774-77199774 on Build GRCh38
rs868466395 Chr.16: 77199682-77199682 on Build GRCh38
rs1041776977 Chr.16: 77199821-77199821 on Build GRCh38
rs114764479 Chr.16: 77199432-77199432 on Build GRCh38
rs745591157 Chr.16: 77199464-77199464 on Build GRCh38

View Full Product Details