Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45151692-45152246 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEXIM1
Entrez Gene ID
10614
Gene Name
hexamethylene bisacetamide inducible 1
Gene Aliases
CLP1, EDG1, HIS1, MAQ1
UniGene
Hs.586945
Gene Chromosome Location
Chr.17: 45147317-45152101 on Build GRCh38
Transcripts
NM_006460.2

SNP Detail

SNP ID Chromosome Location
rs564389311 Chr.17: 45151882-45151882 on Build GRCh38
rs1011282417 Chr.17: 45151770-45151770 on Build GRCh38
rs765506412 Chr.17: 45151952-45151952 on Build GRCh38
rs371346761 Chr.17: 45151831-45151831 on Build GRCh38
rs961790162 Chr.17: 45151996-45151996 on Build GRCh38
rs1014020546 Chr.17: 45151784-45151784 on Build GRCh38
rs928077495 Chr.17: 45152134-45152134 on Build GRCh38
rs552757269 Chr.17: 45151880-45151880 on Build GRCh38
rs1006659247 Chr.17: 45151840-45151840 on Build GRCh38
rs911680364 Chr.17: 45151972-45151972 on Build GRCh38
rs751257872 Chr.17: 45152159-45152159 on Build GRCh38
rs8070447 Chr.17: 45151932-45151932 on Build GRCh38
rs140741522 Chr.17: 45152114-45152114 on Build GRCh38
rs1042800348 Chr.17: 45151807-45151807 on Build GRCh38
rs541309244 Chr.17: 45152053-45152056 on Build GRCh38
rs971769518 Chr.17: 45152045-45152045 on Build GRCh38
rs894323774 Chr.17: 45151783-45151783 on Build GRCh38
rs1028962598 Chr.17: 45151933-45151933 on Build GRCh38
rs538370739 Chr.17: 45152009-45152009 on Build GRCh38
rs984550386 Chr.17: 45152051-45152051 on Build GRCh38
rs868705204 Chr.17: 45152157-45152157 on Build GRCh38
rs563646328 Chr.17: 45151782-45151782 on Build GRCh38
rs969848678 Chr.17: 45152079-45152079 on Build GRCh38
rs139863956 Chr.17: 45152192-45152192 on Build GRCh38
rs986945752 Chr.17: 45151957-45151957 on Build GRCh38
rs952699054 Chr.17: 45152046-45152046 on Build GRCh38
rs192174625 Chr.17: 45151832-45151832 on Build GRCh38
rs542155091 Chr.17: 45151728-45151728 on Build GRCh38
rs947018189 Chr.17: 45152180-45152180 on Build GRCh38
rs767408545 Chr.17: 45152208-45152208 on Build GRCh38
rs533506415 Chr.17: 45151748-45151748 on Build GRCh38
rs764624640 Chr.17: 45151775-45151775 on Build GRCh38
rs150674338 Chr.17: 45152133-45152133 on Build GRCh38
rs879139392 Chr.17: 45151913-45151913 on Build GRCh38
rs535860984 Chr.17: 45152028-45152028 on Build GRCh38
rs994007179 Chr.17: 45151928-45151928 on Build GRCh38
rs78399262 Chr.17: 45152152-45152152 on Build GRCh38
rs28485688 Chr.17: 45151884-45151884 on Build GRCh38
rs546654817 Chr.17: 45151912-45151912 on Build GRCh38
rs891464500 Chr.17: 45151753-45151753 on Build GRCh38
rs935535460 Chr.17: 45151717-45151717 on Build GRCh38
rs556351991 Chr.17: 45152062-45152062 on Build GRCh38
rs1056084943 Chr.17: 45151742-45151742 on Build GRCh38
rs915526129 Chr.17: 45152178-45152178 on Build GRCh38
rs1024367583 Chr.17: 45151797-45151797 on Build GRCh38
rs755634464 Chr.17: 45152043-45152046 on Build GRCh38

View Full Product Details