Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 37363139-37363722 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HKR1
Entrez Gene ID
284459
Gene Name
HKR1, GLI-Kruppel zinc finger family member
Gene Aliases
ZNF875
UniGene
Hs.657027
Gene Chromosome Location
Chr.19: 37317900-37364455 on Build GRCh38
Transcripts
NM_181786.2 XM_017026672.1 XM_017026673.1 XM_017026674.1 XM_017026675.1 XM_017026676.1 XM_017026677.1 XM_017026678.1 XM_017026679.1 XM_017026680.1 XM_017026681.1 XM_017026682.1 XM_017026683.1 XM_017026684.1 XM_017026685.1 XM_017026686.1 XM_017026687.1 XM_017026688.1 XM_017026689.1 XM_017026690.1 XM_017026691.1 XM_017026692.1 XM_017026693.1 XM_017026694.1 XM_017026695.1 XM_017026696.1 XM_017026697.1 XM_017026698.1 XM_017026699.1 XM_017026700.1 XM_017026701.1 XM_017026702.1 XM_017026703.1 XM_017026704.1 XM_017026705.1 XM_017026707.1 XM_017026708.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6680671 c.1466G>A p.R489Q Chr.19: 37363318-37363318 on Build GRCh38
4690594 c.1476delG p.E494fs*19 Chr.19: 37363271-37363271 on Build GRCh38
5069611 c.1418C>T p.T473M Chr.19: 37363270-37363270 on Build GRCh38
4077555 c.1619G>A p.G540D Chr.19: 37363471-37363471 on Build GRCh38
995704 c.1593A>G p.G531G Chr.19: 37363388-37363388 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780521811 Chr.19: 37363311-37363311 on Build GRCh38
rs142002230 Chr.19: 37363516-37363516 on Build GRCh38
rs988666008 Chr.19: 37363290-37363290 on Build GRCh38
rs367884776 Chr.19: 37363352-37363352 on Build GRCh38
rs749379348 Chr.19: 37363256-37363256 on Build GRCh38
rs749339904 Chr.19: 37363316-37363316 on Build GRCh38
rs753375552 Chr.19: 37363181-37363181 on Build GRCh38
rs141902493 Chr.19: 37363371-37363371 on Build GRCh38
rs371013801 Chr.19: 37363607-37363607 on Build GRCh38
rs762646335 Chr.19: 37363364-37363364 on Build GRCh38
rs775240804 Chr.19: 37363601-37363602 on Build GRCh38
rs755794469 Chr.19: 37363599-37363599 on Build GRCh38
rs144607320 Chr.19: 37363443-37363443 on Build GRCh38
rs754974287 Chr.19: 37363665-37363665 on Build GRCh38
rs774237721 Chr.19: 37363525-37363525 on Build GRCh38
rs374237980 Chr.19: 37363401-37363401 on Build GRCh38
rs61742682 Chr.19: 37363437-37363437 on Build GRCh38
rs751142690 Chr.19: 37363462-37363462 on Build GRCh38
rs199577439 Chr.19: 37363593-37363593 on Build GRCh38
rs754628248 Chr.19: 37363185-37363185 on Build GRCh38
rs566316422 Chr.19: 37363616-37363616 on Build GRCh38
rs777120639 Chr.19: 37363573-37363573 on Build GRCh38
rs748797776 Chr.19: 37363551-37363551 on Build GRCh38
rs780161833 Chr.19: 37363494-37363494 on Build GRCh38
rs776575992 Chr.19: 37363345-37363345 on Build GRCh38
rs3745765 Chr.19: 37363333-37363333 on Build GRCh38
rs764086534 Chr.19: 37363187-37363188 on Build GRCh38
rs747196296 Chr.19: 37363508-37363508 on Build GRCh38
rs145361971 Chr.19: 37363544-37363544 on Build GRCh38
rs751550901 Chr.19: 37363248-37363250 on Build GRCh38
rs776165881 Chr.19: 37363615-37363615 on Build GRCh38
rs764221735 Chr.19: 37363580-37363580 on Build GRCh38
rs942550230 Chr.19: 37363315-37363315 on Build GRCh38
rs960694686 Chr.19: 37363189-37363189 on Build GRCh38
rs746212508 Chr.19: 37363309-37363309 on Build GRCh38
rs774572333 Chr.19: 37363261-37363261 on Build GRCh38
rs761056289 Chr.19: 37363480-37363480 on Build GRCh38
rs796090434 Chr.19: 37363577-37363578 on Build GRCh38
rs375294602 Chr.19: 37363318-37363318 on Build GRCh38
rs150608212 Chr.19: 37363295-37363295 on Build GRCh38
rs755956167 Chr.19: 37363652-37363652 on Build GRCh38
rs367650192 Chr.19: 37363598-37363598 on Build GRCh38
rs772409109 Chr.19: 37363578-37363580 on Build GRCh38
rs151159394 Chr.19: 37363670-37363670 on Build GRCh38
rs144017676 Chr.19: 37363337-37363337 on Build GRCh38
rs765937686 Chr.19: 37363176-37363176 on Build GRCh38
rs774772297 Chr.19: 37363576-37363578 on Build GRCh38
rs752317648 Chr.19: 37363357-37363357 on Build GRCh38
rs749172960 Chr.19: 37363601-37363601 on Build GRCh38
rs764668896 Chr.19: 37363623-37363623 on Build GRCh38
rs988174718 Chr.19: 37363202-37363202 on Build GRCh38
rs1020944718 Chr.19: 37363466-37363466 on Build GRCh38
rs141484624 Chr.19: 37363272-37363272 on Build GRCh38
rs146330869 Chr.19: 37363538-37363538 on Build GRCh38
rs372754007 Chr.19: 37363484-37363484 on Build GRCh38
rs771416941 Chr.19: 37363675-37363675 on Build GRCh38
rs769348944 Chr.19: 37363685-37363685 on Build GRCh38
rs775229107 Chr.19: 37363546-37363546 on Build GRCh38
rs147968672 Chr.19: 37363449-37363449 on Build GRCh38
rs138119894 Chr.19: 37363305-37363305 on Build GRCh38
rs761515101 Chr.19: 37363270-37363270 on Build GRCh38
rs755715246 Chr.19: 37363174-37363174 on Build GRCh38
rs749049450 Chr.19: 37363658-37363658 on Build GRCh38
rs551626575 Chr.19: 37363206-37363206 on Build GRCh38
rs752883395 Chr.19: 37363490-37363490 on Build GRCh38
rs757951400 Chr.19: 37363167-37363178 on Build GRCh38
rs201940720 Chr.19: 37363587-37363587 on Build GRCh38
rs373792167 Chr.19: 37363681-37363681 on Build GRCh38
rs763746070 Chr.19: 37363457-37363457 on Build GRCh38
rs374064292 Chr.19: 37363612-37363612 on Build GRCh38
rs370580679 Chr.19: 37363648-37363648 on Build GRCh38
rs773180855 Chr.19: 37363336-37363336 on Build GRCh38
rs774928747 Chr.19: 37363695-37363695 on Build GRCh38
rs183513467 Chr.19: 37363647-37363647 on Build GRCh38
rs567055803 Chr.19: 37363677-37363677 on Build GRCh38
rs755034166 Chr.19: 37363390-37363390 on Build GRCh38
rs373907427 Chr.19: 37363605-37363605 on Build GRCh38
rs771050112 Chr.19: 37363342-37363342 on Build GRCh38
rs954697131 Chr.19: 37363671-37363671 on Build GRCh38
rs753356332 Chr.19: 37363285-37363285 on Build GRCh38
rs779995889 Chr.19: 37363600-37363600 on Build GRCh38
rs766387092 Chr.19: 37363283-37363283 on Build GRCh38
rs150628887 Chr.19: 37363386-37363386 on Build GRCh38
rs375811052 Chr.19: 37363562-37363562 on Build GRCh38
rs764749526 Chr.19: 37363598-37363599 on Build GRCh38
rs964209827 Chr.19: 37363496-37363496 on Build GRCh38
rs749838821 Chr.19: 37363615-37363616 on Build GRCh38
rs759193950 Chr.19: 37363581-37363582 on Build GRCh38
rs148655088 Chr.19: 37363349-37363349 on Build GRCh38
rs752330599 Chr.19: 37363304-37363304 on Build GRCh38
rs761501792 Chr.19: 37363561-37363561 on Build GRCh38
rs779871167 Chr.19: 37363653-37363653 on Build GRCh38
rs769889310 Chr.19: 37363218-37363218 on Build GRCh38
rs748667305 Chr.19: 37363577-37363578 on Build GRCh38
rs755549174 Chr.19: 37363483-37363483 on Build GRCh38
rs768712085 Chr.19: 37363317-37363317 on Build GRCh38
rs764766874 Chr.19: 37363353-37363353 on Build GRCh38
rs532502526 Chr.19: 37363579-37363579 on Build GRCh38
rs746100700 Chr.19: 37363355-37363356 on Build GRCh38
rs746336952 Chr.19: 37363212-37363212 on Build GRCh38
rs968056137 Chr.19: 37363468-37363468 on Build GRCh38
rs754497347 Chr.19: 37363294-37363294 on Build GRCh38
rs757585585 Chr.19: 37363205-37363205 on Build GRCh38
rs746814247 Chr.19: 37363413-37363413 on Build GRCh38
rs754327590 Chr.19: 37363389-37363389 on Build GRCh38
rs201417458 Chr.19: 37363328-37363328 on Build GRCh38
rs767378675 Chr.19: 37363271-37363271 on Build GRCh38
rs1050520680 Chr.19: 37363299-37363299 on Build GRCh38
rs929637623 Chr.19: 37363586-37363586 on Build GRCh38
rs781669442 Chr.19: 37363209-37363209 on Build GRCh38
rs757065327 Chr.19: 37363248-37363251 on Build GRCh38
rs760742788 Chr.19: 37363281-37363281 on Build GRCh38
rs749276780 Chr.19: 37363520-37363521 on Build GRCh38
rs768556501 Chr.19: 37363603-37363603 on Build GRCh38
rs199813545 Chr.19: 37363555-37363555 on Build GRCh38
rs1046697272 Chr.19: 37363589-37363589 on Build GRCh38
rs776241793 Chr.19: 37363573-37363574 on Build GRCh38
rs778623168 Chr.19: 37363190-37363190 on Build GRCh38
rs3745764 Chr.19: 37363678-37363678 on Build GRCh38
rs375688770 Chr.19: 37363522-37363522 on Build GRCh38
rs149184536 Chr.19: 37363576-37363576 on Build GRCh38
rs768814758 Chr.19: 37363257-37363257 on Build GRCh38
rs377589608 Chr.19: 37363630-37363630 on Build GRCh38
rs756333054 Chr.19: 37363467-37363479 on Build GRCh38
rs140257135 Chr.19: 37363672-37363672 on Build GRCh38
rs200083813 Chr.19: 37363680-37363680 on Build GRCh38
rs765480496 Chr.19: 37363614-37363614 on Build GRCh38
rs762432660 Chr.19: 37363456-37363456 on Build GRCh38
rs775598633 Chr.19: 37363220-37363220 on Build GRCh38
rs779097107 Chr.19: 37363400-37363400 on Build GRCh38
rs572154364 Chr.19: 37363410-37363410 on Build GRCh38
rs201964801 Chr.19: 37363306-37363306 on Build GRCh38
rs555809509 Chr.19: 37363684-37363684 on Build GRCh38
rs756409153 Chr.19: 37363375-37363375 on Build GRCh38
rs552933332 Chr.19: 37363482-37363482 on Build GRCh38
rs774012104 Chr.19: 37363560-37363560 on Build GRCh38
rs758494686 Chr.19: 37363499-37363499 on Build GRCh38
rs368777635 Chr.19: 37363402-37363402 on Build GRCh38
rs919476532 Chr.19: 37363564-37363564 on Build GRCh38
rs376866386 Chr.19: 37363606-37363606 on Build GRCh38
rs746467134 Chr.19: 37363507-37363507 on Build GRCh38
rs747392346 Chr.19: 37363191-37363191 on Build GRCh38
rs756677165 Chr.19: 37363310-37363310 on Build GRCh38
rs757542961 Chr.19: 37363591-37363591 on Build GRCh38
rs781191343 Chr.19: 37363302-37363302 on Build GRCh38
rs868053386 Chr.19: 37363227-37363227 on Build GRCh38
rs963514152 Chr.19: 37363696-37363696 on Build GRCh38
rs757418769 Chr.19: 37363241-37363241 on Build GRCh38
rs565301921 Chr.19: 37363575-37363575 on Build GRCh38

View Full Product Details