Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 192797479-192798065 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MB21D2
Entrez Gene ID
151963
Gene Name
Mab-21 domain containing 2
Gene Aliases
C3orf59
UniGene
Hs.151443
Gene Chromosome Location
Chr.3: 192796815-192918161 on Build GRCh38
Transcripts
NM_178496.3

SNP Detail

SNP ID Chromosome Location
rs904014460 Chr.3: 192797806-192797806 on Build GRCh38
rs747430794 Chr.3: 192797508-192797508 on Build GRCh38
rs979813636 Chr.3: 192798011-192798011 on Build GRCh38
rs978037267 Chr.3: 192797543-192797543 on Build GRCh38
rs931756893 Chr.3: 192797597-192797597 on Build GRCh38
rs909707563 Chr.3: 192797572-192797572 on Build GRCh38
rs757160786 Chr.3: 192798010-192798010 on Build GRCh38
rs1036631179 Chr.3: 192797697-192797697 on Build GRCh38
rs778535588 Chr.3: 192797569-192797571 on Build GRCh38
rs764918406 Chr.3: 192797733-192797733 on Build GRCh38
rs887454885 Chr.3: 192797637-192797639 on Build GRCh38
rs921743957 Chr.3: 192797595-192797595 on Build GRCh38
rs1031680579 Chr.3: 192797857-192797857 on Build GRCh38
rs918476528 Chr.3: 192797698-192797698 on Build GRCh38
rs6780822 Chr.3: 192797721-192797721 on Build GRCh38
rs906877208 Chr.3: 192798028-192798028 on Build GRCh38
rs917011255 Chr.3: 192797544-192797544 on Build GRCh38
rs537978273 Chr.3: 192797735-192797735 on Build GRCh38
rs751495229 Chr.3: 192797928-192797928 on Build GRCh38
rs967423562 Chr.3: 192797802-192797802 on Build GRCh38
rs1003061198 Chr.3: 192797701-192797701 on Build GRCh38
rs914612338 Chr.3: 192797503-192797503 on Build GRCh38
rs909759939 Chr.3: 192798023-192798023 on Build GRCh38
rs908928893 Chr.3: 192797963-192797963 on Build GRCh38
rs977796319 Chr.3: 192797821-192797821 on Build GRCh38
rs771471287 Chr.3: 192797516-192797516 on Build GRCh38
rs566775873 Chr.3: 192797688-192797688 on Build GRCh38
rs923830417 Chr.3: 192797551-192797551 on Build GRCh38
rs868354631 Chr.3: 192797756-192797756 on Build GRCh38
rs1049255568 Chr.3: 192797603-192797603 on Build GRCh38
rs933998572 Chr.3: 192797716-192797716 on Build GRCh38
rs140395231 Chr.3: 192797893-192797893 on Build GRCh38
rs1014356625 Chr.3: 192797910-192797910 on Build GRCh38
rs575408556 Chr.3: 192797722-192797722 on Build GRCh38
rs975992290 Chr.3: 192797857-192797857 on Build GRCh38
rs777124063 Chr.3: 192797583-192797583 on Build GRCh38
rs532255315 Chr.3: 192797596-192797596 on Build GRCh38
rs969810531 Chr.3: 192797952-192797952 on Build GRCh38
rs73890330 Chr.3: 192797672-192797672 on Build GRCh38
rs573513924 Chr.3: 192797935-192797935 on Build GRCh38
rs1061860 Chr.3: 192797768-192797768 on Build GRCh38
rs540629586 Chr.3: 192797956-192797956 on Build GRCh38
rs773839694 Chr.3: 192797842-192797842 on Build GRCh38
rs770828402 Chr.3: 192797665-192797665 on Build GRCh38
rs1023909075 Chr.3: 192797931-192797931 on Build GRCh38
rs1019611971 Chr.3: 192797833-192797833 on Build GRCh38
rs546399848 Chr.3: 192797851-192797851 on Build GRCh38
rs147843165 Chr.3: 192797755-192797755 on Build GRCh38
rs992475084 Chr.3: 192797533-192797533 on Build GRCh38
rs10908 Chr.3: 192797967-192797967 on Build GRCh38
rs369621026 Chr.3: 192797581-192797581 on Build GRCh38
rs571375255 Chr.3: 192797983-192797991 on Build GRCh38
rs537113300 Chr.3: 192798012-192798012 on Build GRCh38
rs74624844 Chr.3: 192797547-192797547 on Build GRCh38
rs772630459 Chr.3: 192797776-192797776 on Build GRCh38
rs6780697 Chr.3: 192797564-192797564 on Build GRCh38
rs931765152 Chr.3: 192797859-192797859 on Build GRCh38
rs775081387 Chr.3: 192797759-192797759 on Build GRCh38
rs541797877 Chr.3: 192798036-192798036 on Build GRCh38
rs767355223 Chr.3: 192798025-192798025 on Build GRCh38

View Full Product Details