Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 120456383-120456979 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTBP
Entrez Gene ID
27085
Gene Name
MDM2 binding protein
Gene Aliases
MDM2BP
UniGene
Hs.657656
Gene Chromosome Location
Chr.8: 120445398-120523635 on Build GRCh38
Transcripts
NM_022045.4 XM_011516962.2 XM_011516963.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6895962 c.704C>G p.S235* Chr.8: 120456627-120456627 on Build GRCh38
3663536 c.659C>T p.S220F Chr.8: 120456582-120456582 on Build GRCh38
1623564 c.687C>T p.L229L Chr.8: 120456610-120456610 on Build GRCh38
5835505 c.678_680delAGA p.E227delE Chr.8: 120456601-120456603 on Build GRCh38
7377802 c.736T>C p.W246R Chr.8: 120456659-120456659 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750369375 Chr.8: 120456630-120456630 on Build GRCh38
rs756243593 Chr.8: 120456638-120456638 on Build GRCh38
rs201795878 Chr.8: 120456706-120456706 on Build GRCh38
rs374735189 Chr.8: 120456762-120456762 on Build GRCh38
rs748501231 Chr.8: 120456537-120456537 on Build GRCh38
rs977483908 Chr.8: 120456843-120456843 on Build GRCh38
rs749497320 Chr.8: 120456506-120456506 on Build GRCh38
rs573097647 Chr.8: 120456819-120456819 on Build GRCh38
rs201107399 Chr.8: 120456524-120456524 on Build GRCh38
rs773654116 Chr.8: 120456692-120456692 on Build GRCh38
rs767395595 Chr.8: 120456620-120456620 on Build GRCh38
rs780319687 Chr.8: 120456503-120456503 on Build GRCh38
rs1003130886 Chr.8: 120456742-120456742 on Build GRCh38
rs1008472144 Chr.8: 120456416-120456416 on Build GRCh38
rs756154046 Chr.8: 120456502-120456502 on Build GRCh38
rs780464353 Chr.8: 120456564-120456565 on Build GRCh38
rs373413929 Chr.8: 120456685-120456685 on Build GRCh38
rs750625402 Chr.8: 120456540-120456541 on Build GRCh38
rs554951306 Chr.8: 120456435-120456436 on Build GRCh38
rs371694989 Chr.8: 120456496-120456496 on Build GRCh38
rs979804472 Chr.8: 120456530-120456530 on Build GRCh38
rs976644159 Chr.8: 120456881-120456881 on Build GRCh38
rs777477675 Chr.8: 120456837-120456837 on Build GRCh38
rs913153426 Chr.8: 120456943-120456943 on Build GRCh38
rs1021365489 Chr.8: 120456833-120456833 on Build GRCh38
rs77532040 Chr.8: 120456927-120456927 on Build GRCh38
rs762700935 Chr.8: 120456595-120456595 on Build GRCh38
rs774700329 Chr.8: 120456532-120456532 on Build GRCh38
rs952544197 Chr.8: 120456494-120456494 on Build GRCh38
rs1014963618 Chr.8: 120456726-120456726 on Build GRCh38
rs751277953 Chr.8: 120456577-120456577 on Build GRCh38
rs947058804 Chr.8: 120456454-120456454 on Build GRCh38
rs759038102 Chr.8: 120456558-120456558 on Build GRCh38
rs747567018 Chr.8: 120456693-120456693 on Build GRCh38
rs564040894 Chr.8: 120456722-120456722 on Build GRCh38
rs751333769 Chr.8: 120456604-120456604 on Build GRCh38
rs1026744702 Chr.8: 120456488-120456488 on Build GRCh38
rs199522024 Chr.8: 120456711-120456711 on Build GRCh38
rs371878396 Chr.8: 120456935-120456935 on Build GRCh38
rs897378543 Chr.8: 120456802-120456802 on Build GRCh38
rs542515964 Chr.8: 120456712-120456712 on Build GRCh38
rs749551484 Chr.8: 120456589-120456591 on Build GRCh38
rs769080109 Chr.8: 120456595-120456601 on Build GRCh38
rs914211125 Chr.8: 120456450-120456450 on Build GRCh38
rs779240326 Chr.8: 120456647-120456647 on Build GRCh38
rs1001672348 Chr.8: 120456922-120456926 on Build GRCh38
rs756388976 Chr.8: 120456541-120456541 on Build GRCh38
rs924338629 Chr.8: 120456869-120456869 on Build GRCh38
rs762504842 Chr.8: 120456709-120456709 on Build GRCh38
rs76213660 Chr.8: 120456643-120456643 on Build GRCh38
rs757004357 Chr.8: 120456948-120456948 on Build GRCh38
rs1040851202 Chr.8: 120456657-120456657 on Build GRCh38
rs546557913 Chr.8: 120456926-120456926 on Build GRCh38
rs531000089 Chr.8: 120456729-120456729 on Build GRCh38
rs958875206 Chr.8: 120456775-120456775 on Build GRCh38
rs771501523 Chr.8: 120456702-120456702 on Build GRCh38
rs185275861 Chr.8: 120456730-120456730 on Build GRCh38
rs144931356 Chr.8: 120456941-120456941 on Build GRCh38
rs369863168 Chr.8: 120456648-120456648 on Build GRCh38
rs375131015 Chr.8: 120456569-120456569 on Build GRCh38
rs200194891 Chr.8: 120456543-120456543 on Build GRCh38
rs373848089 Chr.8: 120456707-120456707 on Build GRCh38
rs752715572 Chr.8: 120456625-120456625 on Build GRCh38
rs1027536620 Chr.8: 120456906-120456906 on Build GRCh38
rs748506280 Chr.8: 120456679-120456679 on Build GRCh38
rs945889900 Chr.8: 120456613-120456613 on Build GRCh38
rs952519484 Chr.8: 120456910-120456910 on Build GRCh38
rs752763844 Chr.8: 120456763-120456763 on Build GRCh38
rs139138361 Chr.8: 120456659-120456659 on Build GRCh38
rs745478175 Chr.8: 120456445-120456445 on Build GRCh38
rs369289571 Chr.8: 120456546-120456546 on Build GRCh38
rs991212078 Chr.8: 120456821-120456821 on Build GRCh38
rs528574515 Chr.8: 120456890-120456890 on Build GRCh38
rs1035827371 Chr.8: 120456767-120456767 on Build GRCh38
rs763801842 Chr.8: 120456596-120456596 on Build GRCh38
rs367990727 Chr.8: 120456803-120456803 on Build GRCh38
rs943912130 Chr.8: 120456874-120456874 on Build GRCh38
rs775084504 Chr.8: 120456580-120456580 on Build GRCh38
rs189854594 Chr.8: 120456549-120456549 on Build GRCh38
rs190381381 Chr.8: 120456939-120456939 on Build GRCh38
rs767515188 Chr.8: 120456490-120456490 on Build GRCh38
rs779143464 Chr.8: 120456664-120456664 on Build GRCh38
rs757054093 Chr.8: 120456606-120456606 on Build GRCh38

View Full Product Details