Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 11841664-11842257 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP47
Entrez Gene ID
55031
Gene Name
ubiquitin specific peptidase 47
Gene Aliases
TRFP
UniGene
Hs.577256
Gene Chromosome Location
Chr.11: 11841423-11959325 on Build GRCh38
Transcripts
NM_001282659.1 NM_017944.3 XM_005252997.1 XM_017017953.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6243685 c.13G>T p.E5* Chr.11: 11842198-11842198 on Build GRCh38
1351712 c.12C>T p.G4G Chr.11: 11842197-11842197 on Build GRCh38
5983222 c.39+1G>A p.? Chr.11: 11842225-11842225 on Build GRCh38
5983221 c.39+1G>A p.? Chr.11: 11842225-11842225 on Build GRCh38
6243684 c.13G>T p.E5* Chr.11: 11842198-11842198 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs997324569 Chr.11: 11842194-11842194 on Build GRCh38
rs1013940654 Chr.11: 11841689-11841689 on Build GRCh38
rs967203437 Chr.11: 11842012-11842012 on Build GRCh38
rs948246137 Chr.11: 11842166-11842166 on Build GRCh38
rs944543622 Chr.11: 11841790-11841790 on Build GRCh38
rs115302419 Chr.11: 11841819-11841819 on Build GRCh38
rs896646265 Chr.11: 11841901-11841901 on Build GRCh38
rs1045207873 Chr.11: 11842170-11842170 on Build GRCh38
rs938264716 Chr.11: 11841881-11841881 on Build GRCh38
rs534171969 Chr.11: 11841850-11841850 on Build GRCh38
rs933402584 Chr.11: 11842019-11842019 on Build GRCh38
rs1055276380 Chr.11: 11842067-11842067 on Build GRCh38
rs774900121 Chr.11: 11841885-11841885 on Build GRCh38
rs769737454 Chr.11: 11842206-11842206 on Build GRCh38
rs1027724853 Chr.11: 11841949-11841949 on Build GRCh38
rs773379680 Chr.11: 11841851-11841851 on Build GRCh38
rs771158460 Chr.11: 11842176-11842176 on Build GRCh38
rs867072911 Chr.11: 11842018-11842018 on Build GRCh38
rs965839023 Chr.11: 11841999-11841999 on Build GRCh38
rs113672631 Chr.11: 11841852-11841852 on Build GRCh38
rs913986778 Chr.11: 11841836-11841836 on Build GRCh38
rs925268265 Chr.11: 11842036-11842036 on Build GRCh38
rs993146196 Chr.11: 11841943-11841943 on Build GRCh38
rs918188475 Chr.11: 11842091-11842091 on Build GRCh38
rs776472757 Chr.11: 11842191-11842191 on Build GRCh38
rs762397774 Chr.11: 11842235-11842235 on Build GRCh38
rs1048863905 Chr.11: 11842145-11842145 on Build GRCh38
rs1008547295 Chr.11: 11841853-11841853 on Build GRCh38
rs1027534582 Chr.11: 11841738-11841738 on Build GRCh38
rs952961377 Chr.11: 11841707-11841707 on Build GRCh38
rs1017509239 Chr.11: 11842171-11842171 on Build GRCh38
rs970715364 Chr.11: 11841893-11841893 on Build GRCh38
rs1007245456 Chr.11: 11842154-11842154 on Build GRCh38
rs1037616003 Chr.11: 11842108-11842108 on Build GRCh38
rs781621325 Chr.11: 11842139-11842139 on Build GRCh38
rs989754975 Chr.11: 11841934-11841934 on Build GRCh38
rs1055327300 Chr.11: 11841890-11841890 on Build GRCh38
rs952846055 Chr.11: 11841888-11841888 on Build GRCh38
rs755273076 Chr.11: 11842138-11842138 on Build GRCh38
rs1050300687 Chr.11: 11841846-11841846 on Build GRCh38
rs779222167 Chr.11: 11842185-11842185 on Build GRCh38
rs569690745 Chr.11: 11841823-11841823 on Build GRCh38
rs377184530 Chr.11: 11842152-11842152 on Build GRCh38
rs368586788 Chr.11: 11841939-11841939 on Build GRCh38
rs773163820 Chr.11: 11842216-11842216 on Build GRCh38
rs1044117900 Chr.11: 11841861-11841861 on Build GRCh38
rs762046586 Chr.11: 11841907-11841907 on Build GRCh38
rs1023818937 Chr.11: 11841892-11841892 on Build GRCh38
rs537769822 Chr.11: 11841768-11841768 on Build GRCh38
rs982121122 Chr.11: 11841895-11841895 on Build GRCh38
rs572493763 Chr.11: 11842006-11842006 on Build GRCh38
rs976339109 Chr.11: 11842000-11842000 on Build GRCh38
rs746093600 Chr.11: 11842189-11842189 on Build GRCh38
rs879556356 Chr.11: 11841990-11841991 on Build GRCh38
rs12418935 Chr.11: 11841863-11841863 on Build GRCh38
rs961708523 Chr.11: 11841713-11841713 on Build GRCh38
rs1023869678 Chr.11: 11842007-11842007 on Build GRCh38
rs972161425 Chr.11: 11841723-11841723 on Build GRCh38
rs28364721 Chr.11: 11841979-11841979 on Build GRCh38
rs966358835 Chr.11: 11842175-11842175 on Build GRCh38
rs994780635 Chr.11: 11841876-11841876 on Build GRCh38
rs951953070 Chr.11: 11841746-11841746 on Build GRCh38
rs556408401 Chr.11: 11841929-11841929 on Build GRCh38
rs772313350 Chr.11: 11841813-11841813 on Build GRCh38
rs911784925 Chr.11: 11841954-11841954 on Build GRCh38
rs916586377 Chr.11: 11841745-11841745 on Build GRCh38
rs1024037697 Chr.11: 11841927-11841927 on Build GRCh38
rs915086091 Chr.11: 11841940-11841940 on Build GRCh38
rs944400424 Chr.11: 11841972-11841972 on Build GRCh38
rs574518727 Chr.11: 11842153-11842153 on Build GRCh38
rs565756857 Chr.11: 11841838-11841838 on Build GRCh38
rs761562169 Chr.11: 11842198-11842198 on Build GRCh38
rs924044903 Chr.11: 11841904-11841904 on Build GRCh38
rs1030995211 Chr.11: 11842013-11842013 on Build GRCh38
rs558673933 Chr.11: 11841998-11841998 on Build GRCh38
rs1027671044 Chr.11: 11841703-11841703 on Build GRCh38
rs749987996 Chr.11: 11842157-11842157 on Build GRCh38
rs990812016 Chr.11: 11841709-11841709 on Build GRCh38
rs1024504526 Chr.11: 11841692-11841692 on Build GRCh38
rs944630338 Chr.11: 11842085-11842085 on Build GRCh38
rs953208090 Chr.11: 11842035-11842035 on Build GRCh38
rs993649049 Chr.11: 11842130-11842130 on Build GRCh38
rs911837443 Chr.11: 11842072-11842072 on Build GRCh38
rs910382137 Chr.11: 11841789-11841789 on Build GRCh38
rs536976841 Chr.11: 11841864-11841864 on Build GRCh38
rs930278889 Chr.11: 11842128-11842128 on Build GRCh38
rs1014640179 Chr.11: 11841690-11841690 on Build GRCh38
rs552333899 Chr.11: 11841831-11841831 on Build GRCh38
rs374241185 Chr.11: 11842147-11842147 on Build GRCh38
rs938043575 Chr.11: 11842053-11842053 on Build GRCh38
rs935189452 Chr.11: 11841817-11841817 on Build GRCh38
rs538627749 Chr.11: 11841990-11841990 on Build GRCh38
rs887427305 Chr.11: 11842149-11842149 on Build GRCh38
rs897573594 Chr.11: 11841941-11841941 on Build GRCh38
rs897602722 Chr.11: 11841982-11841982 on Build GRCh38
rs541126107 Chr.11: 11842020-11842020 on Build GRCh38
rs369504560 Chr.11: 11842125-11842125 on Build GRCh38
rs532110346 Chr.11: 11841824-11841824 on Build GRCh38
rs913948508 Chr.11: 11842011-11842011 on Build GRCh38
rs749342378 Chr.11: 11842173-11842173 on Build GRCh38
rs888632397 Chr.11: 11841991-11841993 on Build GRCh38
rs772250799 Chr.11: 11842190-11842190 on Build GRCh38
rs949542193 Chr.11: 11842101-11842101 on Build GRCh38
rs112339156 Chr.11: 11841967-11841967 on Build GRCh38

View Full Product Details