Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 15497106-15497677 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPRO
Entrez Gene ID
5800
Gene Name
protein tyrosine phosphatase, receptor type O
Gene Aliases
GLEPP1, NPHS6, PTP-OC, PTP-U2, PTPROT, PTPU2, R-PTP-O
UniGene
Hs.160871
Gene Chromosome Location
Chr.12: 15322257-15598331 on Build GRCh38
Transcripts
NM_002848.3 NM_030667.2 XM_017019725.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
937596 c.447T>G p.V149V Chr.12: 15497342-15497342 on Build GRCh38
937595 c.395C>G p.S132C Chr.12: 15497290-15497290 on Build GRCh38
140685 c.490C>T p.R164W Chr.12: 15497385-15497385 on Build GRCh38
383093 c.452C>A p.T151K Chr.12: 15497347-15497347 on Build GRCh38
284390 c.412C>A p.L138M Chr.12: 15497307-15497307 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs890448236 Chr.12: 15497303-15497303 on Build GRCh38
rs945265178 Chr.12: 15497593-15497593 on Build GRCh38
rs1001630924 Chr.12: 15497523-15497523 on Build GRCh38
rs753254239 Chr.12: 15497443-15497443 on Build GRCh38
rs1034312888 Chr.12: 15497557-15497557 on Build GRCh38
rs376998542 Chr.12: 15497476-15497476 on Build GRCh38
rs376857700 Chr.12: 15497392-15497392 on Build GRCh38
rs980454035 Chr.12: 15497507-15497507 on Build GRCh38
rs138770041 Chr.12: 15497422-15497422 on Build GRCh38
rs528219515 Chr.12: 15497520-15497520 on Build GRCh38
rs770458073 Chr.12: 15497247-15497247 on Build GRCh38
rs745367539 Chr.12: 15497258-15497258 on Build GRCh38
rs199522094 Chr.12: 15497204-15497204 on Build GRCh38
rs571289140 Chr.12: 15497548-15497548 on Build GRCh38
rs993081955 Chr.12: 15497301-15497301 on Build GRCh38
rs750721345 Chr.12: 15497307-15497307 on Build GRCh38
rs901060336 Chr.12: 15497160-15497160 on Build GRCh38
rs770002949 Chr.12: 15497445-15497445 on Build GRCh38
rs761932153 Chr.12: 15497231-15497236 on Build GRCh38
rs779442056 Chr.12: 15497207-15497207 on Build GRCh38
rs561205461 Chr.12: 15497142-15497142 on Build GRCh38
rs1011178806 Chr.12: 15497463-15497463 on Build GRCh38
rs762901864 Chr.12: 15497261-15497261 on Build GRCh38
rs538366251 Chr.12: 15497592-15497592 on Build GRCh38
rs764129374 Chr.12: 15497193-15497193 on Build GRCh38
rs540081228 Chr.12: 15497260-15497260 on Build GRCh38
rs758633834 Chr.12: 15497444-15497444 on Build GRCh38
rs998108987 Chr.12: 15497165-15497165 on Build GRCh38
rs746632844 Chr.12: 15497231-15497231 on Build GRCh38
rs761511360 Chr.12: 15497419-15497419 on Build GRCh38
rs372239091 Chr.12: 15497383-15497383 on Build GRCh38
rs774407287 Chr.12: 15497411-15497411 on Build GRCh38
rs910058875 Chr.12: 15497656-15497656 on Build GRCh38
rs769300317 Chr.12: 15497259-15497259 on Build GRCh38
rs746543150 Chr.12: 15497425-15497425 on Build GRCh38
rs951682011 Chr.12: 15497634-15497634 on Build GRCh38
rs775013254 Chr.12: 15497385-15497385 on Build GRCh38
rs368339318 Chr.12: 15497319-15497319 on Build GRCh38
rs780700188 Chr.12: 15497198-15497198 on Build GRCh38
rs779603502 Chr.12: 15497345-15497345 on Build GRCh38
rs528278152 Chr.12: 15497191-15497191 on Build GRCh38
rs758853940 Chr.12: 15497367-15497367 on Build GRCh38
rs77846183 Chr.12: 15497226-15497226 on Build GRCh38
rs767624514 Chr.12: 15497306-15497306 on Build GRCh38
rs989271085 Chr.12: 15497562-15497562 on Build GRCh38
rs575611533 Chr.12: 15497263-15497263 on Build GRCh38
rs960066655 Chr.12: 15497589-15497589 on Build GRCh38
rs761719227 Chr.12: 15497302-15497302 on Build GRCh38
rs926334675 Chr.12: 15497521-15497521 on Build GRCh38
rs748436787 Chr.12: 15497358-15497358 on Build GRCh38
rs867253741 Chr.12: 15497591-15497591 on Build GRCh38
rs879787029 Chr.12: 15497473-15497476 on Build GRCh38
rs770282329 Chr.12: 15497480-15497481 on Build GRCh38
rs551711163 Chr.12: 15497328-15497328 on Build GRCh38
rs1025876669 Chr.12: 15497605-15497605 on Build GRCh38
rs958016232 Chr.12: 15497551-15497551 on Build GRCh38
rs760970168 Chr.12: 15497428-15497428 on Build GRCh38
rs138083650 Chr.12: 15497380-15497380 on Build GRCh38
rs757323345 Chr.12: 15497190-15497190 on Build GRCh38
rs553313937 Chr.12: 15497546-15497546 on Build GRCh38
rs549256983 Chr.12: 15497429-15497429 on Build GRCh38
rs569717831 Chr.12: 15497397-15497397 on Build GRCh38
rs746769371 Chr.12: 15497654-15497654 on Build GRCh38
rs980484819 Chr.12: 15497240-15497240 on Build GRCh38
rs780317142 Chr.12: 15497435-15497435 on Build GRCh38
rs774106115 Chr.12: 15497274-15497274 on Build GRCh38
rs933893435 Chr.12: 15497433-15497433 on Build GRCh38
rs1022144797 Chr.12: 15497471-15497471 on Build GRCh38
rs776393054 Chr.12: 15497254-15497254 on Build GRCh38
rs956565365 Chr.12: 15497182-15497182 on Build GRCh38
rs141410112 Chr.12: 15497286-15497286 on Build GRCh38
rs201529265 Chr.12: 15497321-15497321 on Build GRCh38
rs759737379 Chr.12: 15497438-15497438 on Build GRCh38
rs555785100 Chr.12: 15497201-15497201 on Build GRCh38
rs140315694 Chr.12: 15497479-15497479 on Build GRCh38
rs371687990 Chr.12: 15497439-15497439 on Build GRCh38
rs754327869 Chr.12: 15497177-15497177 on Build GRCh38
rs556871126 Chr.12: 15497649-15497649 on Build GRCh38
rs1051597556 Chr.12: 15497622-15497622 on Build GRCh38
rs780643607 Chr.12: 15497376-15497376 on Build GRCh38
rs755573133 Chr.12: 15497340-15497340 on Build GRCh38
rs983868771 Chr.12: 15497386-15497386 on Build GRCh38
rs548002612 Chr.12: 15497602-15497602 on Build GRCh38
rs145570875 Chr.12: 15497312-15497312 on Build GRCh38

View Full Product Details