Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 160884795-160885376 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITLN1
Entrez Gene ID
55600
Gene Name
intelectin 1
Gene Aliases
HL-1, HL1, INTL, ITLN, LFR, hIntL, omentin
UniGene
Hs.50813
Gene Chromosome Location
Chr.1: 160876540-160885170 on Build GRCh38
Transcripts
NM_017625.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3477056 c.18C>T p.F6F Chr.1: 160884860-160884860 on Build GRCh38
6541134 c.35C>A p.A12E Chr.1: 160884843-160884843 on Build GRCh38
3385348 c.35C>T p.A12V Chr.1: 160884843-160884843 on Build GRCh38
7190304 c.58+9C>A p.? Chr.1: 160884811-160884811 on Build GRCh38
7112001 c.30C>T p.L10L Chr.1: 160884848-160884848 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1003763089 Chr.1: 160885278-160885278 on Build GRCh38
rs747105662 Chr.1: 160884889-160884889 on Build GRCh38
rs986101486 Chr.1: 160884911-160884911 on Build GRCh38
rs201799346 Chr.1: 160884843-160884843 on Build GRCh38
rs978598642 Chr.1: 160885323-160885323 on Build GRCh38
rs948950695 Chr.1: 160885025-160885025 on Build GRCh38
rs750348147 Chr.1: 160884855-160884855 on Build GRCh38
rs184899158 Chr.1: 160885347-160885347 on Build GRCh38
rs965977621 Chr.1: 160885266-160885266 on Build GRCh38
rs201053875 Chr.1: 160884881-160884881 on Build GRCh38
rs749033434 Chr.1: 160884875-160884876 on Build GRCh38
rs772095207 Chr.1: 160884828-160884828 on Build GRCh38
rs746410437 Chr.1: 160884879-160884879 on Build GRCh38
rs1055029740 Chr.1: 160884981-160884981 on Build GRCh38
rs894677141 Chr.1: 160885012-160885012 on Build GRCh38
rs758380112 Chr.1: 160884858-160884858 on Build GRCh38
rs112656766 Chr.1: 160885146-160885146 on Build GRCh38
rs878953426 Chr.1: 160885136-160885136 on Build GRCh38
rs544121078 Chr.1: 160885028-160885028 on Build GRCh38
rs772608692 Chr.1: 160884880-160884880 on Build GRCh38
rs74315633 Chr.1: 160884913-160884913 on Build GRCh38
rs372445635 Chr.1: 160885334-160885334 on Build GRCh38
rs749389758 Chr.1: 160885089-160885089 on Build GRCh38
rs371815410 Chr.1: 160885077-160885077 on Build GRCh38
rs1014572842 Chr.1: 160885127-160885127 on Build GRCh38
rs764100689 Chr.1: 160884847-160884847 on Build GRCh38
rs139267617 Chr.1: 160884877-160884877 on Build GRCh38
rs765290429 Chr.1: 160884848-160884848 on Build GRCh38
rs191377321 Chr.1: 160884920-160884920 on Build GRCh38
rs771225454 Chr.1: 160884912-160884912 on Build GRCh38
rs922615029 Chr.1: 160885088-160885088 on Build GRCh38
rs936062363 Chr.1: 160885287-160885287 on Build GRCh38
rs41266939 Chr.1: 160885282-160885282 on Build GRCh38
rs377032743 Chr.1: 160885357-160885357 on Build GRCh38
rs77824633 Chr.1: 160884959-160884959 on Build GRCh38
rs754657259 Chr.1: 160884870-160884870 on Build GRCh38
rs777681746 Chr.1: 160884873-160884873 on Build GRCh38
rs1033819124 Chr.1: 160884952-160884952 on Build GRCh38
rs916024583 Chr.1: 160885319-160885319 on Build GRCh38
rs970985392 Chr.1: 160884930-160884930 on Build GRCh38
rs1011069748 Chr.1: 160885295-160885295 on Build GRCh38
rs961824941 Chr.1: 160884838-160884838 on Build GRCh38
rs944706889 Chr.1: 160884931-160884931 on Build GRCh38
rs527357763 Chr.1: 160885166-160885166 on Build GRCh38
rs764363137 Chr.1: 160884840-160884840 on Build GRCh38
rs533679017 Chr.1: 160885020-160885020 on Build GRCh38
rs878903652 Chr.1: 160885060-160885060 on Build GRCh38
rs570269927 Chr.1: 160885221-160885221 on Build GRCh38
rs1047368782 Chr.1: 160885253-160885253 on Build GRCh38
rs1046377436 Chr.1: 160885339-160885339 on Build GRCh38
rs537963511 Chr.1: 160884957-160884957 on Build GRCh38
rs79209815 Chr.1: 160885137-160885137 on Build GRCh38
rs953248135 Chr.1: 160885169-160885169 on Build GRCh38
rs761632239 Chr.1: 160884846-160884846 on Build GRCh38
rs35779394 Chr.1: 160884830-160884830 on Build GRCh38
rs547191896 Chr.1: 160885174-160885174 on Build GRCh38
rs780834029 Chr.1: 160884872-160884872 on Build GRCh38
rs879248612 Chr.1: 160885057-160885057 on Build GRCh38
rs1022913152 Chr.1: 160885301-160885301 on Build GRCh38
rs150060331 Chr.1: 160885086-160885086 on Build GRCh38
rs760854751 Chr.1: 160884832-160884832 on Build GRCh38
rs765857065 Chr.1: 160884864-160884864 on Build GRCh38
rs749669242 Chr.1: 160884875-160884875 on Build GRCh38
rs542469872 Chr.1: 160885214-160885214 on Build GRCh38
rs1003177103 Chr.1: 160885096-160885096 on Build GRCh38
rs367626564 Chr.1: 160885048-160885048 on Build GRCh38
rs766340485 Chr.1: 160884928-160884928 on Build GRCh38
rs765163926 Chr.1: 160884910-160884910 on Build GRCh38
rs878975512 Chr.1: 160885058-160885059 on Build GRCh38
rs763046109 Chr.1: 160884926-160884926 on Build GRCh38
rs541657124 Chr.1: 160884958-160884958 on Build GRCh38
rs768763855 Chr.1: 160884893-160884893 on Build GRCh38
rs990257693 Chr.1: 160885288-160885288 on Build GRCh38
rs570435172 Chr.1: 160884876-160884876 on Build GRCh38
rs745966521 Chr.1: 160884827-160884827 on Build GRCh38
rs959650138 Chr.1: 160885291-160885291 on Build GRCh38
rs367996311 Chr.1: 160884932-160884932 on Build GRCh38
rs2297560 Chr.1: 160884908-160884908 on Build GRCh38
rs558790888 Chr.1: 160885277-160885277 on Build GRCh38
rs751071494 Chr.1: 160884869-160884869 on Build GRCh38

View Full Product Details