Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57756205-57756798 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF776
Entrez Gene ID
284309
Gene Name
zinc finger protein 776
Gene Aliases
-
UniGene
Hs.720254
Gene Chromosome Location
Chr.19: 57746796-57758159 on Build GRCh38
Transcripts
NM_173632.3

SNP Detail

SNP ID Chromosome Location
rs765565683 Chr.19: 57756640-57756640 on Build GRCh38
rs558214783 Chr.19: 57756358-57756358 on Build GRCh38
rs915368202 Chr.19: 57756439-57756439 on Build GRCh38
rs1029636385 Chr.19: 57756393-57756393 on Build GRCh38
rs557116580 Chr.19: 57756458-57756458 on Build GRCh38
rs769145468 Chr.19: 57756310-57756310 on Build GRCh38
rs1056791875 Chr.19: 57756566-57756566 on Build GRCh38
rs996968993 Chr.19: 57756529-57756529 on Build GRCh38
rs984776415 Chr.19: 57756605-57756605 on Build GRCh38
rs573955064 Chr.19: 57756670-57756670 on Build GRCh38
rs978599821 Chr.19: 57756518-57756518 on Build GRCh38
rs773628244 Chr.19: 57756659-57756659 on Build GRCh38
rs199744931 Chr.19: 57756277-57756277 on Build GRCh38
rs949514291 Chr.19: 57756706-57756706 on Build GRCh38
rs190953239 Chr.19: 57756756-57756756 on Build GRCh38
rs368418857 Chr.19: 57756261-57756261 on Build GRCh38
rs187879974 Chr.19: 57756676-57756676 on Build GRCh38
rs964269981 Chr.19: 57756315-57756315 on Build GRCh38
rs780119521 Chr.19: 57756624-57756624 on Build GRCh38
rs375325856 Chr.19: 57756591-57756591 on Build GRCh38
rs1005756817 Chr.19: 57756285-57756285 on Build GRCh38
rs1042583555 Chr.19: 57756346-57756346 on Build GRCh38
rs956785018 Chr.19: 57756413-57756413 on Build GRCh38
rs777039754 Chr.19: 57756396-57756396 on Build GRCh38
rs972648791 Chr.19: 57756565-57756565 on Build GRCh38
rs374316381 Chr.19: 57756275-57756275 on Build GRCh38
rs888236009 Chr.19: 57756594-57756594 on Build GRCh38
rs929679780 Chr.19: 57756582-57756582 on Build GRCh38
rs537449279 Chr.19: 57756435-57756435 on Build GRCh38
rs927156985 Chr.19: 57756542-57756542 on Build GRCh38
rs918085937 Chr.19: 57756677-57756677 on Build GRCh38
rs763158286 Chr.19: 57756735-57756735 on Build GRCh38
rs1055682956 Chr.19: 57756273-57756273 on Build GRCh38
rs998101615 Chr.19: 57756370-57756370 on Build GRCh38
rs762426096 Chr.19: 57756569-57756569 on Build GRCh38
rs866678035 Chr.19: 57756347-57756347 on Build GRCh38
rs148832270 Chr.19: 57756761-57756761 on Build GRCh38
rs375237952 Chr.19: 57756644-57756644 on Build GRCh38
rs1019858494 Chr.19: 57756481-57756481 on Build GRCh38
rs11290757 Chr.19: 57756277-57756277 on Build GRCh38
rs578053314 Chr.19: 57756395-57756395 on Build GRCh38
rs938221635 Chr.19: 57756265-57756265 on Build GRCh38
rs1032081809 Chr.19: 57756385-57756385 on Build GRCh38
rs371940815 Chr.19: 57756324-57756324 on Build GRCh38
rs756398308 Chr.19: 57756442-57756442 on Build GRCh38
rs937261811 Chr.19: 57756557-57756557 on Build GRCh38

View Full Product Details