Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 70212615-70213203 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TIA1
Entrez Gene ID
7072
Gene Name
TIA1 cytotoxic granule-associated RNA binding protein
Gene Aliases
TIA-1, WDM
UniGene
Hs.413123
Gene Chromosome Location
Chr.2: 70209444-70248793 on Build GRCh38
Transcripts
NM_022037.2 NM_022173.2 XM_005264527.1 XM_005264528.4 XM_005264531.1 XM_011533077.1 XM_011533078.1 XM_011533081.2 XM_011533082.2 XM_017004789.1 XM_017004790.1 XM_017004791.1 XM_017004792.1 XM_017004793.1 XM_017004794.1 XM_017004795.1 XM_017004796.1 XM_017004797.1 XM_017004798.1 XM_017004799.1 XM_017004800.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1669003 c.1150G>A p.E384K Chr.2: 70212730-70212730 on Build GRCh38
3695577 c.1070A>G p.N357S Chr.2: 70212810-70212810 on Build GRCh38
5621996 c.1148A>G p.Y383C Chr.2: 70212732-70212732 on Build GRCh38
6579676 c.1150G>C p.E384Q Chr.2: 70212730-70212730 on Build GRCh38
1483211 c.1059G>C p.W353C Chr.2: 70212821-70212821 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765460029 Chr.2: 70212686-70212686 on Build GRCh38
rs71414814 Chr.2: 70212918-70212918 on Build GRCh38
rs113167846 Chr.2: 70212893-70212894 on Build GRCh38
rs926676952 Chr.2: 70213084-70213084 on Build GRCh38
rs768554955 Chr.2: 70212739-70212739 on Build GRCh38
rs765281803 Chr.2: 70212898-70212902 on Build GRCh38
rs752073368 Chr.2: 70212880-70212880 on Build GRCh38
rs116621885 Chr.2: 70212810-70212810 on Build GRCh38
rs954645706 Chr.2: 70213179-70213179 on Build GRCh38
rs202071613 Chr.2: 70212680-70212680 on Build GRCh38
rs912090965 Chr.2: 70213017-70213017 on Build GRCh38
rs754801947 Chr.2: 70213008-70213008 on Build GRCh38
rs771864817 Chr.2: 70212814-70212814 on Build GRCh38
rs1028042053 Chr.2: 70213000-70213000 on Build GRCh38
rs939446117 Chr.2: 70213109-70213109 on Build GRCh38
rs766155218 Chr.2: 70212675-70212675 on Build GRCh38
rs572347540 Chr.2: 70212948-70212948 on Build GRCh38
rs71414815 Chr.2: 70212927-70212927 on Build GRCh38
rs762305513 Chr.2: 70212885-70212885 on Build GRCh38
rs72902461 Chr.2: 70212794-70212794 on Build GRCh38
rs781188920 Chr.2: 70212797-70212797 on Build GRCh38
rs1045149836 Chr.2: 70213083-70213083 on Build GRCh38
rs753303341 Chr.2: 70212689-70212689 on Build GRCh38
rs7591895 Chr.2: 70213134-70213134 on Build GRCh38
rs1032271334 Chr.2: 70213096-70213096 on Build GRCh38
rs912146134 Chr.2: 70212782-70212782 on Build GRCh38
rs199700349 Chr.2: 70212723-70212723 on Build GRCh38
rs764374187 Chr.2: 70212877-70212877 on Build GRCh38
rs1017008375 Chr.2: 70212934-70212934 on Build GRCh38
rs145782010 Chr.2: 70212912-70212912 on Build GRCh38
rs893604269 Chr.2: 70213114-70213114 on Build GRCh38
rs765923218 Chr.2: 70212755-70212755 on Build GRCh38
rs534696291 Chr.2: 70212756-70212756 on Build GRCh38
rs776571655 Chr.2: 70212893-70212897 on Build GRCh38
rs757277744 Chr.2: 70212732-70212732 on Build GRCh38
rs368330131 Chr.2: 70212858-70212858 on Build GRCh38
rs760297134 Chr.2: 70212749-70212749 on Build GRCh38
rs202093855 Chr.2: 70212852-70212852 on Build GRCh38
rs368903273 Chr.2: 70212890-70212890 on Build GRCh38
rs372889101 Chr.2: 70212817-70212817 on Build GRCh38
rs759094935 Chr.2: 70212757-70212757 on Build GRCh38
rs367908555 Chr.2: 70212671-70212671 on Build GRCh38
rs542190157 Chr.2: 70212857-70212857 on Build GRCh38
rs757332023 Chr.2: 70212795-70212795 on Build GRCh38
rs774014016 Chr.2: 70212744-70212744 on Build GRCh38
rs151174422 Chr.2: 70213077-70213077 on Build GRCh38
rs1016630302 Chr.2: 70212672-70212672 on Build GRCh38
rs752969561 Chr.2: 70212772-70212772 on Build GRCh38
rs375719059 Chr.2: 70213135-70213135 on Build GRCh38
rs556545503 Chr.2: 70212798-70212798 on Build GRCh38
rs371477184 Chr.2: 70212978-70212978 on Build GRCh38
rs761639780 Chr.2: 70212894-70212896 on Build GRCh38
rs915195272 Chr.2: 70213068-70213068 on Build GRCh38
rs533119744 Chr.2: 70213035-70213035 on Build GRCh38
rs767625458 Chr.2: 70212888-70212888 on Build GRCh38
rs999416646 Chr.2: 70213081-70213081 on Build GRCh38
rs529184118 Chr.2: 70213125-70213125 on Build GRCh38
rs1014671596 Chr.2: 70212678-70212678 on Build GRCh38
rs763258598 Chr.2: 70212874-70212874 on Build GRCh38
rs748157704 Chr.2: 70212745-70212745 on Build GRCh38
rs895926303 Chr.2: 70213043-70213043 on Build GRCh38
rs563557906 Chr.2: 70213122-70213122 on Build GRCh38
rs746887914 Chr.2: 70212819-70212819 on Build GRCh38
rs1042136180 Chr.2: 70213066-70213066 on Build GRCh38
rs777946206 Chr.2: 70212726-70212726 on Build GRCh38
rs957176939 Chr.2: 70213036-70213036 on Build GRCh38
rs953520941 Chr.2: 70213002-70213002 on Build GRCh38
rs201905164 Chr.2: 70212802-70212802 on Build GRCh38
rs200990761 Chr.2: 70212779-70212779 on Build GRCh38
rs200542970 Chr.2: 70212733-70212733 on Build GRCh38
rs937518391 Chr.2: 70213011-70213012 on Build GRCh38
rs772046624 Chr.2: 70212805-70212806 on Build GRCh38
rs778817360 Chr.2: 70212803-70212803 on Build GRCh38
rs745901576 Chr.2: 70212686-70212687 on Build GRCh38
rs541087717 Chr.2: 70212952-70212952 on Build GRCh38
rs746021823 Chr.2: 70212734-70212734 on Build GRCh38
rs988415613 Chr.2: 70213051-70213051 on Build GRCh38
rs940962406 Chr.2: 70212917-70212919 on Build GRCh38
rs950070651 Chr.2: 70213061-70213061 on Build GRCh38
rs773238298 Chr.2: 70212747-70212747 on Build GRCh38
rs1056323243 Chr.2: 70212870-70212870 on Build GRCh38
rs935618716 Chr.2: 70212784-70212784 on Build GRCh38
rs996518250 Chr.2: 70212995-70212995 on Build GRCh38
rs372228986 Chr.2: 70212859-70212859 on Build GRCh38
rs76710635 Chr.2: 70212758-70212758 on Build GRCh38
rs199993142 Chr.2: 70212873-70212873 on Build GRCh38
rs747068278 Chr.2: 70212730-70212730 on Build GRCh38
rs974458784 Chr.2: 70212843-70212843 on Build GRCh38
rs879881587 Chr.2: 70212913-70212913 on Build GRCh38
rs999343044 Chr.2: 70212777-70212777 on Build GRCh38

View Full Product Details