Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 181558445-181558883 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CERKL
Entrez Gene ID
375298
Gene Name
ceramide kinase like
Gene Aliases
RP26
UniGene
Hs.732358
Gene Chromosome Location
Chr.2: 181536672-181657107 on Build GRCh38
Transcripts
NM_001030311.2 NM_001030312.2 NM_001030313.2 NM_001160277.1 NM_201548.4 NR_027689.1 NR_027690.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6627321 c.779G>T p.G260V Chr.2: 181558685-181558685 on Build GRCh38
4510941 c.852C>T p.I284I Chr.2: 181558612-181558612 on Build GRCh38
6627322 c.701G>T p.G234V Chr.2: 181558685-181558685 on Build GRCh38
5697528 c.789C>T p.S263S Chr.2: 181558675-181558675 on Build GRCh38
1148861 c.884G>C p.G295A Chr.2: 181558580-181558580 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576790010 Chr.2: 181558506-181558506 on Build GRCh38
rs968625844 Chr.2: 181558789-181558789 on Build GRCh38
rs747702553 Chr.2: 181558545-181558545 on Build GRCh38
rs751149105 Chr.2: 181558628-181558628 on Build GRCh38
rs1022225994 Chr.2: 181558782-181558782 on Build GRCh38
rs1047916483 Chr.2: 181558649-181558649 on Build GRCh38
rs946831250 Chr.2: 181558492-181558492 on Build GRCh38
rs145515805 Chr.2: 181558752-181558752 on Build GRCh38
rs750466083 Chr.2: 181558607-181558607 on Build GRCh38
rs534791684 Chr.2: 181558738-181558738 on Build GRCh38
rs532973636 Chr.2: 181558674-181558674 on Build GRCh38
rs199796256 Chr.2: 181558563-181558563 on Build GRCh38
rs786205545 Chr.2: 181558574-181558574 on Build GRCh38
rs398122964 Chr.2: 181558606-181558606 on Build GRCh38
rs560118661 Chr.2: 181558720-181558720 on Build GRCh38
rs761420671 Chr.2: 181558484-181558484 on Build GRCh38
rs1056709588 Chr.2: 181558568-181558568 on Build GRCh38
rs140898616 Chr.2: 181558651-181558651 on Build GRCh38
rs755482464 Chr.2: 181558769-181558769 on Build GRCh38
rs769531618 Chr.2: 181558550-181558550 on Build GRCh38
rs114440118 Chr.2: 181558818-181558818 on Build GRCh38
rs189638090 Chr.2: 181558557-181558557 on Build GRCh38
rs748685359 Chr.2: 181558738-181558739 on Build GRCh38
rs182279813 Chr.2: 181558794-181558794 on Build GRCh38
rs770502010 Chr.2: 181558665-181558665 on Build GRCh38
rs1023142358 Chr.2: 181558848-181558848 on Build GRCh38
rs561718044 Chr.2: 181558592-181558592 on Build GRCh38
rs773354519 Chr.2: 181558743-181558743 on Build GRCh38
rs939349382 Chr.2: 181558562-181558562 on Build GRCh38
rs776832766 Chr.2: 181558729-181558729 on Build GRCh38
rs78484040 Chr.2: 181558692-181558692 on Build GRCh38
rs968149114 Chr.2: 181558852-181558852 on Build GRCh38
rs756538383 Chr.2: 181558647-181558647 on Build GRCh38
rs992634255 Chr.2: 181558847-181558847 on Build GRCh38
rs1031534164 Chr.2: 181558799-181558799 on Build GRCh38
rs762307171 Chr.2: 181558731-181558731 on Build GRCh38
rs368210950 Chr.2: 181558543-181558543 on Build GRCh38
rs780632196 Chr.2: 181558721-181558721 on Build GRCh38
rs541121385 Chr.2: 181558623-181558623 on Build GRCh38
rs752573519 Chr.2: 181558648-181558648 on Build GRCh38
rs914415083 Chr.2: 181558821-181558821 on Build GRCh38
rs759433647 Chr.2: 181558758-181558758 on Build GRCh38
rs780823438 Chr.2: 181558627-181558627 on Build GRCh38
rs777113677 Chr.2: 181558556-181558556 on Build GRCh38
rs375777581 Chr.2: 181558571-181558571 on Build GRCh38
rs915322469 Chr.2: 181558474-181558474 on Build GRCh38
rs751684520 Chr.2: 181558754-181558754 on Build GRCh38
rs769255785 Chr.2: 181558687-181558687 on Build GRCh38
rs77741297 Chr.2: 181558603-181558603 on Build GRCh38
rs977239895 Chr.2: 181558536-181558536 on Build GRCh38
rs761272654 Chr.2: 181558583-181558583 on Build GRCh38
rs549548530 Chr.2: 181558744-181558744 on Build GRCh38
rs965655071 Chr.2: 181558679-181558679 on Build GRCh38
rs778601169 Chr.2: 181558670-181558670 on Build GRCh38
rs769105750 Chr.2: 181558588-181558588 on Build GRCh38
rs772696039 Chr.2: 181558565-181558565 on Build GRCh38
rs121909398 Chr.2: 181558617-181558617 on Build GRCh38
rs765537516 Chr.2: 181558604-181558604 on Build GRCh38
rs367791203 Chr.2: 181558737-181558737 on Build GRCh38
rs754490297 Chr.2: 181558699-181558702 on Build GRCh38
rs1021500276 Chr.2: 181558475-181558475 on Build GRCh38

View Full Product Details