Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 121916139-121916731 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSMD9
Entrez Gene ID
5715
Gene Name
proteasome 26S subunit, non-ATPase 9
Gene Aliases
Rpn4, p27
UniGene
Hs.131151
Gene Chromosome Location
Chr.12: 121888731-121917865 on Build GRCh38
Transcripts
NM_001261400.2 NM_002813.6 NR_048555.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4718503 c.668G>T p.R223I Chr.12: 121916307-121916307 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs777359445 Chr.12: 121916312-121916312 on Build GRCh38
rs762108833 Chr.12: 121916280-121916280 on Build GRCh38
rs746700358 Chr.12: 121916241-121916241 on Build GRCh38
rs201745629 Chr.12: 121916325-121916325 on Build GRCh38
rs555608220 Chr.12: 121916268-121916268 on Build GRCh38
rs551601737 Chr.12: 121916445-121916445 on Build GRCh38
rs553228163 Chr.12: 121916322-121916322 on Build GRCh38
rs1040054788 Chr.12: 121916564-121916564 on Build GRCh38
rs1004341337 Chr.12: 121916599-121916601 on Build GRCh38
rs117351080 Chr.12: 121916571-121916571 on Build GRCh38
rs370191768 Chr.12: 121916347-121916347 on Build GRCh38
rs987536383 Chr.12: 121916705-121916705 on Build GRCh38
rs527756062 Chr.12: 121916289-121916289 on Build GRCh38
rs951218218 Chr.12: 121916538-121916538 on Build GRCh38
rs922006315 Chr.12: 121916351-121916351 on Build GRCh38
rs780762107 Chr.12: 121916334-121916334 on Build GRCh38
rs971686639 Chr.12: 121916338-121916338 on Build GRCh38
rs542956253 Chr.12: 121916482-121916482 on Build GRCh38
rs754376167 Chr.12: 121916297-121916297 on Build GRCh38
rs973367549 Chr.12: 121916594-121916594 on Build GRCh38
rs373402172 Chr.12: 121916353-121916353 on Build GRCh38
rs562129475 Chr.12: 121916253-121916253 on Build GRCh38
rs999967060 Chr.12: 121916196-121916196 on Build GRCh38
rs3182919 Chr.12: 121916529-121916529 on Build GRCh38
rs569987275 Chr.12: 121916242-121916242 on Build GRCh38
rs1041941560 Chr.12: 121916450-121916450 on Build GRCh38
rs993063598 Chr.12: 121916221-121916221 on Build GRCh38
rs200207711 Chr.12: 121916317-121916317 on Build GRCh38
rs771544882 Chr.12: 121916530-121916531 on Build GRCh38
rs567386937 Chr.12: 121916277-121916277 on Build GRCh38
rs780566522 Chr.12: 121916570-121916570 on Build GRCh38
rs890587777 Chr.12: 121916417-121916417 on Build GRCh38
rs138686848 Chr.12: 121916530-121916530 on Build GRCh38
rs958946993 Chr.12: 121916669-121916669 on Build GRCh38
rs980451747 Chr.12: 121916626-121916626 on Build GRCh38
rs559198088 Chr.12: 121916565-121916565 on Build GRCh38
rs764264680 Chr.12: 121916271-121916271 on Build GRCh38
rs776485580 Chr.12: 121916244-121916244 on Build GRCh38
rs769838709 Chr.12: 121916257-121916257 on Build GRCh38
rs970825855 Chr.12: 121916601-121916601 on Build GRCh38
rs1020370904 Chr.12: 121916166-121916166 on Build GRCh38
rs775526435 Chr.12: 121916355-121916355 on Build GRCh38
rs375242237 Chr.12: 121916219-121916219 on Build GRCh38
rs904214992 Chr.12: 121916476-121916476 on Build GRCh38
rs142086448 Chr.12: 121916354-121916354 on Build GRCh38
rs755895796 Chr.12: 121916364-121916364 on Build GRCh38
rs990271696 Chr.12: 121916457-121916457 on Build GRCh38
rs149241828 Chr.12: 121916663-121916663 on Build GRCh38
rs766842397 Chr.12: 121916291-121916291 on Build GRCh38
rs188620272 Chr.12: 121916170-121916170 on Build GRCh38
rs551398110 Chr.12: 121916652-121916652 on Build GRCh38
rs537195659 Chr.12: 121916258-121916258 on Build GRCh38
rs1024661358 Chr.12: 121916599-121916604 on Build GRCh38
rs373134121 Chr.12: 121916299-121916299 on Build GRCh38
rs767698577 Chr.12: 121916288-121916288 on Build GRCh38
rs557187231 Chr.12: 121916376-121916376 on Build GRCh38
rs372200500 Chr.12: 121916211-121916211 on Build GRCh38
rs768948872 Chr.12: 121916694-121916694 on Build GRCh38
rs575543796 Chr.12: 121916406-121916406 on Build GRCh38
rs891094952 Chr.12: 121916497-121916497 on Build GRCh38
rs1041731085 Chr.12: 121916493-121916493 on Build GRCh38
rs932184734 Chr.12: 121916400-121916400 on Build GRCh38
rs932060149 Chr.12: 121916679-121916679 on Build GRCh38
rs1013967033 Chr.12: 121916509-121916509 on Build GRCh38
rs561143471 Chr.12: 121916484-121916484 on Build GRCh38
rs2272134 Chr.12: 121916498-121916498 on Build GRCh38
rs530814813 Chr.12: 121916696-121916696 on Build GRCh38

View Full Product Details