Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76735262-76735790 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SRSF2
Entrez Gene ID
6427
Gene Name
serine and arginine rich splicing factor 2
Gene Aliases
PR264, SC-35, SC35, SFRS2, SFRS2A, SRp30b
UniGene
Hs.584801
Gene Chromosome Location
Chr.17: 76734115-76737431 on Build GRCh38
Transcripts
NM_001195427.1 NM_003016.4 NR_036608.1 XM_017024942.1

SNP Detail

SNP ID Chromosome Location
rs968206787 Chr.17: 76735354-76735355 on Build GRCh38
rs1022516938 Chr.17: 76735461-76735461 on Build GRCh38
rs540418831 Chr.17: 76735405-76735406 on Build GRCh38
rs779357693 Chr.17: 76735561-76735561 on Build GRCh38
rs960185396 Chr.17: 76735304-76735304 on Build GRCh38
rs944798038 Chr.17: 76735436-76735436 on Build GRCh38
rs1018913779 Chr.17: 76735424-76735424 on Build GRCh38
rs948623170 Chr.17: 76735396-76735396 on Build GRCh38
rs886281458 Chr.17: 76735732-76735732 on Build GRCh38
rs545981405 Chr.17: 76735460-76735460 on Build GRCh38
rs967999665 Chr.17: 76735333-76735333 on Build GRCh38
rs369068156 Chr.17: 76735459-76735459 on Build GRCh38
rs948402642 Chr.17: 76735594-76735594 on Build GRCh38
rs937023047 Chr.17: 76735484-76735484 on Build GRCh38
rs1044350428 Chr.17: 76735402-76735405 on Build GRCh38
rs895527857 Chr.17: 76735526-76735526 on Build GRCh38
rs912016970 Chr.17: 76735418-76735418 on Build GRCh38
rs924387027 Chr.17: 76735374-76735374 on Build GRCh38
rs898968506 Chr.17: 76735404-76735405 on Build GRCh38
rs554649789 Chr.17: 76735329-76735329 on Build GRCh38
rs978454234 Chr.17: 76735359-76735359 on Build GRCh38
rs534983708 Chr.17: 76735695-76735695 on Build GRCh38
rs953408224 Chr.17: 76735384-76735384 on Build GRCh38
rs979050549 Chr.17: 76735352-76735352 on Build GRCh38
rs372039497 Chr.17: 76735709-76735709 on Build GRCh38
rs986056946 Chr.17: 76735686-76735686 on Build GRCh38
rs916895980 Chr.17: 76735586-76735587 on Build GRCh38
rs1031829895 Chr.17: 76735515-76735515 on Build GRCh38
rs918442289 Chr.17: 76735456-76735456 on Build GRCh38
rs1018911309 Chr.17: 76735706-76735706 on Build GRCh38
rs900504123 Chr.17: 76735449-76735449 on Build GRCh38
rs375839667 Chr.17: 76735371-76735371 on Build GRCh38
rs747729103 Chr.17: 76735562-76735562 on Build GRCh38
rs980000174 Chr.17: 76735653-76735653 on Build GRCh38
rs771793423 Chr.17: 76735564-76735564 on Build GRCh38
rs750543548 Chr.17: 76735298-76735298 on Build GRCh38
rs570513126 Chr.17: 76735455-76735455 on Build GRCh38
rs185312656 Chr.17: 76735660-76735660 on Build GRCh38
rs755257044 Chr.17: 76735467-76735467 on Build GRCh38
rs1055549685 Chr.17: 76735494-76735494 on Build GRCh38
rs1000842652 Chr.17: 76735588-76735588 on Build GRCh38
rs986172903 Chr.17: 76735406-76735406 on Build GRCh38
rs1009307719 Chr.17: 76735420-76735420 on Build GRCh38
rs757522285 Chr.17: 76735295-76735295 on Build GRCh38
rs543184419 Chr.17: 76735400-76735400 on Build GRCh38
rs368597985 Chr.17: 76735415-76735416 on Build GRCh38
rs772841082 Chr.17: 76735622-76735622 on Build GRCh38
rs955781494 Chr.17: 76735539-76735539 on Build GRCh38
rs1014020386 Chr.17: 76735556-76735556 on Build GRCh38
rs188462893 Chr.17: 76735480-76735480 on Build GRCh38
rs1047826813 Chr.17: 76735406-76735409 on Build GRCh38
rs1056833520 Chr.17: 76735380-76735380 on Build GRCh38
rs756611477 Chr.17: 76735313-76735313 on Build GRCh38
rs781091708 Chr.17: 76735332-76735332 on Build GRCh38
rs192545116 Chr.17: 76735655-76735655 on Build GRCh38
rs966073912 Chr.17: 76735714-76735714 on Build GRCh38
rs541020530 Chr.17: 76735301-76735301 on Build GRCh38
rs574560751 Chr.17: 76735375-76735375 on Build GRCh38
rs547366739 Chr.17: 76735680-76735680 on Build GRCh38
rs930461286 Chr.17: 76735662-76735662 on Build GRCh38
rs953377199 Chr.17: 76735664-76735664 on Build GRCh38

View Full Product Details