Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40440752-40441345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERTAD3
Entrez Gene ID
29946
Gene Name
SERTA domain containing 3
Gene Aliases
RBT1
UniGene
Hs.515412
Gene Chromosome Location
Chr.19: 40440841-40444375 on Build GRCh38
Transcripts
NM_013368.3 NM_203344.2 XM_005258832.3 XM_006723179.3

SNP Detail

SNP ID Chromosome Location
rs896015704 Chr.19: 40440817-40440817 on Build GRCh38
rs34185184 Chr.19: 40440886-40440887 on Build GRCh38
rs957758273 Chr.19: 40440908-40440908 on Build GRCh38
rs1007709751 Chr.19: 40441137-40441137 on Build GRCh38
rs1031765947 Chr.19: 40441159-40441159 on Build GRCh38
rs963250205 Chr.19: 40441273-40441276 on Build GRCh38
rs182375818 Chr.19: 40440942-40440942 on Build GRCh38
rs1037320488 Chr.19: 40441047-40441047 on Build GRCh38
rs756861074 Chr.19: 40441063-40441063 on Build GRCh38
rs75951686 Chr.19: 40441029-40441029 on Build GRCh38
rs969232050 Chr.19: 40440876-40440879 on Build GRCh38
rs2368579 Chr.19: 40441054-40441054 on Build GRCh38
rs950229463 Chr.19: 40441057-40441057 on Build GRCh38
rs7259593 Chr.19: 40441169-40441169 on Build GRCh38
rs971074162 Chr.19: 40441226-40441226 on Build GRCh38
rs10419182 Chr.19: 40440775-40440775 on Build GRCh38
rs983426071 Chr.19: 40441287-40441287 on Build GRCh38
rs1017475292 Chr.19: 40441191-40441191 on Build GRCh38
rs773723615 Chr.19: 40441205-40441205 on Build GRCh38
rs1058041 Chr.19: 40440938-40440938 on Build GRCh38
rs755652132 Chr.19: 40441127-40441127 on Build GRCh38
rs149562030 Chr.19: 40440856-40440856 on Build GRCh38
rs1032049988 Chr.19: 40440907-40440907 on Build GRCh38
rs1033963933 Chr.19: 40441245-40441246 on Build GRCh38
rs10660252 Chr.19: 40440902-40440903 on Build GRCh38
rs781395976 Chr.19: 40440992-40440992 on Build GRCh38
rs143239771 Chr.19: 40440915-40440915 on Build GRCh38
rs954492211 Chr.19: 40441165-40441165 on Build GRCh38
rs1014436980 Chr.19: 40440825-40440825 on Build GRCh38
rs1002269808 Chr.19: 40440859-40440859 on Build GRCh38
rs542682405 Chr.19: 40440887-40440887 on Build GRCh38
rs543007336 Chr.19: 40441264-40441264 on Build GRCh38
rs1045004558 Chr.19: 40440826-40440826 on Build GRCh38
rs1031712137 Chr.19: 40440918-40440918 on Build GRCh38
rs538540951 Chr.19: 40441026-40441026 on Build GRCh38
rs1020731876 Chr.19: 40440937-40440937 on Build GRCh38
rs1017676358 Chr.19: 40440873-40440874 on Build GRCh38
rs565094894 Chr.19: 40440972-40440972 on Build GRCh38
rs376289897 Chr.19: 40441024-40441024 on Build GRCh38
rs910790067 Chr.19: 40440881-40440881 on Build GRCh38
rs370974128 Chr.19: 40441151-40441151 on Build GRCh38
rs993744033 Chr.19: 40440780-40440780 on Build GRCh38
rs192624391 Chr.19: 40441138-40441138 on Build GRCh38
rs547262323 Chr.19: 40440902-40440902 on Build GRCh38
rs866970531 Chr.19: 40441049-40441049 on Build GRCh38
rs749460123 Chr.19: 40441188-40441188 on Build GRCh38
rs184499405 Chr.19: 40441315-40441315 on Build GRCh38
rs970555768 Chr.19: 40440970-40440970 on Build GRCh38
rs771785806 Chr.19: 40441289-40441289 on Build GRCh38
rs111689476 Chr.19: 40441079-40441079 on Build GRCh38
rs1009255993 Chr.19: 40441139-40441139 on Build GRCh38
rs1000655884 Chr.19: 40441150-40441153 on Build GRCh38
rs1008154151 Chr.19: 40440869-40440869 on Build GRCh38
rs372697105 Chr.19: 40441037-40441037 on Build GRCh38
rs747762827 Chr.19: 40441270-40441270 on Build GRCh38
rs1053438705 Chr.19: 40441134-40441134 on Build GRCh38
rs920618344 Chr.19: 40440985-40440985 on Build GRCh38
rs572125420 Chr.19: 40441090-40441090 on Build GRCh38
rs910424775 Chr.19: 40440961-40440961 on Build GRCh38
rs750958747 Chr.19: 40441085-40441085 on Build GRCh38
rs973373904 Chr.19: 40440924-40440924 on Build GRCh38
rs554049973 Chr.19: 40441078-40441078 on Build GRCh38
rs575681541 Chr.19: 40441235-40441235 on Build GRCh38
rs746171340 Chr.19: 40441042-40441042 on Build GRCh38
rs112924689 Chr.19: 40440898-40440902 on Build GRCh38
rs768387108 Chr.19: 40441256-40441256 on Build GRCh38
rs950201876 Chr.19: 40441317-40441317 on Build GRCh38
rs1046909890 Chr.19: 40441055-40441055 on Build GRCh38
rs1047866706 Chr.19: 40440799-40440799 on Build GRCh38
rs377391999 Chr.19: 40441286-40441286 on Build GRCh38
rs148262714 Chr.19: 40440979-40440979 on Build GRCh38
rs377221600 Chr.19: 40441169-40441169 on Build GRCh38
rs986346880 Chr.19: 40440877-40440877 on Build GRCh38
rs7259255 Chr.19: 40441073-40441073 on Build GRCh38
rs917856535 Chr.19: 40440968-40440974 on Build GRCh38
rs963643778 Chr.19: 40440884-40440884 on Build GRCh38
rs1020674873 Chr.19: 40441164-40441164 on Build GRCh38
rs973086895 Chr.19: 40441278-40441278 on Build GRCh38
rs920524486 Chr.19: 40441280-40441280 on Build GRCh38

View Full Product Details