Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 91907840-91908370 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPC5
Entrez Gene ID
2262
Gene Name
glypican 5
Gene Aliases
-
UniGene
Hs.655675
Gene Chromosome Location
Chr.13: 91398619-92867237 on Build GRCh38
Transcripts
NM_004466.5 XM_011521054.2 XM_017020435.1 XM_017020437.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5882448 c.1281T>G p.S427R Chr.13: 91907937-91907937 on Build GRCh38
4048755 c.1393G>C p.V465L Chr.13: 91908049-91908049 on Build GRCh38
4048754 c.1368T>A p.N456K Chr.13: 91908024-91908024 on Build GRCh38
3980083 c.1341A>T p.K447N Chr.13: 91907997-91907997 on Build GRCh38
4048753 c.1317C>A p.A439A Chr.13: 91907973-91907973 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879793779 Chr.13: 91907866-91907866 on Build GRCh38
rs746069979 Chr.13: 91907902-91907902 on Build GRCh38
rs768674288 Chr.13: 91907942-91907942 on Build GRCh38
rs143619996 Chr.13: 91908306-91908306 on Build GRCh38
rs766072631 Chr.13: 91907955-91907955 on Build GRCh38
rs774179295 Chr.13: 91907945-91907945 on Build GRCh38
rs749419445 Chr.13: 91907914-91907914 on Build GRCh38
rs547561820 Chr.13: 91908303-91908303 on Build GRCh38
rs746591444 Chr.13: 91907990-91907995 on Build GRCh38
rs781394550 Chr.13: 91907995-91907995 on Build GRCh38
rs151065141 Chr.13: 91908070-91908070 on Build GRCh38
rs548686390 Chr.13: 91908215-91908215 on Build GRCh38
rs747363004 Chr.13: 91907894-91907894 on Build GRCh38
rs746932059 Chr.13: 91908284-91908284 on Build GRCh38
rs752283402 Chr.13: 91907991-91907991 on Build GRCh38
rs758975757 Chr.13: 91908106-91908106 on Build GRCh38
rs753318955 Chr.13: 91907971-91907971 on Build GRCh38
rs772106337 Chr.13: 91908045-91908045 on Build GRCh38
rs536610153 Chr.13: 91908031-91908031 on Build GRCh38
rs187710920 Chr.13: 91908089-91908089 on Build GRCh38
rs141144398 Chr.13: 91907981-91907981 on Build GRCh38
rs760506328 Chr.13: 91908073-91908073 on Build GRCh38
rs545921186 Chr.13: 91908056-91908056 on Build GRCh38
rs192761298 Chr.13: 91908225-91908225 on Build GRCh38
rs753406788 Chr.13: 91907892-91907892 on Build GRCh38
rs1025172375 Chr.13: 91908218-91908218 on Build GRCh38
rs201435726 Chr.13: 91908096-91908096 on Build GRCh38
rs770197708 Chr.13: 91907909-91907909 on Build GRCh38
rs372599859 Chr.13: 91907899-91907899 on Build GRCh38
rs752010478 Chr.13: 91907985-91907985 on Build GRCh38
rs754517667 Chr.13: 91907973-91907973 on Build GRCh38
rs1033977979 Chr.13: 91908264-91908264 on Build GRCh38
rs764645419 Chr.13: 91908108-91908108 on Build GRCh38
rs774278491 Chr.13: 91908016-91908016 on Build GRCh38
rs749331025 Chr.13: 91908004-91908004 on Build GRCh38
rs574701648 Chr.13: 91907893-91907893 on Build GRCh38
rs34433071 Chr.13: 91908012-91908013 on Build GRCh38
rs150087896 Chr.13: 91907871-91907871 on Build GRCh38
rs769173341 Chr.13: 91907989-91907990 on Build GRCh38
rs1002646874 Chr.13: 91908247-91908247 on Build GRCh38
rs780981711 Chr.13: 91908316-91908316 on Build GRCh38
rs958369319 Chr.13: 91908267-91908267 on Build GRCh38
rs1854735 Chr.13: 91908171-91908171 on Build GRCh38
rs767223885 Chr.13: 91907948-91907948 on Build GRCh38
rs961980177 Chr.13: 91908319-91908319 on Build GRCh38
rs775687782 Chr.13: 91907913-91907913 on Build GRCh38
rs113389599 Chr.13: 91908098-91908098 on Build GRCh38
rs184567739 Chr.13: 91908289-91908289 on Build GRCh38
rs771696078 Chr.13: 91907911-91907912 on Build GRCh38
rs567674940 Chr.13: 91907999-91907999 on Build GRCh38
rs201904220 Chr.13: 91908010-91908010 on Build GRCh38
rs778379420 Chr.13: 91907920-91907920 on Build GRCh38
rs76285944 Chr.13: 91907947-91907947 on Build GRCh38
rs936891806 Chr.13: 91908090-91908090 on Build GRCh38
rs779865516 Chr.13: 91907923-91907924 on Build GRCh38
rs532444694 Chr.13: 91907954-91907954 on Build GRCh38
rs764600232 Chr.13: 91907974-91907974 on Build GRCh38
rs958581390 Chr.13: 91907874-91907874 on Build GRCh38
rs372608677 Chr.13: 91908156-91908156 on Build GRCh38
rs188301257 Chr.13: 91908345-91908345 on Build GRCh38
rs779535856 Chr.13: 91907890-91907890 on Build GRCh38
rs540414519 Chr.13: 91907901-91907901 on Build GRCh38
rs760310392 Chr.13: 91907950-91907950 on Build GRCh38
rs780454774 Chr.13: 91908001-91908001 on Build GRCh38
rs1010144427 Chr.13: 91908192-91908192 on Build GRCh38

View Full Product Details