Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 73997229-73997828 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXCL5
Entrez Gene ID
6374
Gene Name
C-X-C motif chemokine ligand 5
Gene Aliases
ENA-78, SCYB5
UniGene
Hs.89714
Gene Chromosome Location
Chr.4: 73995642-73998729 on Build GRCh38
Transcripts
NM_002994.4

SNP Detail

SNP ID Chromosome Location
rs751235100 Chr.4: 73997645-73997645 on Build GRCh38
rs978786473 Chr.4: 73997313-73997313 on Build GRCh38
rs754835574 Chr.4: 73997674-73997674 on Build GRCh38
rs575741163 Chr.4: 73997314-73997314 on Build GRCh38
rs577886302 Chr.4: 73997458-73997458 on Build GRCh38
rs200889054 Chr.4: 73997440-73997440 on Build GRCh38
rs201773715 Chr.4: 73997420-73997420 on Build GRCh38
rs369451005 Chr.4: 73997383-73997383 on Build GRCh38
rs770868935 Chr.4: 73997687-73997687 on Build GRCh38
rs200795293 Chr.4: 73997384-73997384 on Build GRCh38
rs1048160202 Chr.4: 73997463-73997463 on Build GRCh38
rs183686562 Chr.4: 73997378-73997378 on Build GRCh38
rs564533575 Chr.4: 73997485-73997485 on Build GRCh38
rs201088551 Chr.4: 73997488-73997488 on Build GRCh38
rs573856448 Chr.4: 73997392-73997392 on Build GRCh38
rs199801246 Chr.4: 73997373-73997373 on Build GRCh38
rs775872229 Chr.4: 73997634-73997634 on Build GRCh38
rs925144623 Chr.4: 73997329-73997329 on Build GRCh38
rs949299656 Chr.4: 73997408-73997408 on Build GRCh38
rs1017514608 Chr.4: 73997503-73997503 on Build GRCh38
rs201478344 Chr.4: 73997639-73997639 on Build GRCh38
rs780931521 Chr.4: 73997590-73997590 on Build GRCh38
rs199791837 Chr.4: 73997355-73997355 on Build GRCh38
rs202077549 Chr.4: 73997459-73997459 on Build GRCh38
rs200074987 Chr.4: 73997473-73997473 on Build GRCh38
rs930948306 Chr.4: 73997802-73997802 on Build GRCh38
rs969226794 Chr.4: 73997283-73997283 on Build GRCh38
rs2437283 Chr.4: 73997268-73997268 on Build GRCh38
rs201194735 Chr.4: 73997528-73997528 on Build GRCh38
rs1045977188 Chr.4: 73997745-73997746 on Build GRCh38
rs368843287 Chr.4: 73997671-73997671 on Build GRCh38
rs1028284586 Chr.4: 73997276-73997276 on Build GRCh38
rs370353558 Chr.4: 73997605-73997605 on Build GRCh38
rs373774200 Chr.4: 73997607-73997607 on Build GRCh38
rs903201068 Chr.4: 73997416-73997416 on Build GRCh38
rs200359847 Chr.4: 73997632-73997632 on Build GRCh38
rs980687266 Chr.4: 73997740-73997740 on Build GRCh38
rs112228157 Chr.4: 73997368-73997368 on Build GRCh38
rs566035158 Chr.4: 73997753-73997753 on Build GRCh38
rs760110682 Chr.4: 73997622-73997622 on Build GRCh38
rs542618725 Chr.4: 73997713-73997713 on Build GRCh38
rs199840924 Chr.4: 73997683-73997683 on Build GRCh38
rs926313004 Chr.4: 73997786-73997786 on Build GRCh38
rs569345229 Chr.4: 73997433-73997433 on Build GRCh38
rs112950358 Chr.4: 73997427-73997427 on Build GRCh38
rs545229027 Chr.4: 73997476-73997476 on Build GRCh38
rs975998986 Chr.4: 73997525-73997528 on Build GRCh38
rs199779476 Chr.4: 73997644-73997644 on Build GRCh38
rs757203077 Chr.4: 73997588-73997588 on Build GRCh38
rs201865226 Chr.4: 73997676-73997676 on Build GRCh38
rs201132592 Chr.4: 73997353-73997353 on Build GRCh38
rs761157781 Chr.4: 73997646-73997646 on Build GRCh38
rs62313969 Chr.4: 73997344-73997344 on Build GRCh38
rs200225019 Chr.4: 73997511-73997511 on Build GRCh38
rs112731620 Chr.4: 73997337-73997337 on Build GRCh38
rs750273985 Chr.4: 73997656-73997656 on Build GRCh38
rs756121915 Chr.4: 73997659-73997659 on Build GRCh38
rs965990474 Chr.4: 73997515-73997515 on Build GRCh38
rs376987649 Chr.4: 73997702-73997702 on Build GRCh38
rs777989099 Chr.4: 73997612-73997612 on Build GRCh38
rs1007833872 Chr.4: 73997464-73997464 on Build GRCh38
rs531752920 Chr.4: 73997579-73997579 on Build GRCh38
rs200225480 Chr.4: 73997571-73997571 on Build GRCh38
rs975812698 Chr.4: 73997270-73997270 on Build GRCh38
rs910021342 Chr.4: 73997285-73997285 on Build GRCh38
rs113256650 Chr.4: 73997779-73997779 on Build GRCh38
rs887297047 Chr.4: 73997466-73997466 on Build GRCh38
rs1025950739 Chr.4: 73997560-73997560 on Build GRCh38
rs894746750 Chr.4: 73997382-73997382 on Build GRCh38
rs199513293 Chr.4: 73997650-73997650 on Build GRCh38
rs755821323 Chr.4: 73997593-73997593 on Build GRCh38
rs201837496 Chr.4: 73997544-73997544 on Build GRCh38
rs200403272 Chr.4: 73997606-73997606 on Build GRCh38
rs879134758 Chr.4: 73997660-73997660 on Build GRCh38
rs201061472 Chr.4: 73997619-73997619 on Build GRCh38
rs200500877 Chr.4: 73997286-73997286 on Build GRCh38
rs200905597 Chr.4: 73997362-73997362 on Build GRCh38
rs748353488 Chr.4: 73997594-73997594 on Build GRCh38
rs751136073 Chr.4: 73997587-73997587 on Build GRCh38
rs971072363 Chr.4: 73997733-73997733 on Build GRCh38
rs200650135 Chr.4: 73997657-73997657 on Build GRCh38
rs746329455 Chr.4: 73997707-73997707 on Build GRCh38
rs201722538 Chr.4: 73997667-73997667 on Build GRCh38
rs557053868 Chr.4: 73997450-73997450 on Build GRCh38
rs202236190 Chr.4: 73997502-73997502 on Build GRCh38
rs74505573 Chr.4: 73997726-73997726 on Build GRCh38
rs202044784 Chr.4: 73997372-73997372 on Build GRCh38
rs199752112 Chr.4: 73997669-73997669 on Build GRCh38
rs34648742 Chr.4: 73997609-73997610 on Build GRCh38
rs781381640 Chr.4: 73997705-73997705 on Build GRCh38
rs199526761 Chr.4: 73997620-73997620 on Build GRCh38
rs200824893 Chr.4: 73997673-73997673 on Build GRCh38

View Full Product Details