Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 89926256-89926751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BACH2
Entrez Gene ID
60468
Gene Name
BTB domain and CNC homolog 2
Gene Aliases
BTBD25
UniGene
Hs.269764
Gene Chromosome Location
Chr.6: 89926528-90296912 on Build GRCh38
Transcripts
NM_001170794.1 NM_021813.3 XM_005248758.4 XM_005248759.4 XM_017011156.1 XM_017011157.1 XM_017011158.1 XM_017011159.1 XM_017011160.1 XM_017011161.1 XM_017011162.1 XM_017011163.1 XM_017011164.1 XM_017011166.1

SNP Detail

SNP ID Chromosome Location
rs929884477 Chr.6: 89926542-89926542 on Build GRCh38
rs73492610 Chr.6: 89926316-89926316 on Build GRCh38
rs535014348 Chr.6: 89926625-89926625 on Build GRCh38
rs753858780 Chr.6: 89926718-89926721 on Build GRCh38
rs1057486191 Chr.6: 89926707-89926707 on Build GRCh38
rs145472587 Chr.6: 89926691-89926692 on Build GRCh38
rs950431428 Chr.6: 89926460-89926460 on Build GRCh38
rs796505454 Chr.6: 89926420-89926420 on Build GRCh38
rs183477491 Chr.6: 89926322-89926322 on Build GRCh38
rs758959292 Chr.6: 89926304-89926304 on Build GRCh38
rs114115002 Chr.6: 89926405-89926405 on Build GRCh38
rs147897846 Chr.6: 89926298-89926298 on Build GRCh38
rs552992903 Chr.6: 89926673-89926673 on Build GRCh38
rs945348041 Chr.6: 89926601-89926601 on Build GRCh38
rs753177297 Chr.6: 89926297-89926297 on Build GRCh38
rs983467886 Chr.6: 89926546-89926546 on Build GRCh38
rs186996113 Chr.6: 89926472-89926472 on Build GRCh38
rs915201560 Chr.6: 89926598-89926598 on Build GRCh38
rs557070710 Chr.6: 89926562-89926562 on Build GRCh38
rs200683305 Chr.6: 89926698-89926698 on Build GRCh38
rs1020374156 Chr.6: 89926359-89926359 on Build GRCh38
rs978508455 Chr.6: 89926389-89926389 on Build GRCh38
rs1027693778 Chr.6: 89926426-89926426 on Build GRCh38
rs911627320 Chr.6: 89926479-89926479 on Build GRCh38
rs959882695 Chr.6: 89926412-89926412 on Build GRCh38
rs1042436732 Chr.6: 89926613-89926613 on Build GRCh38
rs937466413 Chr.6: 89926675-89926675 on Build GRCh38
rs1018043390 Chr.6: 89926398-89926398 on Build GRCh38
rs548335681 Chr.6: 89926326-89926326 on Build GRCh38
rs1003664743 Chr.6: 89926638-89926638 on Build GRCh38
rs757978334 Chr.6: 89926381-89926381 on Build GRCh38
rs573813761 Chr.6: 89926510-89926510 on Build GRCh38
rs1055264608 Chr.6: 89926649-89926649 on Build GRCh38
rs965789176 Chr.6: 89926482-89926482 on Build GRCh38
rs906613288 Chr.6: 89926635-89926635 on Build GRCh38
rs552623865 Chr.6: 89926515-89926515 on Build GRCh38
rs1009871535 Chr.6: 89926334-89926334 on Build GRCh38
rs942080923 Chr.6: 89926616-89926616 on Build GRCh38
rs921819975 Chr.6: 89926543-89926543 on Build GRCh38
rs752864564 Chr.6: 89926523-89926524 on Build GRCh38
rs976567474 Chr.6: 89926494-89926494 on Build GRCh38
rs968488660 Chr.6: 89926373-89926373 on Build GRCh38
rs1003695787 Chr.6: 89926365-89926365 on Build GRCh38

View Full Product Details