Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 138237308-138237710 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC8
Entrez Gene ID
25852
Gene Name
armadillo repeat containing 8
Gene Aliases
GID5, HSPC056, S863-2, VID28
UniGene
Hs.744868
Gene Chromosome Location
Chr.3: 138187248-138298389 on Build GRCh38
Transcripts
NM_001267041.2 NM_001267042.2 NM_001282342.1 NM_014154.4 NM_015396.5 NM_213654.3 XM_006713564.3 XM_006713565.2 XM_006713566.3 XM_006713567.3 XM_011512632.1 XM_011512633.1 XM_017006095.1 XM_017006096.1 XM_017006097.1 XM_017006098.1 XM_017006099.1 XM_017006100.1 XM_017006101.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6604993 c.731T>C p.L244S Chr.3: 138237527-138237527 on Build GRCh38
6040012 c.636G>A p.L212L Chr.3: 138237377-138237377 on Build GRCh38
3123859 c.627G>A p.S209S Chr.3: 138237368-138237368 on Build GRCh38
7181452 c.623T>C p.V208A Chr.3: 138237364-138237364 on Build GRCh38
7181453 c.623T>C p.V208A Chr.3: 138237364-138237364 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768754177 Chr.3: 138237459-138237461 on Build GRCh38
rs769477742 Chr.3: 138237421-138237421 on Build GRCh38
rs568279572 Chr.3: 138237670-138237670 on Build GRCh38
rs944750282 Chr.3: 138237659-138237659 on Build GRCh38
rs201557056 Chr.3: 138237605-138237605 on Build GRCh38
rs750129504 Chr.3: 138237521-138237521 on Build GRCh38
rs764180277 Chr.3: 138237480-138237480 on Build GRCh38
rs995096481 Chr.3: 138237415-138237415 on Build GRCh38
rs756580461 Chr.3: 138237427-138237427 on Build GRCh38
rs372765310 Chr.3: 138237612-138237612 on Build GRCh38
rs747577403 Chr.3: 138237533-138237533 on Build GRCh38
rs1010889266 Chr.3: 138237677-138237677 on Build GRCh38
rs781254808 Chr.3: 138237474-138237474 on Build GRCh38
rs373520831 Chr.3: 138237363-138237363 on Build GRCh38
rs922218803 Chr.3: 138237678-138237678 on Build GRCh38
rs767209620 Chr.3: 138237474-138237474 on Build GRCh38
rs758678734 Chr.3: 138237405-138237405 on Build GRCh38
rs370567469 Chr.3: 138237492-138237492 on Build GRCh38
rs774032245 Chr.3: 138237461-138237461 on Build GRCh38
rs767315148 Chr.3: 138237511-138237511 on Build GRCh38
rs753874161 Chr.3: 138237491-138237491 on Build GRCh38
rs376816457 Chr.3: 138237401-138237401 on Build GRCh38
rs780678884 Chr.3: 138237406-138237406 on Build GRCh38
rs373138393 Chr.3: 138237411-138237411 on Build GRCh38
rs763950862 Chr.3: 138237609-138237609 on Build GRCh38
rs774117383 Chr.3: 138237588-138237588 on Build GRCh38
rs760503808 Chr.3: 138237478-138237478 on Build GRCh38
rs754108321 Chr.3: 138237356-138237356 on Build GRCh38
rs760637129 Chr.3: 138237607-138237607 on Build GRCh38
rs775533658 Chr.3: 138237477-138237477 on Build GRCh38
rs755505808 Chr.3: 138237550-138237550 on Build GRCh38
rs912018321 Chr.3: 138237616-138237616 on Build GRCh38
rs748016059 Chr.3: 138237578-138237581 on Build GRCh38
rs144945210 Chr.3: 138237443-138237443 on Build GRCh38
rs757421910 Chr.3: 138237358-138237358 on Build GRCh38
rs748752087 Chr.3: 138237581-138237581 on Build GRCh38
rs770643415 Chr.3: 138237444-138237444 on Build GRCh38
rs199891060 Chr.3: 138237599-138237599 on Build GRCh38
rs759430497 Chr.3: 138237464-138237464 on Build GRCh38
rs777429378 Chr.3: 138237579-138237579 on Build GRCh38
rs958031372 Chr.3: 138237643-138237643 on Build GRCh38
rs764124674 Chr.3: 138237339-138237339 on Build GRCh38
rs974732037 Chr.3: 138237661-138237661 on Build GRCh38
rs962334538 Chr.3: 138237540-138237540 on Build GRCh38
rs201763796 Chr.3: 138237507-138237507 on Build GRCh38
rs368286300 Chr.3: 138237425-138237425 on Build GRCh38
rs772804188 Chr.3: 138237422-138237422 on Build GRCh38
rs374327467 Chr.3: 138237506-138237506 on Build GRCh38
rs1001352819 Chr.3: 138237442-138237442 on Build GRCh38
rs375848957 Chr.3: 138237413-138237413 on Build GRCh38
rs184380560 Chr.3: 138237584-138237584 on Build GRCh38
rs769711172 Chr.3: 138237602-138237603 on Build GRCh38
rs200890393 Chr.3: 138237509-138237509 on Build GRCh38
rs771946881 Chr.3: 138237602-138237602 on Build GRCh38
rs1013413921 Chr.3: 138237429-138237429 on Build GRCh38
rs1043023506 Chr.3: 138237688-138237688 on Build GRCh38

View Full Product Details