Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 16602100-16602554 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKMY2
Entrez Gene ID
57037
Gene Name
ankyrin repeat and MYND domain containing 2
Gene Aliases
ZMYND20
UniGene
Hs.157378
Gene Chromosome Location
Chr.7: 16599776-16645817 on Build GRCh38
Transcripts
NM_020319.2 XM_011515458.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5450807 c.1141C>T p.H381Y Chr.7: 16602380-16602380 on Build GRCh38
6260242 c.1129C>A p.Q377K Chr.7: 16602392-16602392 on Build GRCh38
3394647 c.1066A>G p.I356V Chr.7: 16602455-16602455 on Build GRCh38
6600250 c.1122A>G p.E374E Chr.7: 16602399-16602399 on Build GRCh38
5757019 c.1133A>G p.E378G Chr.7: 16602388-16602388 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377500502 Chr.7: 16602521-16602521 on Build GRCh38
rs112788803 Chr.7: 16602376-16602376 on Build GRCh38
rs769928784 Chr.7: 16602499-16602499 on Build GRCh38
rs556289826 Chr.7: 16602331-16602331 on Build GRCh38
rs375531087 Chr.7: 16602240-16602240 on Build GRCh38
rs562753435 Chr.7: 16602229-16602229 on Build GRCh38
rs372115112 Chr.7: 16602332-16602332 on Build GRCh38
rs138056522 Chr.7: 16602396-16602396 on Build GRCh38
rs150352747 Chr.7: 16602394-16602394 on Build GRCh38
rs755463501 Chr.7: 16602302-16602302 on Build GRCh38
rs80143045 Chr.7: 16602144-16602144 on Build GRCh38
rs767206252 Chr.7: 16602406-16602406 on Build GRCh38
rs1056775785 Chr.7: 16602364-16602365 on Build GRCh38
rs369509832 Chr.7: 16602433-16602433 on Build GRCh38
rs373650566 Chr.7: 16602379-16602379 on Build GRCh38
rs774712088 Chr.7: 16602455-16602478 on Build GRCh38
rs949346652 Chr.7: 16602287-16602287 on Build GRCh38
rs762744024 Chr.7: 16602374-16602374 on Build GRCh38
rs1032049218 Chr.7: 16602311-16602311 on Build GRCh38
rs548779277 Chr.7: 16602300-16602300 on Build GRCh38
rs779296388 Chr.7: 16602430-16602430 on Build GRCh38
rs745422390 Chr.7: 16602337-16602337 on Build GRCh38
rs987031439 Chr.7: 16602408-16602408 on Build GRCh38
rs968042459 Chr.7: 16602138-16602138 on Build GRCh38
rs556012461 Chr.7: 16602197-16602197 on Build GRCh38
rs756827215 Chr.7: 16602405-16602405 on Build GRCh38
rs750007807 Chr.7: 16602527-16602527 on Build GRCh38
rs888057189 Chr.7: 16602184-16602184 on Build GRCh38
rs377659277 Chr.7: 16602230-16602230 on Build GRCh38
rs765431045 Chr.7: 16602373-16602373 on Build GRCh38
rs754998343 Chr.7: 16602432-16602432 on Build GRCh38
rs776948367 Chr.7: 16602487-16602487 on Build GRCh38
rs776823361 Chr.7: 16602369-16602369 on Build GRCh38
rs767899542 Chr.7: 16602524-16602524 on Build GRCh38
rs537965997 Chr.7: 16602308-16602308 on Build GRCh38
rs1047347770 Chr.7: 16602264-16602264 on Build GRCh38
rs201218392 Chr.7: 16602362-16602362 on Build GRCh38
rs768267014 Chr.7: 16602353-16602353 on Build GRCh38
rs542525602 Chr.7: 16602428-16602428 on Build GRCh38
rs144175206 Chr.7: 16602425-16602425 on Build GRCh38
rs1030417818 Chr.7: 16602511-16602511 on Build GRCh38
rs569738844 Chr.7: 16602127-16602127 on Build GRCh38
rs893549203 Chr.7: 16602318-16602318 on Build GRCh38
rs375760645 Chr.7: 16602448-16602448 on Build GRCh38
rs924278172 Chr.7: 16602323-16602323 on Build GRCh38
rs775654222 Chr.7: 16602508-16602508 on Build GRCh38
rs531852602 Chr.7: 16602140-16602140 on Build GRCh38
rs376660503 Chr.7: 16602410-16602410 on Build GRCh38
rs372868648 Chr.7: 16602363-16602363 on Build GRCh38
rs953465544 Chr.7: 16602531-16602531 on Build GRCh38
rs377221360 Chr.7: 16602370-16602370 on Build GRCh38
rs746124929 Chr.7: 16602490-16602490 on Build GRCh38
rs887004086 Chr.7: 16602265-16602265 on Build GRCh38
rs200492992 Chr.7: 16602509-16602509 on Build GRCh38
rs35548378 Chr.7: 16602345-16602345 on Build GRCh38
rs570135240 Chr.7: 16602304-16602304 on Build GRCh38
rs764120294 Chr.7: 16602385-16602387 on Build GRCh38
rs374631474 Chr.7: 16602520-16602520 on Build GRCh38
rs779916461 Chr.7: 16602344-16602344 on Build GRCh38
rs372753949 Chr.7: 16602434-16602434 on Build GRCh38
rs756021171 Chr.7: 16602338-16602338 on Build GRCh38
rs140598000 Chr.7: 16602505-16602505 on Build GRCh38
rs997551274 Chr.7: 16602476-16602476 on Build GRCh38
rs564968614 Chr.7: 16602206-16602206 on Build GRCh38
rs201167315 Chr.7: 16602533-16602533 on Build GRCh38
rs182289614 Chr.7: 16602207-16602207 on Build GRCh38
rs780677815 Chr.7: 16602166-16602166 on Build GRCh38
rs879078720 Chr.7: 16602290-16602290 on Build GRCh38
rs368736264 Chr.7: 16602350-16602350 on Build GRCh38
rs149509831 Chr.7: 16602416-16602416 on Build GRCh38
rs779539804 Chr.7: 16602429-16602429 on Build GRCh38
rs523615 Chr.7: 16602124-16602124 on Build GRCh38
rs199780371 Chr.7: 16602357-16602357 on Build GRCh38
rs10247129 Chr.7: 16602122-16602122 on Build GRCh38
rs986000601 Chr.7: 16602274-16602274 on Build GRCh38

View Full Product Details