Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 7987152-7987729 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3F
Entrez Gene ID
8665
Gene Name
eukaryotic translation initiation factor 3 subunit F
Gene Aliases
EIF3S5, eIF3-p47
UniGene
Hs.516023
Gene Chromosome Location
Chr.11: 7987320-7996171 on Build GRCh38
Transcripts
NM_003754.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4680611 c.16G>A p.V6I Chr.11: 7987368-7987368 on Build GRCh38
5791710 c.6T>A p.N2K Chr.11: 7987313-7987313 on Build GRCh38
3687678 c.161C>T p.P54L Chr.11: 7987468-7987468 on Build GRCh38
4037030 c.117C>T p.P39P Chr.11: 7987469-7987469 on Build GRCh38
5682385 c.325C>T p.L109L Chr.11: 7987632-7987632 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758048186 Chr.11: 7987402-7987402 on Build GRCh38
rs777698840 Chr.11: 7987478-7987478 on Build GRCh38
rs756899150 Chr.11: 7987365-7987365 on Build GRCh38
rs143020927 Chr.11: 7987614-7987614 on Build GRCh38
rs762690954 Chr.11: 7987618-7987618 on Build GRCh38
rs766098895 Chr.11: 7987459-7987459 on Build GRCh38
rs781140148 Chr.11: 7987594-7987594 on Build GRCh38
rs776953347 Chr.11: 7987496-7987496 on Build GRCh38
rs1028584608 Chr.11: 7987257-7987257 on Build GRCh38
rs555668027 Chr.11: 7987179-7987179 on Build GRCh38
rs749466204 Chr.11: 7987421-7987421 on Build GRCh38
rs776590320 Chr.11: 7987570-7987570 on Build GRCh38
rs758611755 Chr.11: 7987578-7987578 on Build GRCh38
rs923895867 Chr.11: 7987625-7987625 on Build GRCh38
rs754416494 Chr.11: 7987681-7987681 on Build GRCh38
rs753366278 Chr.11: 7987338-7987338 on Build GRCh38
rs746986851 Chr.11: 7987346-7987346 on Build GRCh38
rs546711035 Chr.11: 7987389-7987389 on Build GRCh38
rs995172032 Chr.11: 7987567-7987567 on Build GRCh38
rs200229110 Chr.11: 7987585-7987585 on Build GRCh38
rs770848871 Chr.11: 7987383-7987383 on Build GRCh38
rs764971812 Chr.11: 7987526-7987526 on Build GRCh38
rs534791539 Chr.11: 7987300-7987300 on Build GRCh38
rs139608264 Chr.11: 7987407-7987407 on Build GRCh38
rs760426275 Chr.11: 7987316-7987316 on Build GRCh38
rs768291543 Chr.11: 7987347-7987347 on Build GRCh38
rs868274743 Chr.11: 7987345-7987345 on Build GRCh38
rs144895933 Chr.11: 7987393-7987393 on Build GRCh38
rs116043166 Chr.11: 7987225-7987225 on Build GRCh38
rs775349466 Chr.11: 7987307-7987307 on Build GRCh38
rs778401041 Chr.11: 7987301-7987301 on Build GRCh38
rs61734576 Chr.11: 7987387-7987387 on Build GRCh38
rs767078108 Chr.11: 7987509-7987509 on Build GRCh38
rs757028511 Chr.11: 7987524-7987524 on Build GRCh38
rs763840207 Chr.11: 7987331-7987331 on Build GRCh38
rs1013010131 Chr.11: 7987525-7987525 on Build GRCh38
rs555069925 Chr.11: 7987583-7987583 on Build GRCh38
rs767622076 Chr.11: 7987652-7987652 on Build GRCh38
rs776014443 Chr.11: 7987611-7987611 on Build GRCh38
rs770082012 Chr.11: 7987498-7987498 on Build GRCh38
rs1023195454 Chr.11: 7987537-7987537 on Build GRCh38
rs747156133 Chr.11: 7987538-7987538 on Build GRCh38
rs200315934 Chr.11: 7987381-7987381 on Build GRCh38
rs1007260487 Chr.11: 7987188-7987188 on Build GRCh38
rs923605739 Chr.11: 7987657-7987657 on Build GRCh38
rs11538182 Chr.11: 7987452-7987452 on Build GRCh38
rs559602280 Chr.11: 7987233-7987233 on Build GRCh38
rs756637809 Chr.11: 7987599-7987599 on Build GRCh38
rs893597466 Chr.11: 7987215-7987215 on Build GRCh38
rs774019277 Chr.11: 7987503-7987503 on Build GRCh38
rs368298850 Chr.11: 7987394-7987394 on Build GRCh38
rs775110839 Chr.11: 7987562-7987562 on Build GRCh38
rs553070956 Chr.11: 7987252-7987252 on Build GRCh38
rs762290700 Chr.11: 7987497-7987497 on Build GRCh38
rs1017272342 Chr.11: 7987208-7987208 on Build GRCh38
rs780091744 Chr.11: 7987528-7987528 on Build GRCh38
rs773463834 Chr.11: 7987554-7987554 on Build GRCh38
rs1035815286 Chr.11: 7987372-7987372 on Build GRCh38
rs750588982 Chr.11: 7987527-7987527 on Build GRCh38
rs144028673 Chr.11: 7987181-7987181 on Build GRCh38
rs767716913 Chr.11: 7987363-7987363 on Build GRCh38
rs764166155 Chr.11: 7987518-7987518 on Build GRCh38
rs768938365 Chr.11: 7987445-7987445 on Build GRCh38
rs917726043 Chr.11: 7987430-7987431 on Build GRCh38
rs777715143 Chr.11: 7987419-7987419 on Build GRCh38
rs201599300 Chr.11: 7987340-7987340 on Build GRCh38
rs755458882 Chr.11: 7987543-7987543 on Build GRCh38
rs748993806 Chr.11: 7987281-7987281 on Build GRCh38
rs763718390 Chr.11: 7987569-7987569 on Build GRCh38
rs760844968 Chr.11: 7987553-7987554 on Build GRCh38
rs781243485 Chr.11: 7987408-7987408 on Build GRCh38
rs760309318 Chr.11: 7987565-7987565 on Build GRCh38
rs545923858 Chr.11: 7987231-7987231 on Build GRCh38
rs762496400 Chr.11: 7987457-7987457 on Build GRCh38
rs1052019716 Chr.11: 7987334-7987334 on Build GRCh38
rs751151154 Chr.11: 7987406-7987406 on Build GRCh38
rs770212034 Chr.11: 7987357-7987357 on Build GRCh38
rs745998594 Chr.11: 7987429-7987429 on Build GRCh38
rs780199714 Chr.11: 7987548-7987548 on Build GRCh38
rs151005116 Chr.11: 7987391-7987391 on Build GRCh38
rs766590381 Chr.11: 7987361-7987361 on Build GRCh38
rs771243784 Chr.11: 7987422-7987422 on Build GRCh38
rs946429530 Chr.11: 7987450-7987450 on Build GRCh38
rs767170137 Chr.11: 7987469-7987469 on Build GRCh38
rs749656019 Chr.11: 7987600-7987600 on Build GRCh38
rs755934469 Chr.11: 7987416-7987416 on Build GRCh38
rs1043738 Chr.11: 7987468-7987468 on Build GRCh38
rs201444896 Chr.11: 7987486-7987486 on Build GRCh38
rs755015234 Chr.11: 7987590-7987590 on Build GRCh38
rs1042095845 Chr.11: 7987651-7987651 on Build GRCh38
rs370643885 Chr.11: 7987376-7987376 on Build GRCh38
rs747455483 Chr.11: 7987490-7987490 on Build GRCh38
rs765746768 Chr.11: 7987405-7987405 on Build GRCh38
rs375415343 Chr.11: 7987577-7987577 on Build GRCh38
rs1038981151 Chr.11: 7987434-7987434 on Build GRCh38
rs748371751 Chr.11: 7987350-7987350 on Build GRCh38
rs751937805 Chr.11: 7987271-7987271 on Build GRCh38
rs903298030 Chr.11: 7987192-7987192 on Build GRCh38
rs773422499 Chr.11: 7987546-7987557 on Build GRCh38
rs748122436 Chr.11: 7987414-7987414 on Build GRCh38
rs200904167 Chr.11: 7987579-7987579 on Build GRCh38
rs750593385 Chr.11: 7987332-7987332 on Build GRCh38
rs771964348 Chr.11: 7987344-7987344 on Build GRCh38
rs772240804 Chr.11: 7987500-7987505 on Build GRCh38
rs772055960 Chr.11: 7987306-7987306 on Build GRCh38
rs960269595 Chr.11: 7987286-7987286 on Build GRCh38
rs374733620 Chr.11: 7987622-7987622 on Build GRCh38
rs757388352 Chr.11: 7987483-7987483 on Build GRCh38
rs370284767 Chr.11: 7987339-7987339 on Build GRCh38
rs774619981 Chr.11: 7987434-7987435 on Build GRCh38
rs867328795 Chr.11: 7987571-7987571 on Build GRCh38
rs749554050 Chr.11: 7987382-7987382 on Build GRCh38
rs755763578 Chr.11: 7987277-7987277 on Build GRCh38
rs765020257 Chr.11: 7987575-7987575 on Build GRCh38
rs753754616 Chr.11: 7987481-7987481 on Build GRCh38
rs369055254 Chr.11: 7987703-7987703 on Build GRCh38
rs200048982 Chr.11: 7987371-7987371 on Build GRCh38
rs748631927 Chr.11: 7987596-7987596 on Build GRCh38
rs745376176 Chr.11: 7987296-7987296 on Build GRCh38
rs12421289 Chr.11: 7987694-7987694 on Build GRCh38
rs762123131 Chr.11: 7987328-7987328 on Build GRCh38
rs750034387 Chr.11: 7987401-7987401 on Build GRCh38
rs753948836 Chr.11: 7987572-7987572 on Build GRCh38
rs377017616 Chr.11: 7987336-7987336 on Build GRCh38
rs757654224 Chr.11: 7987685-7987685 on Build GRCh38
rs745521038 Chr.11: 7987556-7987556 on Build GRCh38
rs149749747 Chr.11: 7987691-7987691 on Build GRCh38
rs370821155 Chr.11: 7987266-7987266 on Build GRCh38
rs749564196 Chr.11: 7987337-7987339 on Build GRCh38
rs746100232 Chr.11: 7987485-7987485 on Build GRCh38
rs113790013 Chr.11: 7987456-7987456 on Build GRCh38
rs769094533 Chr.11: 7987344-7987344 on Build GRCh38
rs372055041 Chr.11: 7987514-7987514 on Build GRCh38
rs748638890 Chr.11: 7987549-7987549 on Build GRCh38
rs746689810 Chr.11: 7987607-7987607 on Build GRCh38
rs986515217 Chr.11: 7987267-7987267 on Build GRCh38
rs749911888 Chr.11: 7987368-7987368 on Build GRCh38
rs766364853 Chr.11: 7987604-7987609 on Build GRCh38
rs148453570 Chr.11: 7987373-7987373 on Build GRCh38
rs569582445 Chr.11: 7987461-7987461 on Build GRCh38
rs752787711 Chr.11: 7987664-7987664 on Build GRCh38
rs373853999 Chr.11: 7987310-7987310 on Build GRCh38
rs771918883 Chr.11: 7987559-7987559 on Build GRCh38
rs771826730 Chr.11: 7987601-7987601 on Build GRCh38
rs1026730373 Chr.11: 7987563-7987563 on Build GRCh38
rs781629866 Chr.11: 7987544-7987544 on Build GRCh38
rs761792741 Chr.11: 7987612-7987612 on Build GRCh38
rs775988880 Chr.11: 7987311-7987311 on Build GRCh38
rs756614698 Chr.11: 7987670-7987670 on Build GRCh38
rs369481212 Chr.11: 7987610-7987610 on Build GRCh38
rs768898165 Chr.11: 7987324-7987324 on Build GRCh38
rs200663616 Chr.11: 7987349-7987349 on Build GRCh38
rs534364559 Chr.11: 7987489-7987489 on Build GRCh38
rs764907812 Chr.11: 7987367-7987367 on Build GRCh38
rs759115707 Chr.11: 7987508-7987508 on Build GRCh38
rs760687210 Chr.11: 7987516-7987516 on Build GRCh38
rs770133621 Chr.11: 7987553-7987553 on Build GRCh38
rs774504825 Chr.11: 7987386-7987386 on Build GRCh38
rs765125440 Chr.11: 7987178-7987178 on Build GRCh38
rs773397905 Chr.11: 7987358-7987358 on Build GRCh38
rs917646916 Chr.11: 7987593-7987593 on Build GRCh38
rs748001646 Chr.11: 7987380-7987380 on Build GRCh38
rs775231750 Chr.11: 7987604-7987604 on Build GRCh38
rs762074883 Chr.11: 7987452-7987469 on Build GRCh38
rs761656459 Chr.11: 7987574-7987574 on Build GRCh38
rs778853342 Chr.11: 7987428-7987428 on Build GRCh38
rs72847449 Chr.11: 7987224-7987224 on Build GRCh38
rs759772480 Chr.11: 7987628-7987628 on Build GRCh38
rs776665566 Chr.11: 7987326-7987326 on Build GRCh38
rs745746880 Chr.11: 7987302-7987302 on Build GRCh38
rs753162508 Chr.11: 7987364-7987364 on Build GRCh38
rs778195094 Chr.11: 7987679-7987679 on Build GRCh38
rs376569697 Chr.11: 7987440-7987440 on Build GRCh38
rs552010351 Chr.11: 7987475-7987475 on Build GRCh38
rs772436208 Chr.11: 7987431-7987431 on Build GRCh38
rs781644785 Chr.11: 7987337-7987337 on Build GRCh38
rs763177140 Chr.11: 7987359-7987359 on Build GRCh38
rs553134625 Chr.11: 7987519-7987519 on Build GRCh38
rs777342205 Chr.11: 7987280-7987280 on Build GRCh38
rs199691512 Chr.11: 7987333-7987333 on Build GRCh38
rs367735033 Chr.11: 7987362-7987362 on Build GRCh38
rs752615692 Chr.11: 7987471-7987471 on Build GRCh38

View Full Product Details