Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 28330138-28330586 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
METTL15
Entrez Gene ID
196074
Gene Name
methyltransferase like 15
Gene Aliases
METT5D1
UniGene
Hs.243326
Gene Chromosome Location
Chr.11: 28108251-28527041 on Build GRCh38
Transcripts
NM_001113528.1 NM_001297775.1 NM_152636.2 XM_011519935.2 XM_011519937.2 XM_011519940.2 XM_011519941.2 XM_017017297.1 XM_017017298.1 XM_017017299.1 XM_017017300.1 XM_017017302.1 XM_017017303.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4599657 c.829T>A p.S277T Chr.11: 28330446-28330446 on Build GRCh38
2113586 c.836A>G p.H279R Chr.11: 28330453-28330453 on Build GRCh38
926180 c.841G>A p.A281T Chr.11: 28330458-28330458 on Build GRCh38
2113592 c.923A>G p.K308R Chr.11: 28330540-28330540 on Build GRCh38
6998049 c.940G>A p.G314S Chr.11: 28330557-28330557 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371016812 Chr.11: 28330561-28330561 on Build GRCh38
rs369114838 Chr.11: 28330425-28330425 on Build GRCh38
rs934986967 Chr.11: 28330222-28330222 on Build GRCh38
rs540488675 Chr.11: 28330354-28330354 on Build GRCh38
rs915847039 Chr.11: 28330388-28330388 on Build GRCh38
rs572903336 Chr.11: 28330474-28330474 on Build GRCh38
rs754756005 Chr.11: 28330520-28330520 on Build GRCh38
rs562027928 Chr.11: 28330360-28330360 on Build GRCh38
rs764274954 Chr.11: 28330468-28330468 on Build GRCh38
rs769313609 Chr.11: 28330555-28330555 on Build GRCh38
rs1051012655 Chr.11: 28330311-28330311 on Build GRCh38
rs769308866 Chr.11: 28330406-28330406 on Build GRCh38
rs377363969 Chr.11: 28330554-28330554 on Build GRCh38
rs11602768 Chr.11: 28330239-28330239 on Build GRCh38
rs144364236 Chr.11: 28330446-28330446 on Build GRCh38
rs1049528180 Chr.11: 28330526-28330526 on Build GRCh38
rs374959611 Chr.11: 28330385-28330385 on Build GRCh38
rs900764357 Chr.11: 28330499-28330499 on Build GRCh38
rs779961168 Chr.11: 28330514-28330514 on Build GRCh38
rs959893715 Chr.11: 28330337-28330337 on Build GRCh38
rs868096964 Chr.11: 28330447-28330447 on Build GRCh38
rs748437873 Chr.11: 28330463-28330463 on Build GRCh38
rs772919212 Chr.11: 28330558-28330558 on Build GRCh38
rs753892525 Chr.11: 28330471-28330471 on Build GRCh38
rs559762203 Chr.11: 28330487-28330487 on Build GRCh38
rs760001083 Chr.11: 28330426-28330426 on Build GRCh38
rs111934305 Chr.11: 28330450-28330450 on Build GRCh38
rs932145783 Chr.11: 28330305-28330305 on Build GRCh38
rs934400812 Chr.11: 28330428-28330428 on Build GRCh38
rs890650035 Chr.11: 28330315-28330315 on Build GRCh38
rs992685799 Chr.11: 28330386-28330386 on Build GRCh38
rs182659010 Chr.11: 28330482-28330482 on Build GRCh38
rs1027706275 Chr.11: 28330279-28330279 on Build GRCh38
rs199965348 Chr.11: 28330530-28330530 on Build GRCh38
rs900481543 Chr.11: 28330245-28330246 on Build GRCh38
rs780739721 Chr.11: 28330540-28330540 on Build GRCh38
rs748806950 Chr.11: 28330560-28330560 on Build GRCh38
rs549371768 Chr.11: 28330519-28330519 on Build GRCh38
rs77411268 Chr.11: 28330175-28330175 on Build GRCh38
rs768258722 Chr.11: 28330350-28330350 on Build GRCh38
rs890666686 Chr.11: 28330198-28330198 on Build GRCh38
rs188098216 Chr.11: 28330498-28330498 on Build GRCh38
rs776291740 Chr.11: 28330371-28330371 on Build GRCh38
rs774926728 Chr.11: 28330440-28330440 on Build GRCh38
rs773583995 Chr.11: 28330161-28330161 on Build GRCh38
rs116840126 Chr.11: 28330256-28330256 on Build GRCh38
rs752912889 Chr.11: 28330453-28330453 on Build GRCh38
rs1009142820 Chr.11: 28330199-28330199 on Build GRCh38
rs1015834691 Chr.11: 28330318-28330318 on Build GRCh38
rs11030280 Chr.11: 28330416-28330416 on Build GRCh38

View Full Product Details