Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 59104882-59105428 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNB2
Entrez Gene ID
9133
Gene Name
cyclin B2
Gene Aliases
HsT17299
UniGene
Hs.194698
Gene Chromosome Location
Chr.15: 59105085-59125045 on Build GRCh38
Transcripts
NM_004701.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1200145 c.16C>T p.R6C Chr.15: 59105284-59105284 on Build GRCh38
5500366 c.21G>A p.P7P Chr.15: 59105289-59105289 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs144017866 Chr.15: 59105166-59105166 on Build GRCh38
rs574501270 Chr.15: 59105172-59105172 on Build GRCh38
rs371252673 Chr.15: 59105087-59105087 on Build GRCh38
rs184832385 Chr.15: 59105194-59105194 on Build GRCh38
rs779655115 Chr.15: 59105114-59105114 on Build GRCh38
rs773160088 Chr.15: 59105273-59105273 on Build GRCh38
rs865823239 Chr.15: 59105224-59105224 on Build GRCh38
rs963944354 Chr.15: 59105185-59105185 on Build GRCh38
rs761534519 Chr.15: 59105304-59105304 on Build GRCh38
rs182277915 Chr.15: 59104905-59104905 on Build GRCh38
rs946092254 Chr.15: 59105122-59105122 on Build GRCh38
rs771940160 Chr.15: 59105227-59105227 on Build GRCh38
rs28383493 Chr.15: 59105209-59105209 on Build GRCh38
rs760520580 Chr.15: 59105289-59105289 on Build GRCh38
rs536497004 Chr.15: 59105177-59105177 on Build GRCh38
rs564222788 Chr.15: 59105197-59105197 on Build GRCh38
rs1056343762 Chr.15: 59105143-59105143 on Build GRCh38
rs778279942 Chr.15: 59105238-59105238 on Build GRCh38
rs372194721 Chr.15: 59105326-59105326 on Build GRCh38
rs11548099 Chr.15: 59105244-59105244 on Build GRCh38
rs922548053 Chr.15: 59105234-59105234 on Build GRCh38
rs766730272 Chr.15: 59104942-59104943 on Build GRCh38
rs368290983 Chr.15: 59105340-59105340 on Build GRCh38
rs540025884 Chr.15: 59105082-59105082 on Build GRCh38
rs1046722432 Chr.15: 59105358-59105358 on Build GRCh38
rs754652242 Chr.15: 59105158-59105158 on Build GRCh38
rs766621735 Chr.15: 59105306-59105306 on Build GRCh38
rs535585560 Chr.15: 59104974-59104974 on Build GRCh38
rs759107039 Chr.15: 59105261-59105261 on Build GRCh38
rs964263102 Chr.15: 59105106-59105106 on Build GRCh38
rs28383492 Chr.15: 59105183-59105183 on Build GRCh38
rs755135656 Chr.15: 59105309-59105309 on Build GRCh38
rs888124526 Chr.15: 59105402-59105402 on Build GRCh38
rs763932078 Chr.15: 59105291-59105291 on Build GRCh38
rs558126278 Chr.15: 59105115-59105115 on Build GRCh38
rs555085439 Chr.15: 59105181-59105181 on Build GRCh38
rs775535567 Chr.15: 59105281-59105281 on Build GRCh38
rs779526035 Chr.15: 59105345-59105345 on Build GRCh38
rs550199054 Chr.15: 59104955-59104955 on Build GRCh38
rs568191230 Chr.15: 59104956-59104956 on Build GRCh38
rs199506419 Chr.15: 59105239-59105239 on Build GRCh38
rs993914194 Chr.15: 59105050-59105050 on Build GRCh38
rs113384931 Chr.15: 59105337-59105337 on Build GRCh38
rs572275866 Chr.15: 59105039-59105039 on Build GRCh38
rs562910667 Chr.15: 59105096-59105097 on Build GRCh38
rs753651235 Chr.15: 59105303-59105303 on Build GRCh38
rs537844954 Chr.15: 59104943-59104943 on Build GRCh38
rs201180257 Chr.15: 59104943-59104944 on Build GRCh38
rs186802804 Chr.15: 59104927-59104927 on Build GRCh38
rs750920747 Chr.15: 59104996-59104996 on Build GRCh38
rs770235738 Chr.15: 59105236-59105236 on Build GRCh38
rs191642888 Chr.15: 59105008-59105008 on Build GRCh38
rs774104897 Chr.15: 59105250-59105250 on Build GRCh38
rs776616577 Chr.15: 59105333-59105333 on Build GRCh38
rs771457855 Chr.15: 59105266-59105266 on Build GRCh38
rs367760992 Chr.15: 59105387-59105387 on Build GRCh38
rs547438223 Chr.15: 59105347-59105347 on Build GRCh38
rs911073445 Chr.15: 59105100-59105100 on Build GRCh38
rs191048390 Chr.15: 59104978-59104978 on Build GRCh38
rs1025422269 Chr.15: 59105051-59105051 on Build GRCh38
rs541919685 Chr.15: 59105176-59105176 on Build GRCh38
rs565844161 Chr.15: 59105357-59105357 on Build GRCh38
rs576713566 Chr.15: 59105142-59105142 on Build GRCh38
rs906178915 Chr.15: 59105169-59105169 on Build GRCh38
rs926069483 Chr.15: 59105137-59105137 on Build GRCh38
rs759397642 Chr.15: 59105320-59105320 on Build GRCh38
rs894646233 Chr.15: 59105145-59105145 on Build GRCh38
rs562502263 Chr.15: 59105170-59105170 on Build GRCh38
rs781735062 Chr.15: 59105226-59105226 on Build GRCh38
rs367606664 Chr.15: 59105230-59105230 on Build GRCh38
rs1014887347 Chr.15: 59105151-59105151 on Build GRCh38
rs549889117 Chr.15: 59105214-59105214 on Build GRCh38
rs201447729 Chr.15: 59105308-59105308 on Build GRCh38
rs952510776 Chr.15: 59105052-59105052 on Build GRCh38
rs976703351 Chr.15: 59105107-59105107 on Build GRCh38
rs552166989 Chr.15: 59105139-59105139 on Build GRCh38
rs543122162 Chr.15: 59104937-59104937 on Build GRCh38
rs532002854 Chr.15: 59105130-59105130 on Build GRCh38
rs80074818 Chr.15: 59104954-59104955 on Build GRCh38
rs748952223 Chr.15: 59105116-59105116 on Build GRCh38
rs375932622 Chr.15: 59105173-59105173 on Build GRCh38
rs936044132 Chr.15: 59105293-59105293 on Build GRCh38
rs768037854 Chr.15: 59104916-59104916 on Build GRCh38
rs113082333 Chr.15: 59104943-59104943 on Build GRCh38
rs376989309 Chr.15: 59105332-59105332 on Build GRCh38
rs1034595920 Chr.15: 59104940-59104940 on Build GRCh38

View Full Product Details