Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 31356455-31356913 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGAX
Entrez Gene ID
3687
Gene Name
integrin subunit alpha X
Gene Aliases
CD11C, SLEB6
UniGene
Hs.248472
Gene Chromosome Location
Chr.16: 31355134-31382997 on Build GRCh38
Transcripts
NM_000887.4 NM_001286375.1 XM_011545852.1 XM_011545854.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3509156 c.173C>A p.T58K Chr.16: 31356654-31356654 on Build GRCh38
5729579 c.151G>A p.V51I Chr.16: 31356632-31356632 on Build GRCh38
3509157 c.184C>T p.Q62* Chr.16: 31356665-31356665 on Build GRCh38
970201 c.183C>T p.N61N Chr.16: 31356664-31356664 on Build GRCh38
1717348 c.151G>T p.V51F Chr.16: 31356632-31356632 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766218159 Chr.16: 31356761-31356761 on Build GRCh38
rs775681224 Chr.16: 31356734-31356734 on Build GRCh38
rs747591972 Chr.16: 31356716-31356716 on Build GRCh38
rs747281460 Chr.16: 31356739-31356739 on Build GRCh38
rs1050095004 Chr.16: 31356841-31356841 on Build GRCh38
rs7190997 Chr.16: 31356857-31356857 on Build GRCh38
rs566562365 Chr.16: 31356621-31356621 on Build GRCh38
rs1021772825 Chr.16: 31356512-31356512 on Build GRCh38
rs776985312 Chr.16: 31356746-31356746 on Build GRCh38
rs761842960 Chr.16: 31356646-31356646 on Build GRCh38
rs890947082 Chr.16: 31356684-31356684 on Build GRCh38
rs889242734 Chr.16: 31356488-31356488 on Build GRCh38
rs190655496 Chr.16: 31356793-31356793 on Build GRCh38
rs141210290 Chr.16: 31356886-31356886 on Build GRCh38
rs771371278 Chr.16: 31356611-31356611 on Build GRCh38
rs756979759 Chr.16: 31356756-31356756 on Build GRCh38
rs776790495 Chr.16: 31356641-31356641 on Build GRCh38
rs569113470 Chr.16: 31356839-31356839 on Build GRCh38
rs369769040 Chr.16: 31356671-31356671 on Build GRCh38
rs902388272 Chr.16: 31356865-31356865 on Build GRCh38
rs528529833 Chr.16: 31356495-31356495 on Build GRCh38
rs369539640 Chr.16: 31356584-31356584 on Build GRCh38
rs570550660 Chr.16: 31356802-31356802 on Build GRCh38
rs185801809 Chr.16: 31356788-31356788 on Build GRCh38
rs997532084 Chr.16: 31356477-31356477 on Build GRCh38
rs1050490512 Chr.16: 31356663-31356663 on Build GRCh38
rs550396989 Chr.16: 31356825-31356825 on Build GRCh38
rs1042618694 Chr.16: 31356858-31356858 on Build GRCh38
rs267604518 Chr.16: 31356636-31356636 on Build GRCh38
rs140622291 Chr.16: 31356562-31356562 on Build GRCh38
rs752826732 Chr.16: 31356718-31356718 on Build GRCh38
rs749817415 Chr.16: 31356594-31356594 on Build GRCh38
rs566306631 Chr.16: 31356873-31356873 on Build GRCh38
rs1042999022 Chr.16: 31356874-31356874 on Build GRCh38
rs780693689 Chr.16: 31356699-31356699 on Build GRCh38
rs114590772 Chr.16: 31356552-31356552 on Build GRCh38
rs371311261 Chr.16: 31356623-31356623 on Build GRCh38
rs554298187 Chr.16: 31356620-31356620 on Build GRCh38
rs145547326 Chr.16: 31356518-31356518 on Build GRCh38
rs536230535 Chr.16: 31356575-31356575 on Build GRCh38
rs558648760 Chr.16: 31356889-31356889 on Build GRCh38
rs375890125 Chr.16: 31356669-31356669 on Build GRCh38
rs772530717 Chr.16: 31356731-31356731 on Build GRCh38
rs908307828 Chr.16: 31356791-31356791 on Build GRCh38
rs373555641 Chr.16: 31356675-31356675 on Build GRCh38
rs758577391 Chr.16: 31356792-31356792 on Build GRCh38
rs754538346 Chr.16: 31356698-31356698 on Build GRCh38
rs542203063 Chr.16: 31356782-31356782 on Build GRCh38
rs575057406 Chr.16: 31356766-31356766 on Build GRCh38
rs532178889 Chr.16: 31356531-31356531 on Build GRCh38
rs771184675 Chr.16: 31356879-31356879 on Build GRCh38
rs1035921411 Chr.16: 31356551-31356551 on Build GRCh38
rs754555917 Chr.16: 31356577-31356577 on Build GRCh38
rs114050460 Chr.16: 31356483-31356483 on Build GRCh38
rs369540600 Chr.16: 31356745-31356745 on Build GRCh38
rs898378235 Chr.16: 31356665-31356665 on Build GRCh38
rs762039100 Chr.16: 31356748-31356748 on Build GRCh38
rs966794289 Chr.16: 31356723-31356723 on Build GRCh38
rs952530460 Chr.16: 31356567-31356567 on Build GRCh38
rs778488848 Chr.16: 31356747-31356747 on Build GRCh38
rs560325525 Chr.16: 31356787-31356787 on Build GRCh38
rs983756313 Chr.16: 31356781-31356781 on Build GRCh38
rs751255361 Chr.16: 31356693-31356693 on Build GRCh38
rs764269268 Chr.16: 31356639-31356639 on Build GRCh38
rs774704556 Chr.16: 31356615-31356615 on Build GRCh38
rs146557303 Chr.16: 31356820-31356820 on Build GRCh38
rs200902675 Chr.16: 31356677-31356677 on Build GRCh38
rs922427407 Chr.16: 31356826-31356826 on Build GRCh38
rs374476079 Chr.16: 31356722-31356722 on Build GRCh38
rs939847085 Chr.16: 31356595-31356595 on Build GRCh38
rs1045466322 Chr.16: 31356563-31356563 on Build GRCh38
rs765384344 Chr.16: 31356757-31356757 on Build GRCh38
rs376435369 Chr.16: 31356799-31356799 on Build GRCh38
rs529993130 Chr.16: 31356844-31356844 on Build GRCh38
rs754288584 Chr.16: 31356536-31356536 on Build GRCh38
rs1008499513 Chr.16: 31356691-31356691 on Build GRCh38
rs145679140 Chr.16: 31356760-31356760 on Build GRCh38
rs751255782 Chr.16: 31356764-31356764 on Build GRCh38
rs1012073950 Chr.16: 31356489-31356489 on Build GRCh38
rs779813016 Chr.16: 31356574-31356574 on Build GRCh38
rs1032457535 Chr.16: 31356894-31356894 on Build GRCh38
rs143407533 Chr.16: 31356719-31356719 on Build GRCh38

View Full Product Details