Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 20737591-20738177 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EEF1AKMT1
Entrez Gene ID
221143
Gene Name
eukaryotic translation elongation factor 1 alpha lysine methyltransferase 1
Gene Aliases
ESP13, N6AMT2
UniGene
Hs.26674
Gene Chromosome Location
Chr.13: 20728931-20773958 on Build GRCh38
Transcripts
NM_001318939.1 NM_174928.2 NR_134934.1 XM_017020432.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3711013 c.150G>A p.L50L Chr.13: 20737800-20737800 on Build GRCh38
1659191 c.164A>T p.Y55F Chr.13: 20737786-20737786 on Build GRCh38
6857295 c.225C>T p.G75G Chr.13: 20737725-20737725 on Build GRCh38
6457538 c.167G>A p.S56N Chr.13: 20737783-20737783 on Build GRCh38
1365762 c.158T>G p.F53C Chr.13: 20737792-20737792 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778980138 Chr.13: 20737823-20737823 on Build GRCh38
rs550563497 Chr.13: 20737749-20737749 on Build GRCh38
rs1006747148 Chr.13: 20737779-20737779 on Build GRCh38
rs936727154 Chr.13: 20737948-20737948 on Build GRCh38
rs994427943 Chr.13: 20738052-20738052 on Build GRCh38
rs918061601 Chr.13: 20737626-20737626 on Build GRCh38
rs891191811 Chr.13: 20737677-20737677 on Build GRCh38
rs762056820 Chr.13: 20737739-20737739 on Build GRCh38
rs578257418 Chr.13: 20738107-20738107 on Build GRCh38
rs371456921 Chr.13: 20737844-20737844 on Build GRCh38
rs532937395 Chr.13: 20737871-20737871 on Build GRCh38
rs762522651 Chr.13: 20737714-20737714 on Build GRCh38
rs1006797673 Chr.13: 20737992-20737992 on Build GRCh38
rs901052517 Chr.13: 20738043-20738043 on Build GRCh38
rs556549691 Chr.13: 20738008-20738008 on Build GRCh38
rs751899750 Chr.13: 20737731-20737731 on Build GRCh38
rs756279022 Chr.13: 20737791-20737791 on Build GRCh38
rs918125025 Chr.13: 20737903-20737903 on Build GRCh38
rs192177615 Chr.13: 20737979-20737979 on Build GRCh38
rs1001010804 Chr.13: 20738037-20738037 on Build GRCh38
rs903953653 Chr.13: 20738011-20738011 on Build GRCh38
rs549065843 Chr.13: 20737933-20737933 on Build GRCh38
rs969005217 Chr.13: 20738127-20738127 on Build GRCh38
rs372597695 Chr.13: 20737672-20737672 on Build GRCh38
rs567025741 Chr.13: 20737976-20737976 on Build GRCh38
rs732414 Chr.13: 20737838-20737838 on Build GRCh38
rs777585500 Chr.13: 20737673-20737673 on Build GRCh38
rs764394010 Chr.13: 20737728-20737728 on Build GRCh38
rs1006853305 Chr.13: 20737980-20737980 on Build GRCh38
rs889764415 Chr.13: 20737961-20737961 on Build GRCh38
rs949534138 Chr.13: 20737641-20737641 on Build GRCh38
rs765061369 Chr.13: 20738025-20738025 on Build GRCh38
rs1019465705 Chr.13: 20737814-20737814 on Build GRCh38
rs1013881502 Chr.13: 20738087-20738087 on Build GRCh38
rs563074279 Chr.13: 20737689-20737689 on Build GRCh38
rs142274093 Chr.13: 20737802-20737802 on Build GRCh38
rs952416969 Chr.13: 20737870-20737870 on Build GRCh38
rs1039723016 Chr.13: 20738005-20738005 on Build GRCh38
rs750034260 Chr.13: 20738108-20738108 on Build GRCh38
rs750955070 Chr.13: 20737786-20737786 on Build GRCh38
rs775133884 Chr.13: 20737712-20737712 on Build GRCh38
rs969121284 Chr.13: 20737850-20737850 on Build GRCh38
rs759110701 Chr.13: 20737695-20737695 on Build GRCh38
rs755178526 Chr.13: 20737819-20737819 on Build GRCh38
rs768141703 Chr.13: 20737771-20737771 on Build GRCh38
rs766622716 Chr.13: 20737828-20737828 on Build GRCh38
rs750403359 Chr.13: 20738058-20738058 on Build GRCh38
rs891159277 Chr.13: 20737970-20737973 on Build GRCh38
rs781162342 Chr.13: 20737847-20737847 on Build GRCh38
rs566260003 Chr.13: 20737878-20737878 on Build GRCh38
rs980452518 Chr.13: 20738133-20738133 on Build GRCh38
rs773223226 Chr.13: 20737707-20737707 on Build GRCh38
rs926933928 Chr.13: 20737873-20737873 on Build GRCh38
rs1025820707 Chr.13: 20737864-20737864 on Build GRCh38
rs1031228318 Chr.13: 20737750-20737750 on Build GRCh38
rs980485031 Chr.13: 20737869-20737869 on Build GRCh38
rs746942673 Chr.13: 20737679-20737679 on Build GRCh38
rs567109983 Chr.13: 20737946-20737946 on Build GRCh38
rs762950260 Chr.13: 20737823-20737827 on Build GRCh38
rs370181923 Chr.13: 20737688-20737688 on Build GRCh38
rs924036681 Chr.13: 20737947-20737947 on Build GRCh38
rs956976391 Chr.13: 20738074-20738074 on Build GRCh38
rs957188521 Chr.13: 20737828-20737828 on Build GRCh38
rs933257748 Chr.13: 20737945-20737945 on Build GRCh38
rs1042857443 Chr.13: 20737656-20737656 on Build GRCh38
rs1053741882 Chr.13: 20737957-20737957 on Build GRCh38
rs919169291 Chr.13: 20737914-20737914 on Build GRCh38
rs201315425 Chr.13: 20737697-20737697 on Build GRCh38
rs776517864 Chr.13: 20737693-20737693 on Build GRCh38
rs949590465 Chr.13: 20737908-20737908 on Build GRCh38
rs533684293 Chr.13: 20737929-20737929 on Build GRCh38
rs117435800 Chr.13: 20737900-20737900 on Build GRCh38
rs780217440 Chr.13: 20737800-20737800 on Build GRCh38

View Full Product Details