Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 34254893-34255457 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCL2
Entrez Gene ID
6347
Gene Name
C-C motif chemokine ligand 2
Gene Aliases
GDCF-2, HC11, HSMCR30, MCAF, MCP-1, MCP1, SCYA2, SMC-CF
UniGene
Hs.303649
Gene Chromosome Location
Chr.17: 34255277-34257203 on Build GRCh38
Transcripts
NM_002982.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4768869 c.70C>T p.Q24* Chr.17: 34255419-34255419 on Build GRCh38
1200133 c.16G>A p.A6T Chr.17: 34255365-34255365 on Build GRCh38
4839061 c.9C>A p.V3V Chr.17: 34255358-34255358 on Build GRCh38
1200134 c.7G>A p.V3I Chr.17: 34255356-34255356 on Build GRCh38
4500207 c.55C>T p.P19S Chr.17: 34255404-34255404 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs950082296 Chr.17: 34255218-34255218 on Build GRCh38
rs753449998 Chr.17: 34255371-34255371 on Build GRCh38
rs184897210 Chr.17: 34255269-34255269 on Build GRCh38
rs760955962 Chr.17: 34255258-34255258 on Build GRCh38
rs371522420 Chr.17: 34255186-34255186 on Build GRCh38
rs11575010 Chr.17: 34254920-34254920 on Build GRCh38
rs750066113 Chr.17: 34255285-34255285 on Build GRCh38
rs74988151 Chr.17: 34255217-34255217 on Build GRCh38
rs776841122 Chr.17: 34255334-34255334 on Build GRCh38
rs767198628 Chr.17: 34255282-34255282 on Build GRCh38
rs995236627 Chr.17: 34255029-34255029 on Build GRCh38
rs778276611 Chr.17: 34255396-34255396 on Build GRCh38
rs953850275 Chr.17: 34255099-34255099 on Build GRCh38
rs1007290590 Chr.17: 34254987-34254987 on Build GRCh38
rs993298607 Chr.17: 34254924-34254924 on Build GRCh38
rs2857656 Chr.17: 34254988-34254988 on Build GRCh38
rs751464334 Chr.17: 34255274-34255274 on Build GRCh38
rs762685952 Chr.17: 34255270-34255270 on Build GRCh38
rs762750838 Chr.17: 34255335-34255335 on Build GRCh38
rs748658453 Chr.17: 34255387-34255387 on Build GRCh38
rs749627296 Chr.17: 34255300-34255300 on Build GRCh38
rs887561914 Chr.17: 34254982-34254982 on Build GRCh38
rs766065319 Chr.17: 34255370-34255370 on Build GRCh38
rs1015685883 Chr.17: 34254986-34254986 on Build GRCh38
rs755877280 Chr.17: 34255295-34255295 on Build GRCh38
rs773321785 Chr.17: 34255329-34255329 on Build GRCh38
rs763852250 Chr.17: 34255338-34255338 on Build GRCh38
rs1048872723 Chr.17: 34254940-34254940 on Build GRCh38
rs555689420 Chr.17: 34255192-34255192 on Build GRCh38
rs995205364 Chr.17: 34255402-34255402 on Build GRCh38
rs755719349 Chr.17: 34255360-34255360 on Build GRCh38
rs144450837 Chr.17: 34255386-34255386 on Build GRCh38
rs760591942 Chr.17: 34254970-34254970 on Build GRCh38
rs531047832 Chr.17: 34255109-34255109 on Build GRCh38
rs938810884 Chr.17: 34255302-34255302 on Build GRCh38
rs748665636 Chr.17: 34255323-34255323 on Build GRCh38
rs748196460 Chr.17: 34255247-34255247 on Build GRCh38
rs567665641 Chr.17: 34255136-34255136 on Build GRCh38
rs535705688 Chr.17: 34255154-34255154 on Build GRCh38
rs374418259 Chr.17: 34255405-34255405 on Build GRCh38
rs369439824 Chr.17: 34255313-34255313 on Build GRCh38
rs532056815 Chr.17: 34255119-34255126 on Build GRCh38
rs1028115260 Chr.17: 34255065-34255065 on Build GRCh38
rs957711087 Chr.17: 34255206-34255206 on Build GRCh38
rs991577091 Chr.17: 34255211-34255211 on Build GRCh38
rs774898999 Chr.17: 34255404-34255404 on Build GRCh38
rs372302908 Chr.17: 34254939-34254939 on Build GRCh38
rs764986697 Chr.17: 34255261-34255261 on Build GRCh38
rs886691976 Chr.17: 34254957-34254957 on Build GRCh38
rs965751281 Chr.17: 34254995-34254995 on Build GRCh38
rs3917904 Chr.17: 34254951-34254951 on Build GRCh38
rs772062774 Chr.17: 34255253-34255253 on Build GRCh38
rs1020627118 Chr.17: 34255416-34255416 on Build GRCh38
rs773409942 Chr.17: 34255254-34255254 on Build GRCh38
rs758556069 Chr.17: 34255266-34255266 on Build GRCh38
rs749661142 Chr.17: 34255263-34255264 on Build GRCh38
rs747023179 Chr.17: 34255406-34255406 on Build GRCh38
rs370016526 Chr.17: 34255344-34255344 on Build GRCh38
rs774754806 Chr.17: 34255221-34255221 on Build GRCh38
rs771283629 Chr.17: 34255338-34255339 on Build GRCh38
rs780602243 Chr.17: 34255200-34255200 on Build GRCh38
rs779871993 Chr.17: 34255298-34255298 on Build GRCh38
rs756777436 Chr.17: 34255279-34255279 on Build GRCh38
rs967589920 Chr.17: 34255138-34255138 on Build GRCh38
rs779422035 Chr.17: 34255315-34255315 on Build GRCh38
rs749900916 Chr.17: 34255347-34255347 on Build GRCh38
rs758954928 Chr.17: 34255392-34255392 on Build GRCh38
rs11575011 Chr.17: 34255228-34255228 on Build GRCh38
rs755360396 Chr.17: 34255373-34255373 on Build GRCh38
rs749788226 Chr.17: 34255215-34255215 on Build GRCh38
rs898151976 Chr.17: 34255353-34255353 on Build GRCh38
rs997163316 Chr.17: 34255049-34255049 on Build GRCh38
rs774337603 Chr.17: 34255340-34255340 on Build GRCh38
rs977621785 Chr.17: 34255177-34255177 on Build GRCh38
rs141271352 Chr.17: 34255332-34255332 on Build GRCh38
rs550669939 Chr.17: 34255119-34255119 on Build GRCh38
rs557966833 Chr.17: 34255328-34255328 on Build GRCh38
rs534495781 Chr.17: 34255273-34255273 on Build GRCh38
rs11575012 Chr.17: 34255231-34255231 on Build GRCh38

View Full Product Details