Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81845707-81846288 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
P4HB
Entrez Gene ID
5034
Gene Name
prolyl 4-hydroxylase subunit beta
Gene Aliases
CLCRP1, DSI, ERBA2L, GIT, P4Hbeta, PDI, PDIA1, PHDB, PO4DB, PO4HB, PROHB
UniGene
Hs.464336
Gene Chromosome Location
Chr.17: 81843158-81860668 on Build GRCh38
Transcripts
NM_000918.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3932780 c.1132G>T p.E378* Chr.17: 81845916-81845916 on Build GRCh38
1710896 c.1096A>G p.K366E Chr.17: 81845952-81845952 on Build GRCh38
6223347 c.1083G>T p.P361P Chr.17: 81845965-81845965 on Build GRCh38
6404709 c.1063C>G p.L355V Chr.17: 81845985-81845985 on Build GRCh38
3932781 c.1125G>A p.K375K Chr.17: 81845923-81845923 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751705426 Chr.17: 81845819-81845819 on Build GRCh38
rs776525144 Chr.17: 81845849-81845849 on Build GRCh38
rs573816855 Chr.17: 81846112-81846112 on Build GRCh38
rs200948725 Chr.17: 81846011-81846011 on Build GRCh38
rs766591711 Chr.17: 81846016-81846017 on Build GRCh38
rs958407480 Chr.17: 81846241-81846241 on Build GRCh38
rs1035407858 Chr.17: 81846238-81846238 on Build GRCh38
rs775129222 Chr.17: 81845746-81845746 on Build GRCh38
rs549230866 Chr.17: 81845810-81845810 on Build GRCh38
rs201103118 Chr.17: 81845822-81845822 on Build GRCh38
rs775484483 Chr.17: 81845886-81845886 on Build GRCh38
rs36027124 Chr.17: 81846185-81846186 on Build GRCh38
rs878862269 Chr.17: 81845744-81845744 on Build GRCh38
rs759952074 Chr.17: 81845910-81845910 on Build GRCh38
rs958237237 Chr.17: 81845851-81845851 on Build GRCh38
rs755470013 Chr.17: 81846025-81846025 on Build GRCh38
rs894074533 Chr.17: 81846150-81846150 on Build GRCh38
rs763394045 Chr.17: 81845887-81845888 on Build GRCh38
rs11558881 Chr.17: 81845896-81845896 on Build GRCh38
rs201874167 Chr.17: 81845735-81845735 on Build GRCh38
rs1007848780 Chr.17: 81845840-81845840 on Build GRCh38
rs866732299 Chr.17: 81845918-81845918 on Build GRCh38
rs200656778 Chr.17: 81846026-81846026 on Build GRCh38
rs201235381 Chr.17: 81845887-81845887 on Build GRCh38
rs933440983 Chr.17: 81846096-81846096 on Build GRCh38
rs1021563222 Chr.17: 81846198-81846198 on Build GRCh38
rs1038404144 Chr.17: 81846061-81846061 on Build GRCh38
rs766349876 Chr.17: 81845796-81845796 on Build GRCh38
rs786204843 Chr.17: 81845742-81845742 on Build GRCh38
rs769626015 Chr.17: 81845982-81845982 on Build GRCh38
rs773547908 Chr.17: 81845888-81845888 on Build GRCh38
rs373333137 Chr.17: 81845899-81845899 on Build GRCh38
rs1036636977 Chr.17: 81846068-81846068 on Build GRCh38
rs760303701 Chr.17: 81845761-81845761 on Build GRCh38
rs780345020 Chr.17: 81845913-81845913 on Build GRCh38
rs539390519 Chr.17: 81846004-81846004 on Build GRCh38
rs765746564 Chr.17: 81845860-81845861 on Build GRCh38
rs772898869 Chr.17: 81845760-81845760 on Build GRCh38
rs1051848859 Chr.17: 81846110-81846110 on Build GRCh38
rs571302330 Chr.17: 81846003-81846003 on Build GRCh38
rs200774916 Chr.17: 81845878-81845878 on Build GRCh38
rs142731141 Chr.17: 81845890-81845890 on Build GRCh38
rs373496790 Chr.17: 81846022-81846022 on Build GRCh38
rs147664712 Chr.17: 81845795-81845795 on Build GRCh38
rs200719772 Chr.17: 81845953-81845953 on Build GRCh38
rs759495273 Chr.17: 81845779-81845779 on Build GRCh38
rs200027839 Chr.17: 81846021-81846021 on Build GRCh38
rs537224834 Chr.17: 81846074-81846074 on Build GRCh38
rs970078070 Chr.17: 81846234-81846234 on Build GRCh38
rs779254472 Chr.17: 81846028-81846028 on Build GRCh38
rs1032319974 Chr.17: 81846122-81846122 on Build GRCh38
rs980222162 Chr.17: 81846253-81846253 on Build GRCh38
rs766702502 Chr.17: 81846023-81846023 on Build GRCh38
rs754170405 Chr.17: 81846024-81846024 on Build GRCh38
rs1009809141 Chr.17: 81846173-81846173 on Build GRCh38
rs577557755 Chr.17: 81846189-81846189 on Build GRCh38
rs771527087 Chr.17: 81845738-81845738 on Build GRCh38
rs751782521 Chr.17: 81846042-81846043 on Build GRCh38
rs981629638 Chr.17: 81846215-81846215 on Build GRCh38
rs755447281 Chr.17: 81845950-81845950 on Build GRCh38
rs761976262 Chr.17: 81845895-81845895 on Build GRCh38
rs780965978 Chr.17: 81845966-81845966 on Build GRCh38
rs555520209 Chr.17: 81846091-81846091 on Build GRCh38
rs938830413 Chr.17: 81845973-81845973 on Build GRCh38
rs924510885 Chr.17: 81845862-81845862 on Build GRCh38
rs897641718 Chr.17: 81846071-81846071 on Build GRCh38
rs11558882 Chr.17: 81845964-81845964 on Build GRCh38
rs757473328 Chr.17: 81845808-81845808 on Build GRCh38
rs377540224 Chr.17: 81845911-81845911 on Build GRCh38
rs762463974 Chr.17: 81845779-81845780 on Build GRCh38
rs766274545 Chr.17: 81845770-81845770 on Build GRCh38
rs772667046 Chr.17: 81845830-81845830 on Build GRCh38
rs762888503 Chr.17: 81846000-81846000 on Build GRCh38
rs570207013 Chr.17: 81845739-81845739 on Build GRCh38
rs759788630 Chr.17: 81845794-81845794 on Build GRCh38
rs201775800 Chr.17: 81845854-81845854 on Build GRCh38
rs1031560767 Chr.17: 81846156-81846156 on Build GRCh38
rs200244920 Chr.17: 81845839-81845839 on Build GRCh38
rs201350121 Chr.17: 81845965-81845965 on Build GRCh38
rs200298383 Chr.17: 81845765-81845765 on Build GRCh38
rs559438157 Chr.17: 81846126-81846126 on Build GRCh38
rs762973761 Chr.17: 81845772-81845772 on Build GRCh38
rs576163131 Chr.17: 81846067-81846067 on Build GRCh38
rs202079690 Chr.17: 81846029-81846029 on Build GRCh38
rs775151370 Chr.17: 81845998-81845998 on Build GRCh38
rs199695507 Chr.17: 81845861-81845861 on Build GRCh38
rs531086669 Chr.17: 81845805-81845805 on Build GRCh38
rs563771798 Chr.17: 81845801-81845801 on Build GRCh38
rs113417520 Chr.17: 81846113-81846113 on Build GRCh38
rs866190984 Chr.17: 81845917-81845917 on Build GRCh38
rs545379549 Chr.17: 81846213-81846213 on Build GRCh38
rs754368536 Chr.17: 81845945-81845945 on Build GRCh38
rs201832185 Chr.17: 81846012-81846012 on Build GRCh38
rs199828535 Chr.17: 81845825-81845825 on Build GRCh38
rs1007569336 Chr.17: 81846116-81846116 on Build GRCh38
rs201789542 Chr.17: 81845823-81845823 on Build GRCh38
rs907457808 Chr.17: 81845786-81845786 on Build GRCh38
rs373121788 Chr.17: 81845824-81845824 on Build GRCh38
rs765294380 Chr.17: 81845784-81845784 on Build GRCh38

View Full Product Details