Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 51825936-51826423 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FPR3
Entrez Gene ID
2359
Gene Name
formyl peptide receptor 3
Gene Aliases
FML2_HUMAN, FMLP-R-II, FMLPY, FPRH1, FPRH2, FPRL2, RMLP-R-I
UniGene
Hs.445466
Gene Chromosome Location
Chr.19: 51795146-51826207 on Build GRCh38
Transcripts
NM_002030.4 XM_011526687.2

SNP Detail

SNP ID Chromosome Location
rs917988328 Chr.19: 51826397-51826397 on Build GRCh38
rs974212809 Chr.19: 51826300-51826300 on Build GRCh38
rs1056332007 Chr.19: 51826066-51826066 on Build GRCh38
rs938937709 Chr.19: 51826036-51826036 on Build GRCh38
rs532385934 Chr.19: 51826144-51826144 on Build GRCh38
rs767453993 Chr.19: 51826262-51826262 on Build GRCh38
rs752581034 Chr.19: 51826387-51826387 on Build GRCh38
rs949496218 Chr.19: 51826402-51826402 on Build GRCh38
rs1025410700 Chr.19: 51826183-51826183 on Build GRCh38
rs1018495896 Chr.19: 51826266-51826266 on Build GRCh38
rs910917101 Chr.19: 51826263-51826263 on Build GRCh38
rs1001898419 Chr.19: 51826085-51826085 on Build GRCh38
rs964319352 Chr.19: 51826269-51826269 on Build GRCh38
rs1021734435 Chr.19: 51826087-51826087 on Build GRCh38
rs935520174 Chr.19: 51826384-51826384 on Build GRCh38
rs754759952 Chr.19: 51826230-51826230 on Build GRCh38
rs76780350 Chr.19: 51826178-51826178 on Build GRCh38
rs528457662 Chr.19: 51826362-51826362 on Build GRCh38
rs183793149 Chr.19: 51826080-51826080 on Build GRCh38
rs982372903 Chr.19: 51826400-51826400 on Build GRCh38
rs1016761562 Chr.19: 51826123-51826123 on Build GRCh38
rs540292850 Chr.19: 51826046-51826046 on Build GRCh38
rs998888661 Chr.19: 51826092-51826092 on Build GRCh38
rs1013554651 Chr.19: 51825958-51825958 on Build GRCh38
rs529907694 Chr.19: 51826290-51826290 on Build GRCh38
rs955135467 Chr.19: 51826227-51826227 on Build GRCh38
rs567281319 Chr.19: 51826323-51826323 on Build GRCh38
rs562814024 Chr.19: 51826186-51826186 on Build GRCh38
rs188114320 Chr.19: 51826308-51826308 on Build GRCh38
rs142943688 Chr.19: 51826023-51826023 on Build GRCh38

View Full Product Details