Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 92298109-92298702 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOL8
Entrez Gene ID
55035
Gene Name
nucleolar protein 8
Gene Aliases
C9orf34, NOP132, bA62C3.3, bA62C3.4
UniGene
Hs.442199
Gene Chromosome Location
Chr.9: 92297358-92325594 on Build GRCh38
Transcripts
NM_001256394.1 NM_017948.5 NR_046106.1 XM_006717166.3 XM_006717167.3 XM_006717168.3 XM_006717169.3 XM_006717170.3 XM_006717172.3 XM_006717173.3 XM_011518824.2 XM_011518827.2 XM_017014876.1 XM_017014877.1 XM_017014878.1 XM_017014879.1 XM_017014880.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1217570 c.3429G>T p.E1143D Chr.9: 92298281-92298281 on Build GRCh38
1217569 c.3429G>T p.E1143D Chr.9: 92298281-92298281 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758336703 Chr.9: 92298276-92298276 on Build GRCh38
rs551969244 Chr.9: 92298342-92298342 on Build GRCh38
rs771676320 Chr.9: 92298261-92298261 on Build GRCh38
rs3831004 Chr.9: 92298212-92298213 on Build GRCh38
rs889279626 Chr.9: 92298525-92298525 on Build GRCh38
rs927123224 Chr.9: 92298265-92298265 on Build GRCh38
rs887298113 Chr.9: 92298660-92298660 on Build GRCh38
rs192937852 Chr.9: 92298628-92298628 on Build GRCh38
rs145927690 Chr.9: 92298503-92298505 on Build GRCh38
rs59813350 Chr.9: 92298213-92298214 on Build GRCh38
rs188560839 Chr.9: 92298208-92298208 on Build GRCh38
rs755634812 Chr.9: 92298272-92298272 on Build GRCh38
rs759588984 Chr.9: 92298262-92298262 on Build GRCh38
rs143594230 Chr.9: 92298141-92298141 on Build GRCh38
rs777197319 Chr.9: 92298280-92298280 on Build GRCh38
rs767088482 Chr.9: 92298676-92298676 on Build GRCh38
rs1032826605 Chr.9: 92298654-92298654 on Build GRCh38
rs780531151 Chr.9: 92298341-92298341 on Build GRCh38
rs1001359878 Chr.9: 92298138-92298138 on Build GRCh38
rs915794144 Chr.9: 92298530-92298530 on Build GRCh38
rs147202001 Chr.9: 92298165-92298165 on Build GRCh38
rs1045363809 Chr.9: 92298620-92298620 on Build GRCh38
rs771332748 Chr.9: 92298358-92298358 on Build GRCh38
rs542946047 Chr.9: 92298178-92298178 on Build GRCh38
rs948439913 Chr.9: 92298592-92298592 on Build GRCh38
rs957402526 Chr.9: 92298476-92298476 on Build GRCh38
rs370348135 Chr.9: 92298274-92298274 on Build GRCh38
rs1017577201 Chr.9: 92298410-92298410 on Build GRCh38
rs924446099 Chr.9: 92298473-92298473 on Build GRCh38
rs35882484 Chr.9: 92298606-92298607 on Build GRCh38
rs906946103 Chr.9: 92298238-92298238 on Build GRCh38
rs879404138 Chr.9: 92298478-92298478 on Build GRCh38
rs778406496 Chr.9: 92298346-92298346 on Build GRCh38
rs1029782228 Chr.9: 92298450-92298450 on Build GRCh38
rs981208144 Chr.9: 92298247-92298247 on Build GRCh38
rs929062539 Chr.9: 92298649-92298649 on Build GRCh38
rs745603184 Chr.9: 92298645-92298645 on Build GRCh38
rs772341519 Chr.9: 92298385-92298385 on Build GRCh38
rs771134255 Chr.9: 92298222-92298222 on Build GRCh38
rs770953695 Chr.9: 92298421-92298421 on Build GRCh38
rs780827901 Chr.9: 92298132-92298132 on Build GRCh38
rs3736836 Chr.9: 92298348-92298348 on Build GRCh38
rs529593284 Chr.9: 92298197-92298197 on Build GRCh38
rs754702663 Chr.9: 92298312-92298312 on Build GRCh38
rs533316262 Chr.9: 92298340-92298340 on Build GRCh38
rs569468208 Chr.9: 92298211-92298211 on Build GRCh38
rs375582698 Chr.9: 92298375-92298375 on Build GRCh38
rs775668524 Chr.9: 92298270-92298270 on Build GRCh38
rs773388386 Chr.9: 92298209-92298209 on Build GRCh38
rs1032466129 Chr.9: 92298140-92298140 on Build GRCh38
rs751309784 Chr.9: 92298300-92298300 on Build GRCh38
rs964096474 Chr.9: 92298422-92298422 on Build GRCh38
rs977464911 Chr.9: 92298464-92298464 on Build GRCh38
rs867569295 Chr.9: 92298148-92298148 on Build GRCh38
rs908993021 Chr.9: 92298277-92298277 on Build GRCh38
rs765236671 Chr.9: 92298273-92298273 on Build GRCh38
rs1047219216 Chr.9: 92298301-92298301 on Build GRCh38
rs397969826 Chr.9: 92298216-92298217 on Build GRCh38
rs139611474 Chr.9: 92298463-92298463 on Build GRCh38
rs779016217 Chr.9: 92298429-92298429 on Build GRCh38
rs774720824 Chr.9: 92298362-92298362 on Build GRCh38
rs377211578 Chr.9: 92298263-92298263 on Build GRCh38
rs1047436958 Chr.9: 92298658-92298658 on Build GRCh38
rs534747421 Chr.9: 92298460-92298460 on Build GRCh38

View Full Product Details