Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 12867193-12867784 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TLR7
Entrez Gene ID
51284
Gene Name
toll like receptor 7
Gene Aliases
TLR7-like
UniGene
Hs.659215
Gene Chromosome Location
Chr.X: 12867083-12890361 on Build GRCh38
Transcripts
NM_016562.3

SNP Detail

SNP ID Chromosome Location
rs755964027 Chr.X: 12867420-12867420 on Build GRCh38
rs1008378964 Chr.X: 12867278-12867278 on Build GRCh38
rs200696034 Chr.X: 12867564-12867564 on Build GRCh38
rs2302267 Chr.X: 12867459-12867459 on Build GRCh38
rs750831973 Chr.X: 12867623-12867623 on Build GRCh38
rs1039641211 Chr.X: 12867415-12867415 on Build GRCh38
rs957905061 Chr.X: 12867231-12867231 on Build GRCh38
rs370277798 Chr.X: 12867702-12867702 on Build GRCh38
rs745983260 Chr.X: 12867508-12867508 on Build GRCh38
rs200095522 Chr.X: 12867618-12867618 on Build GRCh38
rs754668379 Chr.X: 12867483-12867483 on Build GRCh38
rs780631911 Chr.X: 12867610-12867610 on Build GRCh38
rs753362405 Chr.X: 12867467-12867467 on Build GRCh38
rs1048628092 Chr.X: 12867460-12867460 on Build GRCh38
rs112371861 Chr.X: 12867239-12867239 on Build GRCh38
rs1040841862 Chr.X: 12867279-12867279 on Build GRCh38
rs768852040 Chr.X: 12867554-12867554 on Build GRCh38
rs146631851 Chr.X: 12867750-12867750 on Build GRCh38
rs1001960719 Chr.X: 12867573-12867573 on Build GRCh38
rs201834636 Chr.X: 12867571-12867571 on Build GRCh38
rs957686997 Chr.X: 12867620-12867620 on Build GRCh38
rs913860038 Chr.X: 12867304-12867304 on Build GRCh38
rs887298204 Chr.X: 12867223-12867223 on Build GRCh38
rs187864051 Chr.X: 12867687-12867687 on Build GRCh38
rs190715862 Chr.X: 12867339-12867339 on Build GRCh38
rs767534068 Chr.X: 12867214-12867214 on Build GRCh38
rs758301366 Chr.X: 12867635-12867635 on Build GRCh38
rs377006756 Chr.X: 12867481-12867481 on Build GRCh38
rs765562515 Chr.X: 12867621-12867621 on Build GRCh38
rs183372465 Chr.X: 12867362-12867362 on Build GRCh38
rs5743715 Chr.X: 12867233-12867233 on Build GRCh38
rs5743716 Chr.X: 12867317-12867317 on Build GRCh38
rs899572283 Chr.X: 12867286-12867286 on Build GRCh38
rs768210650 Chr.X: 12867331-12867331 on Build GRCh38
rs777361001 Chr.X: 12867443-12867443 on Build GRCh38
rs1021292235 Chr.X: 12867378-12867378 on Build GRCh38
rs186242091 Chr.X: 12867305-12867305 on Build GRCh38
rs769232918 Chr.X: 12867548-12867549 on Build GRCh38
rs968753046 Chr.X: 12867384-12867384 on Build GRCh38
rs201939170 Chr.X: 12867468-12867468 on Build GRCh38
rs199865866 Chr.X: 12867531-12867531 on Build GRCh38
rs867356294 Chr.X: 12867232-12867232 on Build GRCh38
rs760840914 Chr.X: 12867534-12867534 on Build GRCh38
rs779271246 Chr.X: 12867476-12867476 on Build GRCh38

View Full Product Details