Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 70197730-70198204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ODAM
Entrez Gene ID
54959
Gene Name
odontogenic, ameloblast asssociated
Gene Aliases
APIN
UniGene
Hs.143811
Gene Chromosome Location
Chr.4: 70195728-70204576 on Build GRCh38
Transcripts
NM_017855.3 XM_005265698.3 XM_006714245.3 XM_017008331.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6011496 c.164C>T p.P55L Chr.4: 70197946-70197946 on Build GRCh38
6023129 c.163C>G p.P55A Chr.4: 70197945-70197945 on Build GRCh38
1259838 c.156A>C p.S52S Chr.4: 70197938-70197938 on Build GRCh38
3775953 c.227C>G p.S76C Chr.4: 70198009-70198009 on Build GRCh38
3783822 c.270G>T p.Q90H Chr.4: 70198052-70198052 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201051562 Chr.4: 70197980-70197980 on Build GRCh38
rs368917712 Chr.4: 70197912-70197912 on Build GRCh38
rs368391960 Chr.4: 70198130-70198130 on Build GRCh38
rs926857191 Chr.4: 70197994-70197994 on Build GRCh38
rs757913560 Chr.4: 70198032-70198032 on Build GRCh38
rs376525673 Chr.4: 70198036-70198036 on Build GRCh38
rs776688725 Chr.4: 70197947-70197947 on Build GRCh38
rs568425459 Chr.4: 70197792-70197792 on Build GRCh38
rs767529071 Chr.4: 70198077-70198077 on Build GRCh38
rs535956397 Chr.4: 70197812-70197812 on Build GRCh38
rs760807507 Chr.4: 70198081-70198081 on Build GRCh38
rs372172515 Chr.4: 70198127-70198128 on Build GRCh38
rs757196200 Chr.4: 70198031-70198031 on Build GRCh38
rs750580373 Chr.4: 70198003-70198003 on Build GRCh38
rs980308293 Chr.4: 70198079-70198079 on Build GRCh38
rs770822433 Chr.4: 70198040-70198040 on Build GRCh38
rs760548673 Chr.4: 70197934-70197934 on Build GRCh38
rs139440397 Chr.4: 70198125-70198126 on Build GRCh38
rs144151978 Chr.4: 70198082-70198082 on Build GRCh38
rs756433683 Chr.4: 70197895-70197895 on Build GRCh38
rs761009925 Chr.4: 70198155-70198155 on Build GRCh38
rs756518642 Chr.4: 70198004-70198004 on Build GRCh38
rs376631325 Chr.4: 70197916-70197916 on Build GRCh38
rs1019560574 Chr.4: 70198009-70198009 on Build GRCh38
rs558310870 Chr.4: 70197890-70197890 on Build GRCh38
rs772957913 Chr.4: 70197923-70197923 on Build GRCh38
rs139946153 Chr.4: 70198157-70198157 on Build GRCh38
rs753841147 Chr.4: 70197814-70197814 on Build GRCh38
rs1049588129 Chr.4: 70198116-70198116 on Build GRCh38
rs374327281 Chr.4: 70198083-70198083 on Build GRCh38
rs200796815 Chr.4: 70197914-70197914 on Build GRCh38
rs371671692 Chr.4: 70198150-70198150 on Build GRCh38
rs773481430 Chr.4: 70198078-70198078 on Build GRCh38
rs754435821 Chr.4: 70198017-70198017 on Build GRCh38
rs751871150 Chr.4: 70197874-70197874 on Build GRCh38
rs187139206 Chr.4: 70197830-70197830 on Build GRCh38
rs371346159 Chr.4: 70198086-70198086 on Build GRCh38
rs775365143 Chr.4: 70198068-70198068 on Build GRCh38
rs937631772 Chr.4: 70197771-70197771 on Build GRCh38
rs755391402 Chr.4: 70198158-70198159 on Build GRCh38
rs370769697 Chr.4: 70197984-70197984 on Build GRCh38
rs376097083 Chr.4: 70198110-70198110 on Build GRCh38
rs777413127 Chr.4: 70198099-70198099 on Build GRCh38
rs749399644 Chr.4: 70198141-70198141 on Build GRCh38
rs779356815 Chr.4: 70197871-70197871 on Build GRCh38
rs755410154 Chr.4: 70198018-70198018 on Build GRCh38
rs770572842 Chr.4: 70197935-70197935 on Build GRCh38
rs780846600 Chr.4: 70198115-70198115 on Build GRCh38
rs926367588 Chr.4: 70197753-70197753 on Build GRCh38
rs745679794 Chr.4: 70198128-70198128 on Build GRCh38
rs180873603 Chr.4: 70198111-70198111 on Build GRCh38
rs77942053 Chr.4: 70198122-70198122 on Build GRCh38
rs749211746 Chr.4: 70198147-70198147 on Build GRCh38
rs998053718 Chr.4: 70197782-70197782 on Build GRCh38
rs1009696729 Chr.4: 70197762-70197762 on Build GRCh38
rs189965058 Chr.4: 70197907-70197907 on Build GRCh38
rs759383682 Chr.4: 70197948-70197948 on Build GRCh38
rs765841821 Chr.4: 70198085-70198085 on Build GRCh38
rs199550783 Chr.4: 70198056-70198056 on Build GRCh38
rs963421130 Chr.4: 70197913-70197913 on Build GRCh38
rs566295310 Chr.4: 70197835-70197835 on Build GRCh38
rs1027239214 Chr.4: 70198052-70198052 on Build GRCh38
rs767760206 Chr.4: 70197995-70197995 on Build GRCh38
rs893478240 Chr.4: 70197757-70197757 on Build GRCh38
rs182260840 Chr.4: 70197781-70197781 on Build GRCh38
rs776875313 Chr.4: 70198156-70198156 on Build GRCh38
rs953997888 Chr.4: 70197797-70197797 on Build GRCh38
rs778535331 Chr.4: 70198024-70198024 on Build GRCh38
rs1042151259 Chr.4: 70197767-70197767 on Build GRCh38
rs764271002 Chr.4: 70197978-70197978 on Build GRCh38
rs753320098 Chr.4: 70198158-70198158 on Build GRCh38
rs897196914 Chr.4: 70198013-70198013 on Build GRCh38
rs201514554 Chr.4: 70198084-70198084 on Build GRCh38
rs758036498 Chr.4: 70198097-70198097 on Build GRCh38
rs766644492 Chr.4: 70198005-70198005 on Build GRCh38
rs879078141 Chr.4: 70197807-70197807 on Build GRCh38
rs539682341 Chr.4: 70197887-70197887 on Build GRCh38
rs781582342 Chr.4: 70197888-70197888 on Build GRCh38
rs761888136 Chr.4: 70198071-70198071 on Build GRCh38
rs780619310 Chr.4: 70197899-70197899 on Build GRCh38
rs773249663 Chr.4: 70198154-70198154 on Build GRCh38
rs1050246002 Chr.4: 70197960-70197960 on Build GRCh38
rs747844033 Chr.4: 70197915-70197915 on Build GRCh38
rs373877978 Chr.4: 70198044-70198044 on Build GRCh38
rs919370777 Chr.4: 70197939-70197939 on Build GRCh38

View Full Product Details