Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 156772252-156772851 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDGFC
Entrez Gene ID
56034
Gene Name
platelet derived growth factor C
Gene Aliases
FALLOTEIN, SCDGF
UniGene
Hs.570855
Gene Chromosome Location
Chr.4: 156761611-156971394 on Build GRCh38
Transcripts
NM_016205.2 NR_036641.1 XM_011532124.2 XM_017008455.1 XM_017008456.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5925131 c.667C>T p.L223F Chr.4: 156772722-156772722 on Build GRCh38
6743117 c.665T>C p.L222P Chr.4: 156772724-156772724 on Build GRCh38
3428281 c.698C>A p.S233Y Chr.4: 156772691-156772691 on Build GRCh38
6166710 c.703G>T p.V235L Chr.4: 156772686-156772686 on Build GRCh38
733346 c.601C>G p.R201G Chr.4: 156772788-156772788 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs974422705 Chr.4: 156772539-156772539 on Build GRCh38
rs755511812 Chr.4: 156772786-156772786 on Build GRCh38
rs777922904 Chr.4: 156772818-156772818 on Build GRCh38
rs867522358 Chr.4: 156772427-156772427 on Build GRCh38
rs375215057 Chr.4: 156772719-156772719 on Build GRCh38
rs921680190 Chr.4: 156772602-156772602 on Build GRCh38
rs184134579 Chr.4: 156772418-156772418 on Build GRCh38
rs1019082307 Chr.4: 156772470-156772470 on Build GRCh38
rs765903830 Chr.4: 156772764-156772764 on Build GRCh38
rs966188337 Chr.4: 156772475-156772475 on Build GRCh38
rs1054567794 Chr.4: 156772323-156772323 on Build GRCh38
rs907049462 Chr.4: 156772372-156772372 on Build GRCh38
rs368455130 Chr.4: 156772647-156772647 on Build GRCh38
rs749956577 Chr.4: 156772762-156772762 on Build GRCh38
rs762699370 Chr.4: 156772413-156772413 on Build GRCh38
rs770103503 Chr.4: 156772657-156772657 on Build GRCh38
rs375990881 Chr.4: 156772787-156772787 on Build GRCh38
rs1049062190 Chr.4: 156772365-156772365 on Build GRCh38
rs950985993 Chr.4: 156772447-156772447 on Build GRCh38
rs907608501 Chr.4: 156772731-156772731 on Build GRCh38
rs749512195 Chr.4: 156772829-156772829 on Build GRCh38
rs1041301275 Chr.4: 156772401-156772401 on Build GRCh38
rs373950792 Chr.4: 156772767-156772767 on Build GRCh38
rs763913754 Chr.4: 156772729-156772729 on Build GRCh38
rs895227114 Chr.4: 156772751-156772751 on Build GRCh38
rs773016587 Chr.4: 156772345-156772345 on Build GRCh38
rs748422838 Chr.4: 156772656-156772656 on Build GRCh38
rs796900513 Chr.4: 156772732-156772732 on Build GRCh38
rs1010975899 Chr.4: 156772463-156772463 on Build GRCh38
rs927872536 Chr.4: 156772574-156772574 on Build GRCh38
rs781674510 Chr.4: 156772651-156772651 on Build GRCh38
rs1041162351 Chr.4: 156772788-156772788 on Build GRCh38
rs369762309 Chr.4: 156772736-156772736 on Build GRCh38
rs764741287 Chr.4: 156772752-156772752 on Build GRCh38
rs935670541 Chr.4: 156772312-156772312 on Build GRCh38
rs974475035 Chr.4: 156772294-156772294 on Build GRCh38
rs1055669066 Chr.4: 156772717-156772717 on Build GRCh38
rs756407673 Chr.4: 156772807-156772807 on Build GRCh38
rs997262394 Chr.4: 156772445-156772445 on Build GRCh38
rs1003600947 Chr.4: 156772379-156772379 on Build GRCh38
rs989814419 Chr.4: 156772613-156772613 on Build GRCh38
rs1033773173 Chr.4: 156772390-156772390 on Build GRCh38
rs761474940 Chr.4: 156772744-156772744 on Build GRCh38
rs773353932 Chr.4: 156772673-156772673 on Build GRCh38
rs945983087 Chr.4: 156772356-156772356 on Build GRCh38
rs767768346 Chr.4: 156772486-156772486 on Build GRCh38
rs984135103 Chr.4: 156772449-156772449 on Build GRCh38
rs981901342 Chr.4: 156772524-156772526 on Build GRCh38
rs1045551516 Chr.4: 156772357-156772357 on Build GRCh38
rs891703422 Chr.4: 156772348-156772348 on Build GRCh38
rs921565825 Chr.4: 156772299-156772299 on Build GRCh38
rs1028106462 Chr.4: 156772442-156772442 on Build GRCh38
rs199562156 Chr.4: 156772708-156772708 on Build GRCh38
rs994838175 Chr.4: 156772416-156772416 on Build GRCh38
rs1049043716 Chr.4: 156772792-156772792 on Build GRCh38
rs753063495 Chr.4: 156772798-156772798 on Build GRCh38
rs912919039 Chr.4: 156772632-156772632 on Build GRCh38
rs966548492 Chr.4: 156772489-156772489 on Build GRCh38
rs1004307519 Chr.4: 156772383-156772383 on Build GRCh38
rs935793133 Chr.4: 156772612-156772612 on Build GRCh38
rs920370362 Chr.4: 156772316-156772316 on Build GRCh38
rs749258476 Chr.4: 156772674-156772674 on Build GRCh38
rs371889486 Chr.4: 156772703-156772703 on Build GRCh38
rs945956243 Chr.4: 156772636-156772636 on Build GRCh38

View Full Product Details