Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 117521452-117522033 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCBLD1
Entrez Gene ID
285761
Gene Name
discoidin, CUB and LCCL domain containing 1
Gene Aliases
dJ94G16.1
UniGene
Hs.658304
Gene Chromosome Location
Chr.6: 117480346-117569858 on Build GRCh38
Transcripts
NM_173674.2 XM_005266941.1 XM_006715463.1 XM_011535771.1 XM_011535773.2 XM_011535774.2 XM_017010775.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
310483 c.462T>G p.D154E Chr.6: 117521526-117521526 on Build GRCh38
6640612 c.470C>T p.T157I Chr.6: 117521534-117521534 on Build GRCh38
122510 c.470C>A p.T157K Chr.6: 117521534-117521534 on Build GRCh38
3857825 c.483A>C p.R161R Chr.6: 117521547-117521547 on Build GRCh38
3157994 c.512G>A p.S171N Chr.6: 117521576-117521576 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759267010 Chr.6: 117521549-117521549 on Build GRCh38
rs753870741 Chr.6: 117521585-117521585 on Build GRCh38
rs372734932 Chr.6: 117521646-117521646 on Build GRCh38
rs778559072 Chr.6: 117521495-117521495 on Build GRCh38
rs770824764 Chr.6: 117521494-117521494 on Build GRCh38
rs910574349 Chr.6: 117521850-117521850 on Build GRCh38
rs769967776 Chr.6: 117521518-117521518 on Build GRCh38
rs557629224 Chr.6: 117522001-117522002 on Build GRCh38
rs189474342 Chr.6: 117521925-117521925 on Build GRCh38
rs201985351 Chr.6: 117521577-117521577 on Build GRCh38
rs747267001 Chr.6: 117521502-117521502 on Build GRCh38
rs367665212 Chr.6: 117521519-117521519 on Build GRCh38
rs948052081 Chr.6: 117521638-117521638 on Build GRCh38
rs755662504 Chr.6: 117521581-117521581 on Build GRCh38
rs755270015 Chr.6: 117521608-117521608 on Build GRCh38
rs761932795 Chr.6: 117521516-117521516 on Build GRCh38
rs116434032 Chr.6: 117521885-117521885 on Build GRCh38
rs968800218 Chr.6: 117521513-117521513 on Build GRCh38
rs986091471 Chr.6: 117521826-117521826 on Build GRCh38
rs765293087 Chr.6: 117521924-117521924 on Build GRCh38
rs961880176 Chr.6: 117521982-117521982 on Build GRCh38
rs996258464 Chr.6: 117521841-117521841 on Build GRCh38
rs904114939 Chr.6: 117521629-117521629 on Build GRCh38
rs879141330 Chr.6: 117521654-117521654 on Build GRCh38
rs9689349 Chr.6: 117521737-117521737 on Build GRCh38
rs772606276 Chr.6: 117521485-117521486 on Build GRCh38
rs999857602 Chr.6: 117521639-117521639 on Build GRCh38
rs537352059 Chr.6: 117521688-117521688 on Build GRCh38
rs755209643 Chr.6: 117521853-117521853 on Build GRCh38
rs778772134 Chr.6: 117521598-117521598 on Build GRCh38
rs944846782 Chr.6: 117521555-117521555 on Build GRCh38
rs777742377 Chr.6: 117521990-117521990 on Build GRCh38
rs568673608 Chr.6: 117521758-117521758 on Build GRCh38
rs1022508512 Chr.6: 117521781-117521781 on Build GRCh38
rs1011081575 Chr.6: 117521760-117521760 on Build GRCh38
rs954541287 Chr.6: 117521542-117521542 on Build GRCh38
rs9285421 Chr.6: 117521535-117521535 on Build GRCh38
rs958682031 Chr.6: 117521683-117521683 on Build GRCh38
rs907700043 Chr.6: 117521969-117521969 on Build GRCh38
rs767109967 Chr.6: 117521558-117521558 on Build GRCh38
rs866681769 Chr.6: 117521572-117521572 on Build GRCh38
rs529104007 Chr.6: 117521697-117521697 on Build GRCh38
rs894113058 Chr.6: 117521991-117521991 on Build GRCh38
rs776878969 Chr.6: 117521514-117521514 on Build GRCh38
rs555363217 Chr.6: 117521923-117521923 on Build GRCh38
rs369622033 Chr.6: 117521862-117521862 on Build GRCh38
rs945495144 Chr.6: 117521945-117521945 on Build GRCh38
rs944771751 Chr.6: 117521882-117521882 on Build GRCh38
rs777737799 Chr.6: 117521476-117521476 on Build GRCh38
rs766121099 Chr.6: 117521714-117521714 on Build GRCh38
rs1014356446 Chr.6: 117522004-117522005 on Build GRCh38
rs546667191 Chr.6: 117521631-117521631 on Build GRCh38
rs371712539 Chr.6: 117521583-117521583 on Build GRCh38
rs748345404 Chr.6: 117521623-117521623 on Build GRCh38
rs374941271 Chr.6: 117521590-117521590 on Build GRCh38
rs561755413 Chr.6: 117521653-117521653 on Build GRCh38
rs1043953458 Chr.6: 117521957-117521957 on Build GRCh38
rs551583984 Chr.6: 117521790-117521790 on Build GRCh38
rs879783517 Chr.6: 117521970-117521970 on Build GRCh38
rs763359924 Chr.6: 117521530-117521530 on Build GRCh38
rs184790040 Chr.6: 117521545-117521545 on Build GRCh38
rs68145844 Chr.6: 117521600-117521601 on Build GRCh38
rs1024019550 Chr.6: 117522005-117522005 on Build GRCh38
rs749150923 Chr.6: 117521487-117521487 on Build GRCh38
rs968391811 Chr.6: 117521794-117521794 on Build GRCh38
rs745608465 Chr.6: 117521603-117521603 on Build GRCh38
rs1037655357 Chr.6: 117521600-117521600 on Build GRCh38
rs9689345 Chr.6: 117521609-117521609 on Build GRCh38
rs72963484 Chr.6: 117521506-117521506 on Build GRCh38
rs539243578 Chr.6: 117521778-117521778 on Build GRCh38
rs35394851 Chr.6: 117521597-117521598 on Build GRCh38
rs397888035 Chr.6: 117521599-117521600 on Build GRCh38
rs987259153 Chr.6: 117521943-117521943 on Build GRCh38
rs772136862 Chr.6: 117521919-117521919 on Build GRCh38

View Full Product Details