Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 32629974-32630567 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAF1L
Entrez Gene ID
138474
Gene Name
TATA-box binding protein associated factor 1 like
Gene Aliases
TAF(II)210, TAF2A2
UniGene
Hs.591086
Gene Chromosome Location
Chr.9: 32629454-32635669 on Build GRCh38
Transcripts
NM_153809.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
326625 c.5047G>T p.V1683L Chr.9: 32630533-32630533 on Build GRCh38
393853 c.5195C>A p.P1732H Chr.9: 32630385-32630385 on Build GRCh38
5487298 c.5407G>A p.G1803S Chr.9: 32630173-32630173 on Build GRCh38
6530602 c.5461A>G p.K1821E Chr.9: 32630119-32630119 on Build GRCh38
5616120 c.5341G>A p.E1781K Chr.9: 32630239-32630239 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766459966 Chr.9: 32630094-32630095 on Build GRCh38
rs753724498 Chr.9: 32630243-32630243 on Build GRCh38
rs754257933 Chr.9: 32630128-32630137 on Build GRCh38
rs776575588 Chr.9: 32630491-32630491 on Build GRCh38
rs753541946 Chr.9: 32630346-32630346 on Build GRCh38
rs761423467 Chr.9: 32630118-32630118 on Build GRCh38
rs750348291 Chr.9: 32630240-32630240 on Build GRCh38
rs748993855 Chr.9: 32630069-32630069 on Build GRCh38
rs748824872 Chr.9: 32630146-32630146 on Build GRCh38
rs773418235 Chr.9: 32630402-32630402 on Build GRCh38
rs780660367 Chr.9: 32630380-32630380 on Build GRCh38
rs767636067 Chr.9: 32630326-32630326 on Build GRCh38
rs771000652 Chr.9: 32630481-32630481 on Build GRCh38
rs745637776 Chr.9: 32630123-32630123 on Build GRCh38
rs763416441 Chr.9: 32630302-32630302 on Build GRCh38
rs370095170 Chr.9: 32630212-32630212 on Build GRCh38
rs753857467 Chr.9: 32630438-32630438 on Build GRCh38
rs778663929 Chr.9: 32630253-32630253 on Build GRCh38
rs746592555 Chr.9: 32630365-32630365 on Build GRCh38
rs779405153 Chr.9: 32630236-32630238 on Build GRCh38
rs1039296117 Chr.9: 32630059-32630059 on Build GRCh38
rs758042199 Chr.9: 32630424-32630424 on Build GRCh38
rs771518750 Chr.9: 32630270-32630270 on Build GRCh38
rs967727744 Chr.9: 32630028-32630028 on Build GRCh38
rs756746480 Chr.9: 32630448-32630448 on Build GRCh38
rs755926328 Chr.9: 32630130-32630130 on Build GRCh38
rs763776889 Chr.9: 32630426-32630426 on Build GRCh38
rs760877567 Chr.9: 32630403-32630403 on Build GRCh38
rs912081240 Chr.9: 32629998-32629998 on Build GRCh38
rs773721333 Chr.9: 32630161-32630161 on Build GRCh38
rs755451203 Chr.9: 32630458-32630458 on Build GRCh38
rs748586498 Chr.9: 32630469-32630469 on Build GRCh38
rs779654443 Chr.9: 32630242-32630242 on Build GRCh38
rs777168319 Chr.9: 32630172-32630172 on Build GRCh38
rs750542623 Chr.9: 32630332-32630332 on Build GRCh38
rs765026411 Chr.9: 32630408-32630408 on Build GRCh38
rs752304382 Chr.9: 32630354-32630354 on Build GRCh38
rs199701622 Chr.9: 32630064-32630064 on Build GRCh38
rs768739252 Chr.9: 32630308-32630308 on Build GRCh38
rs369679263 Chr.9: 32630070-32630070 on Build GRCh38
rs145944536 Chr.9: 32630356-32630356 on Build GRCh38
rs762034779 Chr.9: 32630231-32630231 on Build GRCh38
rs777255832 Chr.9: 32630359-32630359 on Build GRCh38
rs933786939 Chr.9: 32630322-32630322 on Build GRCh38
rs1033421059 Chr.9: 32630083-32630083 on Build GRCh38
rs759931935 Chr.9: 32630092-32630092 on Build GRCh38
rs748640345 Chr.9: 32630394-32630396 on Build GRCh38
rs762159609 Chr.9: 32630312-32630312 on Build GRCh38
rs760675323 Chr.9: 32630337-32630337 on Build GRCh38
rs764448670 Chr.9: 32630226-32630226 on Build GRCh38
rs779404415 Chr.9: 32630468-32630468 on Build GRCh38
rs56342342 Chr.9: 32630151-32630151 on Build GRCh38
rs759420779 Chr.9: 32630407-32630407 on Build GRCh38
rs903517850 Chr.9: 32630416-32630416 on Build GRCh38
rs775940138 Chr.9: 32630205-32630205 on Build GRCh38
rs992975562 Chr.9: 32630091-32630091 on Build GRCh38
rs765420032 Chr.9: 32630096-32630096 on Build GRCh38
rs754848062 Chr.9: 32630347-32630347 on Build GRCh38
rs202147126 Chr.9: 32630108-32630108 on Build GRCh38
rs761028969 Chr.9: 32630079-32630079 on Build GRCh38
rs966743447 Chr.9: 32630536-32630536 on Build GRCh38
rs34241003 Chr.9: 32630387-32630387 on Build GRCh38
rs760268151 Chr.9: 32630379-32630379 on Build GRCh38
rs749740046 Chr.9: 32630457-32630457 on Build GRCh38
rs944831600 Chr.9: 32630036-32630036 on Build GRCh38
rs760056638 Chr.9: 32630179-32630179 on Build GRCh38
rs546273222 Chr.9: 32630527-32630527 on Build GRCh38
rs780602390 Chr.9: 32630453-32630453 on Build GRCh38
rs150771265 Chr.9: 32630523-32630523 on Build GRCh38
rs56007745 Chr.9: 32630186-32630186 on Build GRCh38
rs61736667 Chr.9: 32630284-32630284 on Build GRCh38
rs757970258 Chr.9: 32630402-32630403 on Build GRCh38
rs752547797 Chr.9: 32630410-32630410 on Build GRCh38
rs772388404 Chr.9: 32630471-32630471 on Build GRCh38
rs747586254 Chr.9: 32630261-32630261 on Build GRCh38
rs757138027 Chr.9: 32630117-32630117 on Build GRCh38
rs769027184 Chr.9: 32630385-32630385 on Build GRCh38
rs755228438 Chr.9: 32630191-32630196 on Build GRCh38
rs111917220 Chr.9: 32630405-32630405 on Build GRCh38
rs759506052 Chr.9: 32630515-32630515 on Build GRCh38
rs767729888 Chr.9: 32630233-32630233 on Build GRCh38
rs751723903 Chr.9: 32630114-32630114 on Build GRCh38
rs775717901 Chr.9: 32630300-32630300 on Build GRCh38
rs575169099 Chr.9: 32630351-32630351 on Build GRCh38
rs147562191 Chr.9: 32630229-32630229 on Build GRCh38
rs558316452 Chr.9: 32630341-32630341 on Build GRCh38
rs568675126 Chr.9: 32630112-32630112 on Build GRCh38
rs562781976 Chr.9: 32630544-32630544 on Build GRCh38
rs754577609 Chr.9: 32630249-32630249 on Build GRCh38
rs540004697 Chr.9: 32630436-32630436 on Build GRCh38
rs777233672 Chr.9: 32630470-32630471 on Build GRCh38
rs772477235 Chr.9: 32630401-32630401 on Build GRCh38
rs780023391 Chr.9: 32630136-32630136 on Build GRCh38
rs16918393 Chr.9: 32630167-32630167 on Build GRCh38
rs770174132 Chr.9: 32630373-32630373 on Build GRCh38
rs773916138 Chr.9: 32630071-32630071 on Build GRCh38
rs10758145 Chr.9: 32630474-32630474 on Build GRCh38
rs267602207 Chr.9: 32630391-32630391 on Build GRCh38
rs1055664019 Chr.9: 32630396-32630396 on Build GRCh38
rs142325458 Chr.9: 32630311-32630311 on Build GRCh38
rs377232652 Chr.9: 32630107-32630107 on Build GRCh38
rs150232564 Chr.9: 32630166-32630166 on Build GRCh38
rs975355208 Chr.9: 32630236-32630236 on Build GRCh38
rs746318773 Chr.9: 32630294-32630294 on Build GRCh38
rs540244683 Chr.9: 32630160-32630160 on Build GRCh38

View Full Product Details